Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 140542491-140543001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMOD2
Entrez Gene ID
255520
Gene Name
ELMO domain containing 2
Gene Aliases
9830169G11Rik
UniGene
Hs.450105
Gene Chromosome Location
Chr.4: 140524146-140553770 on Build GRCh38
Transcripts
NM_153702.3 XM_005262885.3 XM_005262886.2 XM_011531818.2 XM_011531819.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6422843 c.569A>T p.Q190L Chr.4: 140542609-140542609 on Build GRCh38
4680803 c.544G>T p.E182* Chr.4: 140542584-140542584 on Build GRCh38
6430068 c.563C>A p.A188D Chr.4: 140542603-140542603 on Build GRCh38
4773009 c.580C>A p.R194S Chr.4: 140542620-140542620 on Build GRCh38
7115166 c.534-1G>A p.? Chr.4: 140542573-140542573 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778876519 Chr.4: 140542548-140542548 on Build GRCh38
rs770658001 Chr.4: 140542567-140542567 on Build GRCh38
rs770098385 Chr.4: 140542595-140542595 on Build GRCh38
rs3104105 Chr.4: 140542955-140542955 on Build GRCh38
rs199973427 Chr.4: 140542559-140542559 on Build GRCh38
rs544670913 Chr.4: 140542853-140542853 on Build GRCh38
rs533169766 Chr.4: 140542863-140542863 on Build GRCh38
rs575335988 Chr.4: 140542537-140542537 on Build GRCh38
rs748555886 Chr.4: 140542593-140542593 on Build GRCh38
rs926002190 Chr.4: 140542893-140542893 on Build GRCh38
rs779232905 Chr.4: 140542868-140542868 on Build GRCh38
rs374151745 Chr.4: 140542658-140542658 on Build GRCh38
rs1023622357 Chr.4: 140542895-140542895 on Build GRCh38
rs775039262 Chr.4: 140542739-140542739 on Build GRCh38
rs750369242 Chr.4: 140542662-140542662 on Build GRCh38
rs1001115027 Chr.4: 140542928-140542928 on Build GRCh38
rs965603861 Chr.4: 140542850-140542850 on Build GRCh38
rs1037968323 Chr.4: 140542834-140542834 on Build GRCh38
rs540518760 Chr.4: 140542779-140542779 on Build GRCh38
rs559567069 Chr.4: 140542520-140542520 on Build GRCh38
rs967323191 Chr.4: 140542881-140542881 on Build GRCh38
rs937421695 Chr.4: 140542919-140542919 on Build GRCh38
rs775727065 Chr.4: 140542656-140542656 on Build GRCh38
rs768437575 Chr.4: 140542585-140542585 on Build GRCh38
rs566560203 Chr.4: 140542883-140542883 on Build GRCh38
rs980031714 Chr.4: 140542541-140542541 on Build GRCh38
rs768282187 Chr.4: 140542596-140542596 on Build GRCh38
rs774370188 Chr.4: 140542687-140542687 on Build GRCh38
rs370185972 Chr.4: 140542828-140542828 on Build GRCh38
rs113781254 Chr.4: 140542812-140542812 on Build GRCh38
rs781649998 Chr.4: 140542588-140542588 on Build GRCh38
rs746593479 Chr.4: 140542577-140542577 on Build GRCh38
rs756490301 Chr.4: 140542679-140542679 on Build GRCh38
rs910490765 Chr.4: 140542804-140542804 on Build GRCh38
rs969654973 Chr.4: 140542926-140542926 on Build GRCh38
rs745914321 Chr.4: 140542693-140542693 on Build GRCh38
rs1050023993 Chr.4: 140542778-140542778 on Build GRCh38
rs751770630 Chr.4: 140542523-140542523 on Build GRCh38
rs1027372700 Chr.4: 140542781-140542781 on Build GRCh38
rs377194309 Chr.4: 140542668-140542668 on Build GRCh38
rs747854290 Chr.4: 140542676-140542676 on Build GRCh38
rs1036250539 Chr.4: 140542977-140542977 on Build GRCh38
rs779496388 Chr.4: 140542576-140542576 on Build GRCh38
rs749740363 Chr.4: 140542684-140542684 on Build GRCh38
rs548335050 Chr.4: 140542846-140542846 on Build GRCh38
rs371769622 Chr.4: 140542527-140542527 on Build GRCh38
rs190121947 Chr.4: 140542538-140542538 on Build GRCh38
rs533313734 Chr.4: 140542667-140542667 on Build GRCh38
rs754639087 Chr.4: 140542670-140542670 on Build GRCh38
rs573532089 Chr.4: 140542767-140542767 on Build GRCh38
rs763815566 Chr.4: 140542529-140542529 on Build GRCh38
rs991624347 Chr.4: 140542936-140542936 on Build GRCh38
rs763431469 Chr.4: 140542621-140542621 on Build GRCh38
rs759701312 Chr.4: 140542652-140542652 on Build GRCh38
rs750375702 Chr.4: 140542558-140542558 on Build GRCh38
rs200726237 Chr.4: 140542733-140542737 on Build GRCh38
rs771043154 Chr.4: 140542650-140542650 on Build GRCh38
rs949931839 Chr.4: 140542956-140542956 on Build GRCh38
rs144150157 Chr.4: 140542606-140542606 on Build GRCh38
rs780963652 Chr.4: 140542675-140542675 on Build GRCh38
rs771359760 Chr.4: 140542686-140542686 on Build GRCh38
rs112703313 Chr.4: 140542859-140542859 on Build GRCh38

View Full Product Details