Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 164954659-164955192 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM61
Entrez Gene ID
391712
Gene Name
tripartite motif containing 61
Gene Aliases
RNF35
UniGene
Hs.654633
Gene Chromosome Location
Chr.4: 164954446-164977666 on Build GRCh38
Transcripts
NM_001012414.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4969509 c.589C>T p.R197C Chr.4: 164955033-164955033 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs899768445 Chr.4: 164954710-164954710 on Build GRCh38
rs376969248 Chr.4: 164954825-164954825 on Build GRCh38
rs934816839 Chr.4: 164955032-164955032 on Build GRCh38
rs940973973 Chr.4: 164955123-164955123 on Build GRCh38
rs769199597 Chr.4: 164954961-164954961 on Build GRCh38
rs1038099556 Chr.4: 164954720-164954720 on Build GRCh38
rs1047319832 Chr.4: 164955120-164955121 on Build GRCh38
rs947646064 Chr.4: 164955011-164955011 on Build GRCh38
rs1029494929 Chr.4: 164954752-164954752 on Build GRCh38
rs960099642 Chr.4: 164954876-164954876 on Build GRCh38
rs915891209 Chr.4: 164954900-164954900 on Build GRCh38
rs991432130 Chr.4: 164954895-164954895 on Build GRCh38
rs540495146 Chr.4: 164955050-164955050 on Build GRCh38
rs955419858 Chr.4: 164954768-164954768 on Build GRCh38
rs1020852685 Chr.4: 164954826-164954826 on Build GRCh38
rs995395250 Chr.4: 164954735-164954735 on Build GRCh38
rs926000425 Chr.4: 164955015-164955015 on Build GRCh38
rs899466925 Chr.4: 164955122-164955122 on Build GRCh38
rs1048340180 Chr.4: 164955145-164955145 on Build GRCh38
rs1039401906 Chr.4: 164955106-164955106 on Build GRCh38
rs369400329 Chr.4: 164955053-164955053 on Build GRCh38
rs768573538 Chr.4: 164955099-164955100 on Build GRCh38
rs995426129 Chr.4: 164955135-164955135 on Build GRCh38
rs917408753 Chr.4: 164955049-164955049 on Build GRCh38
rs1009474620 Chr.4: 164954777-164954777 on Build GRCh38
rs1026828834 Chr.4: 164954742-164954742 on Build GRCh38
rs748482634 Chr.4: 164955026-164955026 on Build GRCh38
rs773431567 Chr.4: 164955130-164955130 on Build GRCh38
rs776515678 Chr.4: 164955082-164955084 on Build GRCh38
rs908808731 Chr.4: 164955120-164955120 on Build GRCh38
rs80146096 Chr.4: 164955103-164955103 on Build GRCh38
rs1018226842 Chr.4: 164954687-164954687 on Build GRCh38
rs770168839 Chr.4: 164955101-164955101 on Build GRCh38
rs571824778 Chr.4: 164954751-164954751 on Build GRCh38
rs967321436 Chr.4: 164954951-164954951 on Build GRCh38
rs539365021 Chr.4: 164954835-164954835 on Build GRCh38
rs982926987 Chr.4: 164954796-164954796 on Build GRCh38
rs200797852 Chr.4: 164955119-164955120 on Build GRCh38
rs111408210 Chr.4: 164955033-164955033 on Build GRCh38
rs924840787 Chr.4: 164955025-164955025 on Build GRCh38
rs1008295820 Chr.4: 164954683-164954683 on Build GRCh38
rs1005241591 Chr.4: 164954698-164954698 on Build GRCh38
rs886753930 Chr.4: 164954697-164954697 on Build GRCh38
rs899593660 Chr.4: 164954725-164954725 on Build GRCh38
rs996602072 Chr.4: 164955165-164955165 on Build GRCh38
rs781700623 Chr.4: 164954979-164954979 on Build GRCh38
rs978943118 Chr.4: 164955016-164955016 on Build GRCh38

View Full Product Details