Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186196817-186197360 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4V2
Entrez Gene ID
285440
Gene Name
cytochrome P450 family 4 subfamily V member 2
Gene Aliases
BCD, CYP4AH1
UniGene
Hs.587231
Gene Chromosome Location
Chr.4: 186191520-186213463 on Build GRCh38
Transcripts
NM_207352.3 XM_005262935.3 XM_017008037.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6416077 c.546C>A p.N182K Chr.4: 186197072-186197072 on Build GRCh38
3917551 c.478G>A p.E160K Chr.4: 186197004-186197004 on Build GRCh38
1428957 c.565delT p.Y191fs*7 Chr.4: 186197091-186197091 on Build GRCh38
3730921 c.500A>T p.N167I Chr.4: 186197026-186197026 on Build GRCh38
6379016 c.554C>A p.A185E Chr.4: 186197080-186197080 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs957313060 Chr.4: 186196995-186196995 on Build GRCh38
rs1052037375 Chr.4: 186197259-186197259 on Build GRCh38
rs62350518 Chr.4: 186197204-186197204 on Build GRCh38
rs370950185 Chr.4: 186196949-186196949 on Build GRCh38
rs72646260 Chr.4: 186197205-186197205 on Build GRCh38
rs72646257 Chr.4: 186196847-186196847 on Build GRCh38
rs397722245 Chr.4: 186196900-186196901 on Build GRCh38
rs768759783 Chr.4: 186197176-186197176 on Build GRCh38
rs965425263 Chr.4: 186197017-186197017 on Build GRCh38
rs544363983 Chr.4: 186197300-186197300 on Build GRCh38
rs374764984 Chr.4: 186197166-186197166 on Build GRCh38
rs112367913 Chr.4: 186196887-186196888 on Build GRCh38
rs998537843 Chr.4: 186197276-186197276 on Build GRCh38
rs187249105 Chr.4: 186197215-186197215 on Build GRCh38
rs750786252 Chr.4: 186196963-186196963 on Build GRCh38
rs776588424 Chr.4: 186196892-186196892 on Build GRCh38
rs769104744 Chr.4: 186197272-186197274 on Build GRCh38
rs945246638 Chr.4: 186197077-186197077 on Build GRCh38
rs575374497 Chr.4: 186197264-186197264 on Build GRCh38
rs779627569 Chr.4: 186196981-186196981 on Build GRCh38
rs764426507 Chr.4: 186197060-186197060 on Build GRCh38
rs746453302 Chr.4: 186196890-186196890 on Build GRCh38
rs376204847 Chr.4: 186196933-186196933 on Build GRCh38
rs758719879 Chr.4: 186197080-186197080 on Build GRCh38
rs138297987 Chr.4: 186197068-186197068 on Build GRCh38
rs942107523 Chr.4: 186197226-186197226 on Build GRCh38
rs779534809 Chr.4: 186196957-186196957 on Build GRCh38
rs141572111 Chr.4: 186197224-186197224 on Build GRCh38
rs376981886 Chr.4: 186197145-186197145 on Build GRCh38
rs762907849 Chr.4: 186196938-186196938 on Build GRCh38
rs398088141 Chr.4: 186196898-186196899 on Build GRCh38
rs768393090 Chr.4: 186196990-186196990 on Build GRCh38
rs762776506 Chr.4: 186197020-186197020 on Build GRCh38
rs773206217 Chr.4: 186197002-186197002 on Build GRCh38
rs756204549 Chr.4: 186197217-186197217 on Build GRCh38
rs777802828 Chr.4: 186197140-186197140 on Build GRCh38
rs928525124 Chr.4: 186197172-186197172 on Build GRCh38
rs889941184 Chr.4: 186197217-186197217 on Build GRCh38
rs755661211 Chr.4: 186197137-186197137 on Build GRCh38
rs776006079 Chr.4: 186197067-186197068 on Build GRCh38
rs564886125 Chr.4: 186197251-186197251 on Build GRCh38
rs765599061 Chr.4: 186197075-186197075 on Build GRCh38
rs1031419655 Chr.4: 186196985-186196985 on Build GRCh38
rs201006021 Chr.4: 186197067-186197067 on Build GRCh38
rs752979612 Chr.4: 186196948-186196948 on Build GRCh38
rs749271531 Chr.4: 186197143-186197143 on Build GRCh38
rs1007041023 Chr.4: 186197217-186197219 on Build GRCh38
rs11932764 Chr.4: 186197081-186197081 on Build GRCh38
rs897369199 Chr.4: 186196885-186196886 on Build GRCh38
rs1039230003 Chr.4: 186197234-186197234 on Build GRCh38
rs761621320 Chr.4: 186196902-186196902 on Build GRCh38
rs780661245 Chr.4: 186197036-186197036 on Build GRCh38
rs780541319 Chr.4: 186196991-186196991 on Build GRCh38
rs569336882 Chr.4: 186197087-186197087 on Build GRCh38
rs750943905 Chr.4: 186197079-186197079 on Build GRCh38
rs749760428 Chr.4: 186197295-186197295 on Build GRCh38
rs995695271 Chr.4: 186196886-186196887 on Build GRCh38
rs150875391 Chr.4: 186197046-186197046 on Build GRCh38
rs35200327 Chr.4: 186196888-186196889 on Build GRCh38
rs758769153 Chr.4: 186196980-186196980 on Build GRCh38
rs760949006 Chr.4: 186197047-186197047 on Build GRCh38
rs558747441 Chr.4: 186197263-186197263 on Build GRCh38
rs1031744280 Chr.4: 186197340-186197340 on Build GRCh38
rs201407155 Chr.4: 186196888-186196888 on Build GRCh38
rs747545322 Chr.4: 186197121-186197121 on Build GRCh38
rs767779208 Chr.4: 186196939-186196939 on Build GRCh38
rs897447408 Chr.4: 186197183-186197183 on Build GRCh38
rs775816650 Chr.4: 186197035-186197035 on Build GRCh38
rs1014276097 Chr.4: 186196864-186196864 on Build GRCh38
rs770874016 Chr.4: 186197030-186197030 on Build GRCh38
rs746484929 Chr.4: 186197022-186197025 on Build GRCh38
rs72646259 Chr.4: 186197165-186197165 on Build GRCh38
rs753990809 Chr.4: 186196951-186196951 on Build GRCh38
rs145366496 Chr.4: 186197238-186197238 on Build GRCh38
rs79128796 Chr.4: 186197097-186197097 on Build GRCh38
rs746692527 Chr.4: 186196982-186196982 on Build GRCh38
rs78041259 Chr.4: 186197099-186197099 on Build GRCh38
rs199476190 Chr.4: 186197044-186197044 on Build GRCh38
rs769815150 Chr.4: 186196923-186196923 on Build GRCh38
rs780963135 Chr.4: 186197120-186197120 on Build GRCh38
rs747748338 Chr.4: 186196992-186196992 on Build GRCh38
rs1020701260 Chr.4: 186197242-186197243 on Build GRCh38
rs925107016 Chr.4: 186197107-186197108 on Build GRCh38
rs566817829 Chr.4: 186197227-186197227 on Build GRCh38
rs900659963 Chr.4: 186197237-186197237 on Build GRCh38
rs966684558 Chr.4: 186197256-186197256 on Build GRCh38
rs1015861865 Chr.4: 186196881-186196881 on Build GRCh38
rs370543462 Chr.4: 186197012-186197012 on Build GRCh38
rs769612802 Chr.4: 186196998-186196998 on Build GRCh38
rs1054782379 Chr.4: 186197142-186197142 on Build GRCh38
rs757683326 Chr.4: 186196956-186196956 on Build GRCh38

View Full Product Details