Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 314313-314819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHRR
Entrez Gene ID
57491
Gene Name
aryl-hydrocarbon receptor repressor
Gene Aliases
AHH, AHHR, bHLHe77
UniGene
Hs.50823
Gene Chromosome Location
Chr.5: 304176-438291 on Build GRCh38
Transcripts
NM_001242412.1 NM_020731.4 NM_001267556.1 NM_001267557.1 NM_001267558.1 NM_001267559.1 NM_013232.3

Gene Symbol
PDCD6
Entrez Gene ID
10016
Gene Name
programmed cell death 6
Gene Aliases
ALG-2, ALG2, PEF1B
UniGene
Hs.50823
Gene Chromosome Location
Chr.5: 271621-314974 on Build GRCh38
Transcripts
NM_001242412.1 NM_020731.4 NM_001267556.1 NM_001267557.1 NM_001267558.1 NM_001267559.1 NM_013232.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7214065 c.501T>A p.R167R Chr.5: 314440-314440 on Build GRCh38
5759459 c.517G>A p.D173N Chr.5: 314456-314456 on Build GRCh38
6742283 c.515A>G p.Q172R Chr.5: 314454-314454 on Build GRCh38
6742285 c.572T>C p.V191A Chr.5: 314511-314511 on Build GRCh38
5868392 c.499C>T p.R167C Chr.5: 314438-314438 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs901517601 Chr.5: 314630-314630 on Build GRCh38
rs951634157 Chr.5: 314549-314549 on Build GRCh38
rs774240050 Chr.5: 314421-314421 on Build GRCh38
rs749432176 Chr.5: 314691-314691 on Build GRCh38
rs934101317 Chr.5: 314339-314339 on Build GRCh38
rs73734221 Chr.5: 314634-314634 on Build GRCh38
rs774847534 Chr.5: 314693-314693 on Build GRCh38
rs909500893 Chr.5: 314429-314429 on Build GRCh38
rs1017619892 Chr.5: 314610-314610 on Build GRCh38
rs144783509 Chr.5: 314458-314458 on Build GRCh38
rs752050902 Chr.5: 314546-314546 on Build GRCh38
rs146107545 Chr.5: 314479-314479 on Build GRCh38
rs375697789 Chr.5: 314545-314545 on Build GRCh38
rs758754234 Chr.5: 314380-314380 on Build GRCh38
rs763545735 Chr.5: 314567-314567 on Build GRCh38
rs753438782 Chr.5: 314574-314574 on Build GRCh38
rs1035920080 Chr.5: 314767-314767 on Build GRCh38
rs888868367 Chr.5: 314656-314656 on Build GRCh38
rs750925812 Chr.5: 314379-314379 on Build GRCh38
rs778962868 Chr.5: 314504-314504 on Build GRCh38
rs780325757 Chr.5: 314384-314384 on Build GRCh38
rs752856839 Chr.5: 314722-314722 on Build GRCh38
rs139334790 Chr.5: 314439-314439 on Build GRCh38
rs146908777 Chr.5: 314404-314404 on Build GRCh38
rs866975794 Chr.5: 314337-314337 on Build GRCh38
rs1574220 Chr.5: 314403-314403 on Build GRCh38
rs964357289 Chr.5: 314611-314611 on Build GRCh38
rs1025986344 Chr.5: 314518-314518 on Build GRCh38
rs778153237 Chr.5: 314768-314768 on Build GRCh38
rs756806245 Chr.5: 314587-314587 on Build GRCh38
rs766748501 Chr.5: 314449-314449 on Build GRCh38
rs935237667 Chr.5: 314584-314584 on Build GRCh38
rs894280225 Chr.5: 314761-314761 on Build GRCh38
rs758628685 Chr.5: 314738-314738 on Build GRCh38
rs933943769 Chr.5: 314670-314670 on Build GRCh38
rs763389359 Chr.5: 314448-314448 on Build GRCh38
rs148124045 Chr.5: 314572-314572 on Build GRCh38
rs751964638 Chr.5: 314450-314450 on Build GRCh38
rs1041813929 Chr.5: 314341-314341 on Build GRCh38
rs201011657 Chr.5: 314438-314438 on Build GRCh38
rs750604562 Chr.5: 314626-314626 on Build GRCh38
rs780795882 Chr.5: 314402-314402 on Build GRCh38
rs972326665 Chr.5: 314723-314723 on Build GRCh38
rs138896105 Chr.5: 314495-314495 on Build GRCh38
rs761008685 Chr.5: 314424-314424 on Build GRCh38
rs745705216 Chr.5: 314510-314510 on Build GRCh38
rs745795259 Chr.5: 314612-314612 on Build GRCh38
rs758269411 Chr.5: 314624-314624 on Build GRCh38
rs773288149 Chr.5: 314687-314687 on Build GRCh38
rs762992222 Chr.5: 314532-314532 on Build GRCh38
rs770997224 Chr.5: 314692-314692 on Build GRCh38
rs779050494 Chr.5: 314595-314595 on Build GRCh38
rs948705301 Chr.5: 314789-314789 on Build GRCh38
rs773289429 Chr.5: 314528-314528 on Build GRCh38
rs915966826 Chr.5: 314548-314548 on Build GRCh38
rs769772556 Chr.5: 314527-314527 on Build GRCh38
rs370878298 Chr.5: 314480-314480 on Build GRCh38
rs960323215 Chr.5: 314777-314777 on Build GRCh38
rs750895835 Chr.5: 314443-314443 on Build GRCh38
rs755743116 Chr.5: 314564-314564 on Build GRCh38
rs201782998 Chr.5: 314418-314418 on Build GRCh38
rs771978012 Chr.5: 314516-314516 on Build GRCh38
rs149412009 Chr.5: 314444-314444 on Build GRCh38
rs369618664 Chr.5: 314462-314462 on Build GRCh38
rs751222340 Chr.5: 314388-314388 on Build GRCh38
rs553399647 Chr.5: 314686-314686 on Build GRCh38
rs866939105 Chr.5: 314523-314524 on Build GRCh38
rs997692807 Chr.5: 314399-314399 on Build GRCh38
rs759962998 Chr.5: 314552-314552 on Build GRCh38
rs752391766 Chr.5: 314476-314476 on Build GRCh38
rs769040089 Chr.5: 314425-314425 on Build GRCh38
rs749184352 Chr.5: 314410-314410 on Build GRCh38
rs754589073 Chr.5: 314389-314389 on Build GRCh38
rs776781897 Chr.5: 314517-314517 on Build GRCh38
rs759980982 Chr.5: 314704-314704 on Build GRCh38
rs372493961 Chr.5: 314558-314558 on Build GRCh38
rs751855779 Chr.5: 314793-314793 on Build GRCh38
rs199506819 Chr.5: 314551-314551 on Build GRCh38
rs770951720 Chr.5: 314498-314498 on Build GRCh38
rs748336524 Chr.5: 314635-314635 on Build GRCh38

View Full Product Details