Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 61526117-61526634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSWIM6
Entrez Gene ID
57688
Gene Name
zinc finger SWIM-type containing 6
Gene Aliases
AFND
UniGene
Hs.744939
Gene Chromosome Location
Chr.5: 61332273-61546172 on Build GRCh38
Transcripts
NM_020928.1 XM_017009677.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6470448 c.1697T>C p.V566A Chr.5: 61526256-61526256 on Build GRCh38
6202003 c.1762C>A p.L588I Chr.5: 61526321-61526321 on Build GRCh38
1486885 c.1813G>C p.E605Q Chr.5: 61526372-61526372 on Build GRCh38
4628453 c.1782T>C p.N594N Chr.5: 61526341-61526341 on Build GRCh38
1438073 c.1828_1829insA p.E613fs*5 Chr.5: 61526387-61526388 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760717798 Chr.5: 61526401-61526401 on Build GRCh38
rs564257275 Chr.5: 61526350-61526350 on Build GRCh38
rs754349666 Chr.5: 61526192-61526192 on Build GRCh38
rs183354495 Chr.5: 61526204-61526204 on Build GRCh38
rs187525163 Chr.5: 61526281-61526281 on Build GRCh38
rs752491016 Chr.5: 61526603-61526603 on Build GRCh38
rs781111121 Chr.5: 61526540-61526540 on Build GRCh38
rs887613067 Chr.5: 61526571-61526571 on Build GRCh38
rs929125026 Chr.5: 61526193-61526193 on Build GRCh38
rs968415621 Chr.5: 61526258-61526258 on Build GRCh38
rs990330044 Chr.5: 61526342-61526342 on Build GRCh38
rs954444840 Chr.5: 61526352-61526352 on Build GRCh38
rs918493495 Chr.5: 61526332-61526332 on Build GRCh38
rs888464367 Chr.5: 61526413-61526413 on Build GRCh38
rs887707348 Chr.5: 61526218-61526218 on Build GRCh38
rs918692061 Chr.5: 61526465-61526465 on Build GRCh38
rs968343342 Chr.5: 61526495-61526495 on Build GRCh38
rs1023089963 Chr.5: 61526260-61526260 on Build GRCh38
rs944040878 Chr.5: 61526439-61526439 on Build GRCh38
rs1021768229 Chr.5: 61526349-61526349 on Build GRCh38
rs377424540 Chr.5: 61526557-61526557 on Build GRCh38
rs374403415 Chr.5: 61526442-61526442 on Build GRCh38
rs1003063211 Chr.5: 61526284-61526284 on Build GRCh38
rs755603325 Chr.5: 61526514-61526514 on Build GRCh38
rs1012794487 Chr.5: 61526255-61526255 on Build GRCh38
rs1048891351 Chr.5: 61526505-61526505 on Build GRCh38
rs1031202379 Chr.5: 61526447-61526447 on Build GRCh38
rs1048859301 Chr.5: 61526207-61526207 on Build GRCh38
rs976654292 Chr.5: 61526329-61526329 on Build GRCh38
rs1011132372 Chr.5: 61526460-61526460 on Build GRCh38
rs772566819 Chr.5: 61526388-61526388 on Build GRCh38
rs948677235 Chr.5: 61526578-61526578 on Build GRCh38
rs867459936 Chr.5: 61526469-61526469 on Build GRCh38
rs988403045 Chr.5: 61526227-61526227 on Build GRCh38
rs770726727 Chr.5: 61526151-61526152 on Build GRCh38
rs190475981 Chr.5: 61526420-61526420 on Build GRCh38
rs116036211 Chr.5: 61526288-61526288 on Build GRCh38
rs1044305414 Chr.5: 61526593-61526593 on Build GRCh38
rs542331706 Chr.5: 61526590-61526591 on Build GRCh38
rs34246689 Chr.5: 61526591-61526592 on Build GRCh38
rs758684100 Chr.5: 61526494-61526494 on Build GRCh38
rs764549854 Chr.5: 61526179-61526179 on Build GRCh38
rs1004115960 Chr.5: 61526445-61526445 on Build GRCh38
rs397716099 Chr.5: 61526601-61526602 on Build GRCh38
rs1021250147 Chr.5: 61526253-61526253 on Build GRCh38
rs764942550 Chr.5: 61526387-61526388 on Build GRCh38
rs955750160 Chr.5: 61526333-61526333 on Build GRCh38
rs1021217408 Chr.5: 61526475-61526475 on Build GRCh38
rs560045642 Chr.5: 61526471-61526471 on Build GRCh38
rs776674553 Chr.5: 61526436-61526436 on Build GRCh38
rs191192967 Chr.5: 61526200-61526200 on Build GRCh38
rs529004231 Chr.5: 61526500-61526500 on Build GRCh38
rs185882718 Chr.5: 61526167-61526167 on Build GRCh38
rs761606372 Chr.5: 61526388-61526388 on Build GRCh38

View Full Product Details