Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 73174366-73174861 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM174
Entrez Gene ID
134288
Gene Name
transmembrane protein 174
Gene Aliases
-
UniGene
Hs.508588
Gene Chromosome Location
Chr.5: 73173196-73175143 on Build GRCh38
Transcripts
NM_153217.2

SNP Detail

SNP ID Chromosome Location
rs550280124 Chr.5: 73174776-73174776 on Build GRCh38
rs1034499664 Chr.5: 73174793-73174793 on Build GRCh38
rs370635221 Chr.5: 73174452-73174452 on Build GRCh38
rs546846256 Chr.5: 73174559-73174559 on Build GRCh38
rs1002213965 Chr.5: 73174473-73174473 on Build GRCh38
rs1010406332 Chr.5: 73174623-73174623 on Build GRCh38
rs967955669 Chr.5: 73174402-73174402 on Build GRCh38
rs892175409 Chr.5: 73174634-73174634 on Build GRCh38
rs960694392 Chr.5: 73174721-73174721 on Build GRCh38
rs1001733631 Chr.5: 73174746-73174746 on Build GRCh38
rs903785715 Chr.5: 73174679-73174679 on Build GRCh38
rs918588734 Chr.5: 73174517-73174517 on Build GRCh38
rs535927968 Chr.5: 73174827-73174827 on Build GRCh38
rs745959159 Chr.5: 73174674-73174674 on Build GRCh38
rs1013503941 Chr.5: 73174759-73174759 on Build GRCh38
rs911732906 Chr.5: 73174558-73174558 on Build GRCh38
rs575352520 Chr.5: 73174599-73174599 on Build GRCh38
rs900189637 Chr.5: 73174425-73174425 on Build GRCh38
rs909468439 Chr.5: 73174519-73174520 on Build GRCh38
rs149217273 Chr.5: 73174401-73174401 on Build GRCh38
rs1038828693 Chr.5: 73174569-73174569 on Build GRCh38
rs1054080739 Chr.5: 73174597-73174597 on Build GRCh38
rs757288254 Chr.5: 73174562-73174562 on Build GRCh38
rs944525815 Chr.5: 73174568-73174568 on Build GRCh38
rs1024903473 Chr.5: 73174467-73174467 on Build GRCh38
rs986863666 Chr.5: 73174557-73174557 on Build GRCh38
rs935915999 Chr.5: 73174586-73174586 on Build GRCh38
rs533902226 Chr.5: 73174801-73174801 on Build GRCh38
rs34964537 Chr.5: 73174600-73174601 on Build GRCh38
rs962994725 Chr.5: 73174794-73174794 on Build GRCh38
rs556394968 Chr.5: 73174489-73174489 on Build GRCh38
rs907378602 Chr.5: 73174701-73174701 on Build GRCh38
rs753123246 Chr.5: 73174572-73174572 on Build GRCh38
rs376686636 Chr.5: 73174455-73174455 on Build GRCh38
rs1104891 Chr.5: 73174828-73174828 on Build GRCh38
rs1045464424 Chr.5: 73174661-73174661 on Build GRCh38
rs999845284 Chr.5: 73174449-73174449 on Build GRCh38
rs538951580 Chr.5: 73174668-73174668 on Build GRCh38
rs1033676538 Chr.5: 73174713-73174714 on Build GRCh38
rs561411020 Chr.5: 73174456-73174456 on Build GRCh38
rs58753501 Chr.5: 73174637-73174638 on Build GRCh38
rs568031009 Chr.5: 73174677-73174677 on Build GRCh38
rs548054082 Chr.5: 73174642-73174643 on Build GRCh38
rs1034572672 Chr.5: 73174475-73174475 on Build GRCh38
rs146914922 Chr.5: 73174469-73174469 on Build GRCh38
rs879890959 Chr.5: 73174639-73174639 on Build GRCh38
rs987302016 Chr.5: 73174840-73174840 on Build GRCh38
rs746869066 Chr.5: 73174528-73174528 on Build GRCh38
rs781350233 Chr.5: 73174665-73174665 on Build GRCh38
rs1002056662 Chr.5: 73174698-73174698 on Build GRCh38
rs777741254 Chr.5: 73174516-73174516 on Build GRCh38
rs1039411881 Chr.5: 73174573-73174573 on Build GRCh38
rs747313558 Chr.5: 73174488-73174488 on Build GRCh38
rs962944151 Chr.5: 73174493-73174493 on Build GRCh38
rs921874886 Chr.5: 73174570-73174570 on Build GRCh38
rs775831344 Chr.5: 73174434-73174434 on Build GRCh38

View Full Product Details