Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 96662066-96662558 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAST
Entrez Gene ID
831
Gene Name
calpastatin
Gene Aliases
BS-17, PLACK
UniGene
Hs.436186
Gene Chromosome Location
Chr.5: 96662037-96774683 on Build GRCh38
Transcripts
NM_001042440.3 NM_001190442.1 XM_006714696.3 XM_006714697.3 XM_006714698.3 XM_006714700.3 XM_006714701.3 XM_006714703.3 XM_006714705.3 XM_011543654.2 XM_011543655.2 XM_011543656.2 XM_017009911.1 XM_017009912.1 XM_017009913.1 XM_017009914.1 XM_017009915.1 XM_017009916.1 XM_017009918.1 XM_017009919.1 XM_017009922.1 XM_017009923.1 XM_017009924.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5958242 c.46delC p.R16fs*69 Chr.5: 96662468-96662468 on Build GRCh38
5958241 c.40_41insG p.P14fs*12 Chr.5: 96662462-96662463 on Build GRCh38
5958245 c.48_49insG p.A17fs*9 Chr.5: 96662470-96662471 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs943679938 Chr.5: 96662351-96662351 on Build GRCh38
rs980628252 Chr.5: 96662282-96662282 on Build GRCh38
rs915998655 Chr.5: 96662199-96662199 on Build GRCh38
rs1054199329 Chr.5: 96662453-96662453 on Build GRCh38
rs1057211061 Chr.5: 96662261-96662261 on Build GRCh38
rs978596172 Chr.5: 96662481-96662481 on Build GRCh38
rs975722594 Chr.5: 96662441-96662441 on Build GRCh38
rs921006340 Chr.5: 96662419-96662419 on Build GRCh38
rs557919179 Chr.5: 96662198-96662198 on Build GRCh38
rs1039950195 Chr.5: 96662166-96662166 on Build GRCh38
rs981040992 Chr.5: 96662207-96662207 on Build GRCh38
rs1023570704 Chr.5: 96662228-96662228 on Build GRCh38
rs930966688 Chr.5: 96662105-96662105 on Build GRCh38
rs912446646 Chr.5: 96662245-96662245 on Build GRCh38
rs931270581 Chr.5: 96662222-96662222 on Build GRCh38
rs560335700 Chr.5: 96662536-96662536 on Build GRCh38
rs896042983 Chr.5: 96662268-96662268 on Build GRCh38
rs539689477 Chr.5: 96662178-96662178 on Build GRCh38
rs908486496 Chr.5: 96662309-96662309 on Build GRCh38
rs773457376 Chr.5: 96662321-96662324 on Build GRCh38
rs902972700 Chr.5: 96662259-96662259 on Build GRCh38
rs934411659 Chr.5: 96662523-96662523 on Build GRCh38
rs899552170 Chr.5: 96662177-96662177 on Build GRCh38
rs919793724 Chr.5: 96662284-96662284 on Build GRCh38
rs999071720 Chr.5: 96662264-96662264 on Build GRCh38
rs1031929009 Chr.5: 96662273-96662273 on Build GRCh38
rs769312998 Chr.5: 96662155-96662155 on Build GRCh38
rs565934362 Chr.5: 96662090-96662090 on Build GRCh38
rs919836203 Chr.5: 96662208-96662208 on Build GRCh38
rs916885140 Chr.5: 96662540-96662540 on Build GRCh38
rs544068142 Chr.5: 96662239-96662239 on Build GRCh38
rs574337274 Chr.5: 96662381-96662381 on Build GRCh38
rs145742747 Chr.5: 96662368-96662368 on Build GRCh38
rs899961865 Chr.5: 96662133-96662133 on Build GRCh38
rs904650806 Chr.5: 96662457-96662457 on Build GRCh38
rs998852349 Chr.5: 96662165-96662165 on Build GRCh38
rs947229512 Chr.5: 96662254-96662254 on Build GRCh38
rs1035944803 Chr.5: 96662211-96662211 on Build GRCh38
rs902836667 Chr.5: 96662184-96662184 on Build GRCh38
rs1014237137 Chr.5: 96662501-96662501 on Build GRCh38
rs541952198 Chr.5: 96662396-96662396 on Build GRCh38
rs774209204 Chr.5: 96662424-96662424 on Build GRCh38
rs555904487 Chr.5: 96662290-96662290 on Build GRCh38
rs982624915 Chr.5: 96662304-96662304 on Build GRCh38
rs975249893 Chr.5: 96662248-96662248 on Build GRCh38
rs1053334944 Chr.5: 96662469-96662469 on Build GRCh38
rs768176880 Chr.5: 96662389-96662389 on Build GRCh38
rs943846122 Chr.5: 96662130-96662130 on Build GRCh38
rs1041201142 Chr.5: 96662131-96662131 on Build GRCh38
rs921076297 Chr.5: 96662252-96662252 on Build GRCh38
rs1023240710 Chr.5: 96662297-96662297 on Build GRCh38
rs748702197 Chr.5: 96662376-96662376 on Build GRCh38
rs1042818121 Chr.5: 96662258-96662258 on Build GRCh38
rs1019381736 Chr.5: 96662434-96662434 on Build GRCh38
rs1046740573 Chr.5: 96662110-96662110 on Build GRCh38
rs921224034 Chr.5: 96662460-96662460 on Build GRCh38
rs964765462 Chr.5: 96662435-96662435 on Build GRCh38
rs943966123 Chr.5: 96662246-96662246 on Build GRCh38
rs1007967282 Chr.5: 96662129-96662129 on Build GRCh38
rs968989057 Chr.5: 96662156-96662156 on Build GRCh38
rs934276471 Chr.5: 96662260-96662260 on Build GRCh38
rs1042850053 Chr.5: 96662183-96662183 on Build GRCh38
rs995001771 Chr.5: 96662328-96662328 on Build GRCh38
rs934832851 Chr.5: 96662468-96662468 on Build GRCh38
rs778638665 Chr.5: 96662324-96662325 on Build GRCh38
rs796434517 Chr.5: 96662340-96662340 on Build GRCh38
rs987434447 Chr.5: 96662242-96662242 on Build GRCh38
rs899327600 Chr.5: 96662327-96662327 on Build GRCh38
rs960033576 Chr.5: 96662180-96662180 on Build GRCh38
rs1046952638 Chr.5: 96662125-96662125 on Build GRCh38
rs548216153 Chr.5: 96662320-96662321 on Build GRCh38
rs963407524 Chr.5: 96662232-96662232 on Build GRCh38
rs1026316346 Chr.5: 96662331-96662331 on Build GRCh38
rs527529256 Chr.5: 96662225-96662225 on Build GRCh38
rs774908016 Chr.5: 96662249-96662249 on Build GRCh38
rs969219244 Chr.5: 96662202-96662202 on Build GRCh38
rs1013606133 Chr.5: 96662278-96662278 on Build GRCh38
rs896060616 Chr.5: 96662219-96662219 on Build GRCh38
rs912508060 Chr.5: 96662126-96662126 on Build GRCh38
rs762544496 Chr.5: 96662274-96662274 on Build GRCh38
rs1026387095 Chr.5: 96662236-96662236 on Build GRCh38
rs973441403 Chr.5: 96662235-96662235 on Build GRCh38
rs1021850610 Chr.5: 96662150-96662150 on Build GRCh38
rs924323747 Chr.5: 96662496-96662496 on Build GRCh38
rs997444614 Chr.5: 96662139-96662139 on Build GRCh38
rs199812306 Chr.5: 96662089-96662089 on Build GRCh38
rs878906668 Chr.5: 96662423-96662423 on Build GRCh38
rs896099327 Chr.5: 96662488-96662488 on Build GRCh38
rs968536841 Chr.5: 96662462-96662462 on Build GRCh38

View Full Product Details