Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 126777664-126778163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LMNB1
Entrez Gene ID
4001
Gene Name
lamin B1
Gene Aliases
ADLD, LMN, LMN2, LMNB
UniGene
Hs.89497
Gene Chromosome Location
Chr.5: 126776623-126837020 on Build GRCh38
Transcripts
NM_001198557.1 NM_005573.3 NR_134488.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6360586 c.262G>A p.D88N Chr.5: 126777770-126777770 on Build GRCh38
6381939 c.235A>G p.K79E Chr.5: 126777743-126777743 on Build GRCh38
1186820 c.298C>A p.R100S Chr.5: 126777806-126777806 on Build GRCh38
4769354 c.269G>A p.R90Q Chr.5: 126777777-126777777 on Build GRCh38
6416087 c.277C>T p.L93F Chr.5: 126777785-126777785 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139629550 Chr.5: 126778056-126778056 on Build GRCh38
rs199906985 Chr.5: 126777947-126777948 on Build GRCh38
rs1016919784 Chr.5: 126777812-126777812 on Build GRCh38
rs563194936 Chr.5: 126777835-126777835 on Build GRCh38
rs758523326 Chr.5: 126777947-126777947 on Build GRCh38
rs1050790849 Chr.5: 126778040-126778040 on Build GRCh38
rs1043664541 Chr.5: 126778102-126778102 on Build GRCh38
rs903735021 Chr.5: 126778116-126778116 on Build GRCh38
rs1009469054 Chr.5: 126777862-126777862 on Build GRCh38
rs1029242875 Chr.5: 126777800-126777800 on Build GRCh38
rs765333856 Chr.5: 126777876-126777876 on Build GRCh38
rs769551613 Chr.5: 126777722-126777722 on Build GRCh38
rs560272218 Chr.5: 126777747-126777747 on Build GRCh38
rs1034786910 Chr.5: 126777724-126777724 on Build GRCh38
rs1056110934 Chr.5: 126778078-126778078 on Build GRCh38
rs368286117 Chr.5: 126777714-126777714 on Build GRCh38
rs372142905 Chr.5: 126777735-126777735 on Build GRCh38
rs567374793 Chr.5: 126777966-126777966 on Build GRCh38
rs958741242 Chr.5: 126777913-126777913 on Build GRCh38
rs1033970604 Chr.5: 126778142-126778142 on Build GRCh38
rs759651941 Chr.5: 126777866-126777866 on Build GRCh38
rs896211045 Chr.5: 126777751-126777751 on Build GRCh38
rs933779912 Chr.5: 126778036-126778036 on Build GRCh38
rs1022617034 Chr.5: 126777871-126777871 on Build GRCh38
rs74362780 Chr.5: 126777787-126777787 on Build GRCh38
rs548867767 Chr.5: 126777919-126777919 on Build GRCh38
rs1040240485 Chr.5: 126777994-126777994 on Build GRCh38
rs369733845 Chr.5: 126777867-126777867 on Build GRCh38
rs929947445 Chr.5: 126777937-126777937 on Build GRCh38
rs917288764 Chr.5: 126777936-126777936 on Build GRCh38
rs930251835 Chr.5: 126777691-126777691 on Build GRCh38
rs574016921 Chr.5: 126777719-126777719 on Build GRCh38
rs769735067 Chr.5: 126778137-126778137 on Build GRCh38
rs748913015 Chr.5: 126777725-126777725 on Build GRCh38
rs55750112 Chr.5: 126777895-126777895 on Build GRCh38
rs772419119 Chr.5: 126777774-126777774 on Build GRCh38
rs745556697 Chr.5: 126778125-126778125 on Build GRCh38
rs771565600 Chr.5: 126778138-126778138 on Build GRCh38
rs934051971 Chr.5: 126777933-126777933 on Build GRCh38
rs1039473632 Chr.5: 126777766-126777766 on Build GRCh38
rs943701027 Chr.5: 126777935-126777935 on Build GRCh38
rs892157074 Chr.5: 126778047-126778047 on Build GRCh38
rs910100822 Chr.5: 126777950-126777950 on Build GRCh38
rs546612432 Chr.5: 126778053-126778053 on Build GRCh38
rs796496632 Chr.5: 126777938-126777938 on Build GRCh38
rs746908601 Chr.5: 126777702-126777702 on Build GRCh38
rs964031149 Chr.5: 126777883-126777883 on Build GRCh38
rs941548784 Chr.5: 126777982-126777982 on Build GRCh38
rs562221299 Chr.5: 126777698-126777698 on Build GRCh38
rs189717541 Chr.5: 126777796-126777796 on Build GRCh38
rs534395188 Chr.5: 126778026-126778026 on Build GRCh38
rs913724243 Chr.5: 126777960-126777960 on Build GRCh38
rs946650432 Chr.5: 126777703-126777703 on Build GRCh38
rs892127611 Chr.5: 126777856-126777856 on Build GRCh38
rs780849458 Chr.5: 126777715-126777715 on Build GRCh38
rs777958553 Chr.5: 126778044-126778044 on Build GRCh38
rs77877457 Chr.5: 126777802-126777802 on Build GRCh38
rs747081714 Chr.5: 126778100-126778100 on Build GRCh38
rs922696668 Chr.5: 126777903-126777903 on Build GRCh38
rs1034447326 Chr.5: 126777898-126777898 on Build GRCh38
rs533097546 Chr.5: 126777801-126777801 on Build GRCh38

View Full Product Details