Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 136213232-136213741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRPC7
Entrez Gene ID
57113
Gene Name
transient receptor potential cation channel subfamily C member 7
Gene Aliases
TRP7
UniGene
Hs.591263
Gene Chromosome Location
Chr.5: 136213311-136365476 on Build GRCh38
Transcripts
NM_001167576.1 NM_001167577.1 NM_020389.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1433257 c.2586T>G p.I862M Chr.5: 136213438-136213438 on Build GRCh38
351121 c.2562G>C p.L854L Chr.5: 136213462-136213462 on Build GRCh38
7144614 c.2562G>A p.L854L Chr.5: 136213462-136213462 on Build GRCh38
5011491 c.2473G>C p.E825Q Chr.5: 136213551-136213551 on Build GRCh38
3850274 c.2540A>C p.K847T Chr.5: 136213484-136213484 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779500988 Chr.5: 136213490-136213490 on Build GRCh38
rs377403649 Chr.5: 136213634-136213634 on Build GRCh38
rs969361427 Chr.5: 136213367-136213367 on Build GRCh38
rs753412727 Chr.5: 136213633-136213633 on Build GRCh38
rs1020472664 Chr.5: 136213359-136213359 on Build GRCh38
rs769123609 Chr.5: 136213700-136213704 on Build GRCh38
rs376791511 Chr.5: 136213405-136213405 on Build GRCh38
rs868526545 Chr.5: 136213593-136213593 on Build GRCh38
rs377745502 Chr.5: 136213468-136213468 on Build GRCh38
rs746097813 Chr.5: 136213648-136213648 on Build GRCh38
rs371371913 Chr.5: 136213498-136213498 on Build GRCh38
rs563893550 Chr.5: 136213338-136213338 on Build GRCh38
rs772458087 Chr.5: 136213390-136213390 on Build GRCh38
rs764479274 Chr.5: 136213617-136213617 on Build GRCh38
rs751504622 Chr.5: 136213580-136213580 on Build GRCh38
rs779554156 Chr.5: 136213630-136213630 on Build GRCh38
rs765374203 Chr.5: 136213423-136213423 on Build GRCh38
rs115246677 Chr.5: 136213355-136213355 on Build GRCh38
rs1000396638 Chr.5: 136213557-136213557 on Build GRCh38
rs747388376 Chr.5: 136213504-136213504 on Build GRCh38
rs568707658 Chr.5: 136213375-136213375 on Build GRCh38
rs555431749 Chr.5: 136213695-136213695 on Build GRCh38
rs965716165 Chr.5: 136213689-136213689 on Build GRCh38
rs758679480 Chr.5: 136213430-136213431 on Build GRCh38
rs764158398 Chr.5: 136213531-136213531 on Build GRCh38
rs373487347 Chr.5: 136213393-136213393 on Build GRCh38
rs970248711 Chr.5: 136213319-136213319 on Build GRCh38
rs974375212 Chr.5: 136213702-136213702 on Build GRCh38
rs776755335 Chr.5: 136213515-136213515 on Build GRCh38
rs778191022 Chr.5: 136213640-136213640 on Build GRCh38
rs763959434 Chr.5: 136213441-136213441 on Build GRCh38
rs558151708 Chr.5: 136213642-136213642 on Build GRCh38
rs762992926 Chr.5: 136213537-136213537 on Build GRCh38
rs747518092 Chr.5: 136213641-136213641 on Build GRCh38
rs759737471 Chr.5: 136213421-136213421 on Build GRCh38
rs572917134 Chr.5: 136213279-136213279 on Build GRCh38
rs780768162 Chr.5: 136213458-136213458 on Build GRCh38
rs573133241 Chr.5: 136213647-136213647 on Build GRCh38
rs764009360 Chr.5: 136213539-136213539 on Build GRCh38
rs149040587 Chr.5: 136213651-136213651 on Build GRCh38
rs149909373 Chr.5: 136213293-136213293 on Build GRCh38
rs771303103 Chr.5: 136213508-136213508 on Build GRCh38
rs755715699 Chr.5: 136213612-136213612 on Build GRCh38
rs921676950 Chr.5: 136213388-136213388 on Build GRCh38
rs201400042 Chr.5: 136213518-136213518 on Build GRCh38
rs978923837 Chr.5: 136213331-136213331 on Build GRCh38
rs770012565 Chr.5: 136213521-136213521 on Build GRCh38
rs763005668 Chr.5: 136213652-136213652 on Build GRCh38
rs752709835 Chr.5: 136213426-136213426 on Build GRCh38
rs911957174 Chr.5: 136213424-136213424 on Build GRCh38
rs757074277 Chr.5: 136213450-136213450 on Build GRCh38
rs774888798 Chr.5: 136213387-136213387 on Build GRCh38
rs930381575 Chr.5: 136213720-136213720 on Build GRCh38
rs767313749 Chr.5: 136213602-136213602 on Build GRCh38
rs76887239 Chr.5: 136213344-136213344 on Build GRCh38
rs750058935 Chr.5: 136213459-136213459 on Build GRCh38
rs940826089 Chr.5: 136213431-136213431 on Build GRCh38
rs750108887 Chr.5: 136213603-136213603 on Build GRCh38
rs751419745 Chr.5: 136213639-136213639 on Build GRCh38
rs574338441 Chr.5: 136213704-136213704 on Build GRCh38
rs765070960 Chr.5: 136213372-136213372 on Build GRCh38
rs748677264 Chr.5: 136213497-136213497 on Build GRCh38
rs1034693728 Chr.5: 136213312-136213312 on Build GRCh38
rs762930386 Chr.5: 136213429-136213429 on Build GRCh38
rs761566093 Chr.5: 136213600-136213600 on Build GRCh38
rs866242690 Chr.5: 136213376-136213376 on Build GRCh38
rs536049603 Chr.5: 136213389-136213389 on Build GRCh38
rs751249702 Chr.5: 136213446-136213446 on Build GRCh38
rs368959669 Chr.5: 136213420-136213420 on Build GRCh38
rs373082826 Chr.5: 136213453-136213453 on Build GRCh38
rs753254021 Chr.5: 136213649-136213649 on Build GRCh38
rs767421151 Chr.5: 136213337-136213337 on Build GRCh38
rs904841475 Chr.5: 136213275-136213275 on Build GRCh38
rs921707148 Chr.5: 136213706-136213706 on Build GRCh38

View Full Product Details