Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 146129895-146130405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LARS
Entrez Gene ID
51520
Gene Name
leucyl-tRNA synthetase
Gene Aliases
HSPC192, ILFS1, LARS1, LEURS, LEUS, LFIS, LRS, PIG44, RNTLS, hr025Cl
UniGene
Hs.432674
Gene Chromosome Location
Chr.5: 146113026-146182787 on Build GRCh38
Transcripts
NM_001317964.1 NM_001317965.1 NM_016460.3 NM_020117.10 XM_011537656.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3730156 c.2575_2577delCTC p.L859delL Chr.5: 146130069-146130071 on Build GRCh38
4340415 c.2554A>G p.I852V Chr.5: 146130092-146130092 on Build GRCh38
6276630 c.2579C>A p.A860D Chr.5: 146130067-146130067 on Build GRCh38
1063764 c.2570T>G p.L857R Chr.5: 146130076-146130076 on Build GRCh38
149965 c.2628+8C>T p.? Chr.5: 146130010-146130010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs961374521 Chr.5: 146130229-146130229 on Build GRCh38
rs77219405 Chr.5: 146130119-146130119 on Build GRCh38
rs146505812 Chr.5: 146130155-146130155 on Build GRCh38
rs370418485 Chr.5: 146130069-146130069 on Build GRCh38
rs191151724 Chr.5: 146130288-146130288 on Build GRCh38
rs187036851 Chr.5: 146130107-146130107 on Build GRCh38
rs758426753 Chr.5: 146130026-146130026 on Build GRCh38
rs755733368 Chr.5: 146129992-146129992 on Build GRCh38
rs545928714 Chr.5: 146130030-146130030 on Build GRCh38
rs543004173 Chr.5: 146130006-146130006 on Build GRCh38
rs747191362 Chr.5: 146130037-146130037 on Build GRCh38
rs770802595 Chr.5: 146130142-146130142 on Build GRCh38
rs764452937 Chr.5: 146130004-146130004 on Build GRCh38
rs775052438 Chr.5: 146130156-146130156 on Build GRCh38
rs773047009 Chr.5: 146130051-146130051 on Build GRCh38
rs565853116 Chr.5: 146130373-146130373 on Build GRCh38
rs763708339 Chr.5: 146130097-146130097 on Build GRCh38
rs762201757 Chr.5: 146130101-146130101 on Build GRCh38
rs780107741 Chr.5: 146130120-146130120 on Build GRCh38
rs1051847309 Chr.5: 146130029-146130029 on Build GRCh38
rs371024015 Chr.5: 146129980-146129980 on Build GRCh38
rs760370344 Chr.5: 146130087-146130087 on Build GRCh38
rs768831398 Chr.5: 146130047-146130047 on Build GRCh38
rs75680405 Chr.5: 146130127-146130127 on Build GRCh38
rs927594016 Chr.5: 146130168-146130168 on Build GRCh38
rs563304209 Chr.5: 146130009-146130009 on Build GRCh38
rs570080802 Chr.5: 146130285-146130285 on Build GRCh38
rs368057623 Chr.5: 146130113-146130113 on Build GRCh38
rs770962213 Chr.5: 146130062-146130062 on Build GRCh38
rs200503223 Chr.5: 146130134-146130134 on Build GRCh38
rs112912805 Chr.5: 146130068-146130068 on Build GRCh38
rs144153578 Chr.5: 146130098-146130098 on Build GRCh38
rs781310755 Chr.5: 146130139-146130139 on Build GRCh38
rs548773989 Chr.5: 146130197-146130197 on Build GRCh38
rs200351329 Chr.5: 146130146-146130146 on Build GRCh38
rs764537553 Chr.5: 146130131-146130131 on Build GRCh38
rs752100018 Chr.5: 146130197-146130214 on Build GRCh38
rs765522441 Chr.5: 146130194-146130194 on Build GRCh38
rs751652832 Chr.5: 146130131-146130131 on Build GRCh38
rs373799044 Chr.5: 146130251-146130251 on Build GRCh38
rs1051791475 Chr.5: 146130377-146130377 on Build GRCh38
rs774393338 Chr.5: 146130065-146130065 on Build GRCh38
rs897057450 Chr.5: 146130313-146130313 on Build GRCh38
rs983801903 Chr.5: 146129928-146129928 on Build GRCh38
rs370412649 Chr.5: 146129982-146129982 on Build GRCh38
rs182471010 Chr.5: 146130347-146130347 on Build GRCh38
rs763338654 Chr.5: 146129981-146129982 on Build GRCh38
rs778156602 Chr.5: 146130167-146130167 on Build GRCh38
rs761470843 Chr.5: 146130170-146130170 on Build GRCh38
rs910746454 Chr.5: 146130313-146130314 on Build GRCh38
rs972388626 Chr.5: 146130263-146130263 on Build GRCh38
rs17104268 Chr.5: 146130165-146130165 on Build GRCh38
rs549899494 Chr.5: 146130276-146130276 on Build GRCh38
rs11953517 Chr.5: 146130010-146130010 on Build GRCh38
rs370606654 Chr.5: 146130140-146130140 on Build GRCh38
rs990536417 Chr.5: 146130212-146130212 on Build GRCh38
rs749087544 Chr.5: 146130060-146130060 on Build GRCh38
rs1007576766 Chr.5: 146130110-146130110 on Build GRCh38
rs922375564 Chr.5: 146130005-146130005 on Build GRCh38
rs530434307 Chr.5: 146130267-146130267 on Build GRCh38
rs937755911 Chr.5: 146130184-146130184 on Build GRCh38
rs143025329 Chr.5: 146129991-146129991 on Build GRCh38

View Full Product Details