Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148088327-148088828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPINK5
Entrez Gene ID
11005
Gene Name
serine peptidase inhibitor, Kazal type 5
Gene Aliases
LEKTI, LETKI, NETS, NS, VAKTI
UniGene
Hs.331555
Gene Chromosome Location
Chr.5: 148063972-148137362 on Build GRCh38
Transcripts
NM_001127698.1 NM_001127699.1 NM_006846.3 XM_011537551.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1227379 c.430G>T p.G144C Chr.5: 148088561-148088561 on Build GRCh38
6453623 c.442G>A p.E148K Chr.5: 148088573-148088573 on Build GRCh38
5750958 c.459C>A p.S153R Chr.5: 148088590-148088590 on Build GRCh38
3730667 c.411-6_411-5delTT p.? Chr.5: 148088536-148088537 on Build GRCh38
1227380 c.430G>T p.G144C Chr.5: 148088561-148088561 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748602271 Chr.5: 148088732-148088732 on Build GRCh38
rs748373697 Chr.5: 148088501-148088501 on Build GRCh38
rs897993132 Chr.5: 148088740-148088740 on Build GRCh38
rs971744480 Chr.5: 148088440-148088440 on Build GRCh38
rs959367716 Chr.5: 148088383-148088383 on Build GRCh38
rs575620403 Chr.5: 148088373-148088373 on Build GRCh38
rs990074605 Chr.5: 148088583-148088583 on Build GRCh38
rs945860906 Chr.5: 148088658-148088658 on Build GRCh38
rs372708333 Chr.5: 148088674-148088674 on Build GRCh38
rs1015548421 Chr.5: 148088451-148088451 on Build GRCh38
rs972904416 Chr.5: 148088463-148088463 on Build GRCh38
rs1055744144 Chr.5: 148088701-148088701 on Build GRCh38
rs1006921078 Chr.5: 148088703-148088703 on Build GRCh38
rs7711953 Chr.5: 148088671-148088671 on Build GRCh38
rs758086366 Chr.5: 148088553-148088553 on Build GRCh38
rs540247618 Chr.5: 148088631-148088631 on Build GRCh38
rs768703310 Chr.5: 148088520-148088520 on Build GRCh38
rs1048802304 Chr.5: 148088558-148088558 on Build GRCh38
rs1015145246 Chr.5: 148088704-148088704 on Build GRCh38
rs773392121 Chr.5: 148088671-148088673 on Build GRCh38
rs962230381 Chr.5: 148088457-148088457 on Build GRCh38
rs573158320 Chr.5: 148088549-148088549 on Build GRCh38
rs1001006227 Chr.5: 148088677-148088677 on Build GRCh38
rs887783334 Chr.5: 148088702-148088702 on Build GRCh38
rs990853688 Chr.5: 148088424-148088424 on Build GRCh38
rs752089232 Chr.5: 148088577-148088577 on Build GRCh38
rs778100225 Chr.5: 148088595-148088595 on Build GRCh38
rs997661195 Chr.5: 148088753-148088753 on Build GRCh38
rs764813956 Chr.5: 148088557-148088557 on Build GRCh38
rs777350307 Chr.5: 148088587-148088587 on Build GRCh38
rs1042606502 Chr.5: 148088673-148088673 on Build GRCh38
rs758755812 Chr.5: 148088564-148088564 on Build GRCh38
rs913064627 Chr.5: 148088612-148088612 on Build GRCh38
rs781019996 Chr.5: 148088676-148088676 on Build GRCh38
rs78661210 Chr.5: 148088441-148088441 on Build GRCh38
rs771881177 Chr.5: 148088620-148088620 on Build GRCh38
rs745625344 Chr.5: 148088523-148088523 on Build GRCh38
rs773579939 Chr.5: 148088519-148088519 on Build GRCh38
rs191190941 Chr.5: 148088684-148088684 on Build GRCh38
rs749514810 Chr.5: 148088597-148088597 on Build GRCh38
rs555026185 Chr.5: 148088515-148088515 on Build GRCh38
rs745822158 Chr.5: 148088695-148088695 on Build GRCh38
rs745691257 Chr.5: 148088530-148088530 on Build GRCh38
rs771795224 Chr.5: 148088535-148088535 on Build GRCh38
rs746962500 Chr.5: 148088627-148088627 on Build GRCh38
rs775206561 Chr.5: 148088626-148088626 on Build GRCh38
rs761266841 Chr.5: 148088638-148088638 on Build GRCh38
rs756071381 Chr.5: 148088536-148088537 on Build GRCh38
rs756283031 Chr.5: 148088591-148088591 on Build GRCh38
rs746673766 Chr.5: 148088611-148088611 on Build GRCh38
rs182285422 Chr.5: 148088775-148088775 on Build GRCh38
rs755061592 Chr.5: 148088497-148088497 on Build GRCh38
rs752159491 Chr.5: 148088482-148088482 on Build GRCh38
rs374790471 Chr.5: 148088571-148088571 on Build GRCh38
rs998185502 Chr.5: 148088672-148088672 on Build GRCh38
rs952425881 Chr.5: 148088402-148088402 on Build GRCh38
rs955138560 Chr.5: 148088793-148088793 on Build GRCh38
rs917923135 Chr.5: 148088426-148088426 on Build GRCh38
rs756587236 Chr.5: 148088433-148088433 on Build GRCh38
rs775468293 Chr.5: 148088537-148088537 on Build GRCh38
rs531000352 Chr.5: 148088581-148088581 on Build GRCh38
rs776194215 Chr.5: 148088551-148088551 on Build GRCh38
rs373960413 Chr.5: 148088499-148088499 on Build GRCh38
rs768517498 Chr.5: 148088630-148088630 on Build GRCh38
rs760579391 Chr.5: 148088548-148088548 on Build GRCh38

View Full Product Details