Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148268781-148269293 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC102546294
Entrez Gene ID
102546294
Gene Name
uncharacterized LOC102546294
Gene Aliases
-
UniGene
Hs.660461
Gene Chromosome Location
Chr.5: 148268307-148383850 on Build GRCh38
Transcripts
NR_105057.1 NM_001040129.2

Gene Symbol
SPINK13
Entrez Gene ID
153218
Gene Name
serine peptidase inhibitor, Kazal type 13 (putative)
Gene Aliases
HBVDNAPTP1, HESPINTOR, LiESP6, SPINK5L3
UniGene
Hs.483771
Gene Chromosome Location
Chr.5: 148268860-148286210 on Build GRCh38
Transcripts
NR_105057.1 NM_001040129.2

SNP Detail

SNP ID Chromosome Location
rs181014495 Chr.5: 148269142-148269142 on Build GRCh38
rs937176363 Chr.5: 148269121-148269121 on Build GRCh38
rs759778125 Chr.5: 148269215-148269215 on Build GRCh38
rs1027181452 Chr.5: 148268823-148268823 on Build GRCh38
rs897173580 Chr.5: 148269146-148269146 on Build GRCh38
rs984826083 Chr.5: 148268876-148268876 on Build GRCh38
rs748934859 Chr.5: 148268966-148268966 on Build GRCh38
rs952114516 Chr.5: 148268858-148268858 on Build GRCh38
rs1549886 Chr.5: 148268894-148268894 on Build GRCh38
rs1001672291 Chr.5: 148269071-148269071 on Build GRCh38
rs574448190 Chr.5: 148268990-148268990 on Build GRCh38
rs7733564 Chr.5: 148269187-148269187 on Build GRCh38
rs1027127224 Chr.5: 148268973-148268973 on Build GRCh38
rs541577154 Chr.5: 148269148-148269148 on Build GRCh38
rs935627678 Chr.5: 148268950-148268950 on Build GRCh38
rs765721402 Chr.5: 148269245-148269245 on Build GRCh38
rs140717841 Chr.5: 148268971-148268971 on Build GRCh38
rs974238000 Chr.5: 148268815-148268815 on Build GRCh38
rs755557959 Chr.5: 148268899-148268899 on Build GRCh38
rs1034538459 Chr.5: 148269111-148269111 on Build GRCh38
rs935925516 Chr.5: 148269226-148269226 on Build GRCh38
rs527488250 Chr.5: 148269198-148269198 on Build GRCh38
rs889924681 Chr.5: 148269182-148269182 on Build GRCh38
rs912881218 Chr.5: 148269254-148269254 on Build GRCh38
rs933836136 Chr.5: 148268930-148268930 on Build GRCh38
rs7734352 Chr.5: 148268947-148268947 on Build GRCh38
rs966047923 Chr.5: 148269054-148269054 on Build GRCh38
rs954266421 Chr.5: 148268847-148268847 on Build GRCh38
rs560255036 Chr.5: 148269135-148269135 on Build GRCh38
rs1037023524 Chr.5: 148269144-148269144 on Build GRCh38
rs907910950 Chr.5: 148268879-148268879 on Build GRCh38
rs947177317 Chr.5: 148268924-148268924 on Build GRCh38
rs138513680 Chr.5: 148268946-148268946 on Build GRCh38
rs1013582307 Chr.5: 148268961-148268961 on Build GRCh38
rs576321054 Chr.5: 148269014-148269014 on Build GRCh38
rs111341055 Chr.5: 148268887-148268887 on Build GRCh38
rs79126418 Chr.5: 148269269-148269269 on Build GRCh38
rs115597309 Chr.5: 148269112-148269112 on Build GRCh38
rs976146554 Chr.5: 148269158-148269158 on Build GRCh38
rs145855103 Chr.5: 148268993-148268993 on Build GRCh38
rs912972688 Chr.5: 148268913-148268913 on Build GRCh38
rs1016051546 Chr.5: 148268962-148268962 on Build GRCh38
rs907369729 Chr.5: 148269038-148269038 on Build GRCh38
rs768443409 Chr.5: 148268986-148268986 on Build GRCh38
rs555867581 Chr.5: 148269131-148269131 on Build GRCh38
rs748104865 Chr.5: 148269109-148269109 on Build GRCh38
rs760853712 Chr.5: 148269154-148269154 on Build GRCh38
rs568568213 Chr.5: 148268928-148268928 on Build GRCh38
rs985724049 Chr.5: 148268888-148268888 on Build GRCh38
rs955162512 Chr.5: 148269173-148269173 on Build GRCh38
rs896436526 Chr.5: 148268955-148268955 on Build GRCh38
rs940629076 Chr.5: 148269137-148269137 on Build GRCh38
rs927104800 Chr.5: 148269114-148269114 on Build GRCh38
rs778620024 Chr.5: 148269093-148269093 on Build GRCh38
rs549903597 Chr.5: 148268895-148268895 on Build GRCh38
rs535560491 Chr.5: 148268929-148268929 on Build GRCh38
rs138321621 Chr.5: 148269171-148269171 on Build GRCh38
rs1009940893 Chr.5: 148268985-148268985 on Build GRCh38
rs1019873729 Chr.5: 148269031-148269031 on Build GRCh38
rs758928343 Chr.5: 148268809-148268809 on Build GRCh38
rs188121392 Chr.5: 148268812-148268812 on Build GRCh38

View Full Product Details