Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150648188-150648676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNPO
Entrez Gene ID
11346
Gene Name
synaptopodin
Gene Aliases
-
UniGene
Hs.435228
Gene Chromosome Location
Chr.5: 150586026-150659230 on Build GRCh38
Transcripts
NM_001109974.2 NM_001166208.1 NM_001166209.1 NM_007286.5 XM_005268370.1 XM_005268371.1 XM_006714755.3 XM_011537552.2 XM_017009007.1 XM_017009008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7328050 c.143G>T p.R48L Chr.5: 150648418-150648418 on Build GRCh38
4731642 c.263G>A p.S88N Chr.5: 150648538-150648538 on Build GRCh38
7146878 c.972C>T p.L324L Chr.5: 150648515-150648515 on Build GRCh38
376303 c.366A>G p.Q122Q Chr.5: 150648641-150648641 on Build GRCh38
3244372 c.1113C>A p.G371G Chr.5: 150648656-150648656 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs533483214 Chr.5: 150648656-150648656 on Build GRCh38
rs754284257 Chr.5: 150648228-150648228 on Build GRCh38
rs765720877 Chr.5: 150648312-150648312 on Build GRCh38
rs376542347 Chr.5: 150648347-150648347 on Build GRCh38
rs770054530 Chr.5: 150648382-150648382 on Build GRCh38
rs775082855 Chr.5: 150648515-150648517 on Build GRCh38
rs201766640 Chr.5: 150648440-150648440 on Build GRCh38
rs767411471 Chr.5: 150648274-150648274 on Build GRCh38
rs555455244 Chr.5: 150648512-150648512 on Build GRCh38
rs377264111 Chr.5: 150648261-150648261 on Build GRCh38
rs766223815 Chr.5: 150648207-150648207 on Build GRCh38
rs370835156 Chr.5: 150648269-150648269 on Build GRCh38
rs758625709 Chr.5: 150648344-150648344 on Build GRCh38
rs745821356 Chr.5: 150648391-150648391 on Build GRCh38
rs759972071 Chr.5: 150648543-150648543 on Build GRCh38
rs370095729 Chr.5: 150648351-150648351 on Build GRCh38
rs755256041 Chr.5: 150648356-150648356 on Build GRCh38
rs764365021 Chr.5: 150648618-150648618 on Build GRCh38
rs761471013 Chr.5: 150648511-150648511 on Build GRCh38
rs780484672 Chr.5: 150648348-150648348 on Build GRCh38
rs755439626 Chr.5: 150648444-150648444 on Build GRCh38
rs776616689 Chr.5: 150648400-150648400 on Build GRCh38
rs764040108 Chr.5: 150648555-150648555 on Build GRCh38
rs763556481 Chr.5: 150648292-150648292 on Build GRCh38
rs763541477 Chr.5: 150648439-150648439 on Build GRCh38
rs748815651 Chr.5: 150648578-150648578 on Build GRCh38
rs373212175 Chr.5: 150648598-150648598 on Build GRCh38
rs757296990 Chr.5: 150648627-150648627 on Build GRCh38
rs998711559 Chr.5: 150648457-150648457 on Build GRCh38
rs768275869 Chr.5: 150648416-150648416 on Build GRCh38
rs759236741 Chr.5: 150648270-150648270 on Build GRCh38
rs751009044 Chr.5: 150648314-150648314 on Build GRCh38
rs200631093 Chr.5: 150648499-150648499 on Build GRCh38
rs142079099 Chr.5: 150648447-150648447 on Build GRCh38
rs373632479 Chr.5: 150648401-150648401 on Build GRCh38
rs761363144 Chr.5: 150648310-150648310 on Build GRCh38
rs753932064 Chr.5: 150648557-150648557 on Build GRCh38
rs764733581 Chr.5: 150648423-150648423 on Build GRCh38
rs984981628 Chr.5: 150648605-150648605 on Build GRCh38
rs760428128 Chr.5: 150648405-150648405 on Build GRCh38
rs758857838 Chr.5: 150648242-150648242 on Build GRCh38
rs774852438 Chr.5: 150648591-150648591 on Build GRCh38
rs371173029 Chr.5: 150648229-150648229 on Build GRCh38
rs933104552 Chr.5: 150648516-150648516 on Build GRCh38
rs747912249 Chr.5: 150648657-150648657 on Build GRCh38
rs765248495 Chr.5: 150648637-150648637 on Build GRCh38
rs767996337 Chr.5: 150648544-150648544 on Build GRCh38
rs920962429 Chr.5: 150648404-150648404 on Build GRCh38
rs893799015 Chr.5: 150648247-150648247 on Build GRCh38
rs1045058276 Chr.5: 150648630-150648630 on Build GRCh38
rs754076512 Chr.5: 150648620-150648620 on Build GRCh38
rs751918002 Chr.5: 150648443-150648443 on Build GRCh38
rs368415231 Chr.5: 150648533-150648533 on Build GRCh38
rs778987463 Chr.5: 150648389-150648389 on Build GRCh38
rs573686646 Chr.5: 150648536-150648536 on Build GRCh38
rs759413850 Chr.5: 150648211-150648211 on Build GRCh38
rs761309899 Chr.5: 150648420-150648420 on Build GRCh38
rs542377558 Chr.5: 150648254-150648254 on Build GRCh38
rs753314882 Chr.5: 150648295-150648295 on Build GRCh38
rs754711985 Chr.5: 150648571-150648571 on Build GRCh38
rs530690446 Chr.5: 150648586-150648586 on Build GRCh38
rs754943596 Chr.5: 150648653-150648653 on Build GRCh38
rs780037983 Chr.5: 150648558-150648558 on Build GRCh38
rs192726670 Chr.5: 150648424-150648424 on Build GRCh38
rs376253597 Chr.5: 150648655-150648655 on Build GRCh38
rs774775211 Chr.5: 150648540-150648540 on Build GRCh38
rs776868872 Chr.5: 150648256-150648256 on Build GRCh38
rs770958059 Chr.5: 150648268-150648268 on Build GRCh38
rs776028959 Chr.5: 150648607-150648607 on Build GRCh38
rs750298045 Chr.5: 150648484-150648484 on Build GRCh38
rs369769452 Chr.5: 150648577-150648577 on Build GRCh38
rs367768458 Chr.5: 150648469-150648469 on Build GRCh38
rs762406403 Chr.5: 150648534-150648534 on Build GRCh38
rs376126437 Chr.5: 150648251-150648251 on Build GRCh38
rs866208549 Chr.5: 150648474-150648474 on Build GRCh38
rs560109540 Chr.5: 150648576-150648576 on Build GRCh38
rs746955643 Chr.5: 150648560-150648560 on Build GRCh38
rs201288895 Chr.5: 150648547-150648547 on Build GRCh38
rs767052370 Chr.5: 150648212-150648212 on Build GRCh38
rs774315632 Chr.5: 150648209-150648209 on Build GRCh38
rs151142897 Chr.5: 150648608-150648608 on Build GRCh38
rs370579156 Chr.5: 150648494-150648494 on Build GRCh38
rs147580348 Chr.5: 150648329-150648329 on Build GRCh38
rs748214237 Chr.5: 150648374-150648374 on Build GRCh38
rs759517134 Chr.5: 150648418-150648418 on Build GRCh38
rs770697443 Chr.5: 150648287-150648287 on Build GRCh38
rs771705173 Chr.5: 150648394-150648394 on Build GRCh38
rs760131670 Chr.5: 150648593-150648593 on Build GRCh38
rs373884404 Chr.5: 150648290-150648290 on Build GRCh38
rs779437380 Chr.5: 150648231-150648231 on Build GRCh38
rs752451143 Chr.5: 150648220-150648220 on Build GRCh38
rs78583075 Chr.5: 150648252-150648252 on Build GRCh38
rs779905679 Chr.5: 150648492-150648492 on Build GRCh38
rs746212122 Chr.5: 150648236-150648236 on Build GRCh38
rs781601503 Chr.5: 150648362-150648362 on Build GRCh38
rs751452946 Chr.5: 150648647-150648647 on Build GRCh38
rs145720408 Chr.5: 150648572-150648572 on Build GRCh38
rs199790421 Chr.5: 150648500-150648500 on Build GRCh38
rs756426210 Chr.5: 150648548-150648548 on Build GRCh38
rs753032848 Chr.5: 150648449-150648449 on Build GRCh38
rs1052327589 Chr.5: 150648525-150648525 on Build GRCh38
rs763664246 Chr.5: 150648222-150648222 on Build GRCh38

View Full Product Details