Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150894304-150894811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRGM
Entrez Gene ID
345611
Gene Name
immunity related GTPase M
Gene Aliases
IFI1, IRGM1, LRG-47, LRG47
UniGene
Hs.519680
Gene Chromosome Location
Chr.5: 150846523-150902402 on Build GRCh38
Transcripts
XM_011537641.2 NM_001172831.1 NM_001172832.1 NM_052860.2 XM_011537702.2

Gene Symbol
ZNF300
Entrez Gene ID
91975
Gene Name
zinc finger protein 300
Gene Aliases
-
UniGene
Hs.134885
Gene Chromosome Location
Chr.5: 150892958-150904983 on Build GRCh38
Transcripts
XM_011537641.2 NM_001172831.1 NM_001172832.1 NM_052860.2 XM_011537702.2

SNP Detail

SNP ID Chromosome Location
rs1025585218 Chr.5: 150894431-150894431 on Build GRCh38
rs79899193 Chr.5: 150894546-150894546 on Build GRCh38
rs1025921370 Chr.5: 150894703-150894703 on Build GRCh38
rs936391184 Chr.5: 150894459-150894459 on Build GRCh38
rs897301459 Chr.5: 150894512-150894512 on Build GRCh38
rs966560316 Chr.5: 150894621-150894621 on Build GRCh38
rs146356008 Chr.5: 150894747-150894747 on Build GRCh38
rs528741343 Chr.5: 150894741-150894741 on Build GRCh38
rs186211186 Chr.5: 150894573-150894573 on Build GRCh38
rs970880261 Chr.5: 150894447-150894447 on Build GRCh38
rs913307882 Chr.5: 150894586-150894586 on Build GRCh38
rs142316443 Chr.5: 150894730-150894730 on Build GRCh38
rs190676262 Chr.5: 150894675-150894675 on Build GRCh38
rs866391199 Chr.5: 150894434-150894434 on Build GRCh38
rs535351663 Chr.5: 150894343-150894343 on Build GRCh38
rs1012782525 Chr.5: 150894644-150894644 on Build GRCh38
rs140840542 Chr.5: 150894454-150894454 on Build GRCh38
rs796427291 Chr.5: 150894481-150894481 on Build GRCh38
rs150977382 Chr.5: 150894417-150894417 on Build GRCh38
rs775782434 Chr.5: 150894576-150894576 on Build GRCh38
rs925259268 Chr.5: 150894661-150894661 on Build GRCh38
rs552661833 Chr.5: 150894346-150894346 on Build GRCh38
rs773150865 Chr.5: 150894416-150894416 on Build GRCh38
rs544362428 Chr.5: 150894507-150894507 on Build GRCh38
rs145786914 Chr.5: 150894686-150894687 on Build GRCh38
rs181897244 Chr.5: 150894501-150894501 on Build GRCh38
rs952912155 Chr.5: 150894721-150894721 on Build GRCh38
rs976802061 Chr.5: 150894577-150894578 on Build GRCh38
rs1054802251 Chr.5: 150894685-150894685 on Build GRCh38
rs911343028 Chr.5: 150894369-150894370 on Build GRCh38
rs748581687 Chr.5: 150894336-150894336 on Build GRCh38
rs1016368147 Chr.5: 150894731-150894731 on Build GRCh38
rs918811045 Chr.5: 150894493-150894493 on Build GRCh38
rs943142528 Chr.5: 150894516-150894516 on Build GRCh38
rs146076874 Chr.5: 150894503-150894503 on Build GRCh38
rs1032444889 Chr.5: 150894358-150894358 on Build GRCh38
rs556635350 Chr.5: 150894452-150894452 on Build GRCh38
rs757141999 Chr.5: 150894673-150894673 on Build GRCh38
rs994300129 Chr.5: 150894701-150894701 on Build GRCh38
rs116833167 Chr.5: 150894328-150894328 on Build GRCh38
rs542443834 Chr.5: 150894463-150894463 on Build GRCh38
rs974906305 Chr.5: 150894779-150894779 on Build GRCh38
rs772702516 Chr.5: 150894354-150894354 on Build GRCh38
rs916209161 Chr.5: 150894694-150894697 on Build GRCh38
rs945287276 Chr.5: 150894448-150894448 on Build GRCh38
rs902529068 Chr.5: 150894350-150894350 on Build GRCh38
rs577231557 Chr.5: 150894385-150894385 on Build GRCh38
rs138937564 Chr.5: 150894552-150894552 on Build GRCh38
rs531058943 Chr.5: 150894630-150894630 on Build GRCh38
rs10515643 Chr.5: 150894517-150894517 on Build GRCh38
rs1055193397 Chr.5: 150894489-150894489 on Build GRCh38
rs933121169 Chr.5: 150894542-150894542 on Build GRCh38
rs530326439 Chr.5: 150894530-150894530 on Build GRCh38
rs751406652 Chr.5: 150894566-150894566 on Build GRCh38

View Full Product Details