Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 163456614-163457128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUDCD2
Entrez Gene ID
134492
Gene Name
NudC domain containing 2
Gene Aliases
NudCL2
UniGene
Hs.140443
Gene Chromosome Location
Chr.5: 163453580-163460137 on Build GRCh38
Transcripts
NM_145266.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6418014 c.239A>T p.E80V Chr.5: 163457080-163457080 on Build GRCh38
1733524 c.239-3delT p.? Chr.5: 163457083-163457083 on Build GRCh38
5060901 c.330G>A p.A110A Chr.5: 163456989-163456989 on Build GRCh38
6170310 c.373G>C p.E125Q Chr.5: 163456946-163456946 on Build GRCh38
5060900 c.336T>A p.P112P Chr.5: 163456983-163456983 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs75693765 Chr.5: 163456758-163456758 on Build GRCh38
rs1001375651 Chr.5: 163456668-163456668 on Build GRCh38
rs764231026 Chr.5: 163456989-163456989 on Build GRCh38
rs922859107 Chr.5: 163456829-163456829 on Build GRCh38
rs1057042337 Chr.5: 163456669-163456669 on Build GRCh38
rs200628619 Chr.5: 163456888-163456888 on Build GRCh38
rs907570590 Chr.5: 163456996-163456996 on Build GRCh38
rs778036785 Chr.5: 163456806-163456806 on Build GRCh38
rs749002124 Chr.5: 163457091-163457092 on Build GRCh38
rs773237977 Chr.5: 163456957-163456957 on Build GRCh38
rs372168863 Chr.5: 163457070-163457070 on Build GRCh38
rs369487816 Chr.5: 163457010-163457010 on Build GRCh38
rs769395107 Chr.5: 163457083-163457084 on Build GRCh38
rs746568322 Chr.5: 163457032-163457032 on Build GRCh38
rs150464483 Chr.5: 163457031-163457031 on Build GRCh38
rs767606090 Chr.5: 163456968-163456968 on Build GRCh38
rs932595253 Chr.5: 163457004-163457004 on Build GRCh38
rs200190066 Chr.5: 163457091-163457091 on Build GRCh38
rs747457697 Chr.5: 163457083-163457083 on Build GRCh38
rs755112083 Chr.5: 163456946-163456947 on Build GRCh38
rs778317662 Chr.5: 163457082-163457083 on Build GRCh38
rs376036154 Chr.5: 163456984-163456984 on Build GRCh38
rs1003980330 Chr.5: 163456775-163456775 on Build GRCh38
rs370580721 Chr.5: 163457049-163457049 on Build GRCh38
rs201814363 Chr.5: 163457100-163457100 on Build GRCh38
rs761634543 Chr.5: 163456950-163456950 on Build GRCh38
rs978112776 Chr.5: 163456651-163456651 on Build GRCh38
rs752773367 Chr.5: 163456976-163456976 on Build GRCh38
rs1020733797 Chr.5: 163456746-163456746 on Build GRCh38
rs758910013 Chr.5: 163457041-163457042 on Build GRCh38
rs371791584 Chr.5: 163456944-163456944 on Build GRCh38
rs530503422 Chr.5: 163456873-163456873 on Build GRCh38
rs895740426 Chr.5: 163456681-163456681 on Build GRCh38
rs966652700 Chr.5: 163456861-163456861 on Build GRCh38
rs905655225 Chr.5: 163456645-163456645 on Build GRCh38
rs937134231 Chr.5: 163457060-163457060 on Build GRCh38
rs774901995 Chr.5: 163456902-163456902 on Build GRCh38
rs185970759 Chr.5: 163456778-163456778 on Build GRCh38
rs746651260 Chr.5: 163456917-163456917 on Build GRCh38
rs1024982918 Chr.5: 163456855-163456855 on Build GRCh38
rs139292728 Chr.5: 163457042-163457042 on Build GRCh38
rs1015331195 Chr.5: 163456776-163456779 on Build GRCh38
rs368381995 Chr.5: 163456993-163456993 on Build GRCh38
rs932342750 Chr.5: 163456757-163456757 on Build GRCh38
rs866182845 Chr.5: 163457033-163457033 on Build GRCh38
rs113515886 Chr.5: 163457093-163457094 on Build GRCh38
rs559396604 Chr.5: 163456698-163456698 on Build GRCh38
rs762060193 Chr.5: 163457097-163457097 on Build GRCh38
rs373523422 Chr.5: 163457040-163457043 on Build GRCh38
rs113736533 Chr.5: 163456796-163456797 on Build GRCh38
rs543711948 Chr.5: 163457063-163457063 on Build GRCh38
rs747274702 Chr.5: 163457039-163457042 on Build GRCh38
rs899733619 Chr.5: 163456738-163456738 on Build GRCh38
rs1034381188 Chr.5: 163456799-163456799 on Build GRCh38
rs964148089 Chr.5: 163456784-163456784 on Build GRCh38
rs935813134 Chr.5: 163456688-163456688 on Build GRCh38
rs201931869 Chr.5: 163457093-163457093 on Build GRCh38
rs150010030 Chr.5: 163457067-163457067 on Build GRCh38
rs754127091 Chr.5: 163456902-163456903 on Build GRCh38
rs142159368 Chr.5: 163457007-163457007 on Build GRCh38
rs1000278532 Chr.5: 163456773-163456773 on Build GRCh38
rs140642151 Chr.5: 163456998-163456998 on Build GRCh38
rs749363112 Chr.5: 163456927-163456927 on Build GRCh38
rs779401727 Chr.5: 163457092-163457093 on Build GRCh38
rs774345989 Chr.5: 163457087-163457087 on Build GRCh38
rs749716070 Chr.5: 163456884-163456884 on Build GRCh38
rs951745192 Chr.5: 163456880-163456880 on Build GRCh38
rs757618982 Chr.5: 163456886-163456886 on Build GRCh38
rs769674467 Chr.5: 163457058-163457058 on Build GRCh38
rs762831815 Chr.5: 163456967-163456967 on Build GRCh38
rs150776333 Chr.5: 163457009-163457009 on Build GRCh38
rs768250901 Chr.5: 163456938-163456938 on Build GRCh38
rs1054321483 Chr.5: 163456730-163456730 on Build GRCh38
rs576383191 Chr.5: 163457035-163457036 on Build GRCh38
rs78084002 Chr.5: 163456779-163456779 on Build GRCh38
rs147218174 Chr.5: 163456956-163456956 on Build GRCh38
rs747827504 Chr.5: 163457050-163457050 on Build GRCh38
rs78759977 Chr.5: 163457092-163457092 on Build GRCh38
rs779331736 Chr.5: 163456887-163456887 on Build GRCh38

View Full Product Details