Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 168568513-168569010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PANK3
Entrez Gene ID
79646
Gene Name
pantothenate kinase 3
Gene Aliases
-
UniGene
Hs.388400
Gene Chromosome Location
Chr.5: 168555623-168579609 on Build GRCh38
Transcripts
NM_024594.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1065645 c.145C>T p.R49W Chr.5: 168568882-168568882 on Build GRCh38
218838 c.362T>G p.F121C Chr.5: 168568665-168568665 on Build GRCh38
4392027 c.135A>T p.L45F Chr.5: 168568892-168568892 on Build GRCh38
3393426 c.377G>A p.R126H Chr.5: 168568650-168568650 on Build GRCh38
6102987 c.324G>C p.Q108H Chr.5: 168568703-168568703 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008035336 Chr.5: 168568580-168568580 on Build GRCh38
rs148664216 Chr.5: 168568579-168568579 on Build GRCh38
rs1055428331 Chr.5: 168568618-168568618 on Build GRCh38
rs116218738 Chr.5: 168568847-168568847 on Build GRCh38
rs1011622419 Chr.5: 168568555-168568555 on Build GRCh38
rs1026936399 Chr.5: 168568685-168568685 on Build GRCh38
rs899726905 Chr.5: 168568875-168568875 on Build GRCh38
rs990470602 Chr.5: 168568751-168568751 on Build GRCh38
rs771045313 Chr.5: 168568700-168568700 on Build GRCh38
rs762609025 Chr.5: 168568929-168568929 on Build GRCh38
rs916320111 Chr.5: 168568790-168568790 on Build GRCh38
rs71603869 Chr.5: 168568921-168568921 on Build GRCh38
rs781333716 Chr.5: 168568690-168568690 on Build GRCh38
rs543643077 Chr.5: 168568880-168568880 on Build GRCh38
rs760823330 Chr.5: 168568854-168568854 on Build GRCh38
rs569700809 Chr.5: 168568861-168568861 on Build GRCh38
rs765982316 Chr.5: 168568942-168568942 on Build GRCh38
rs893135385 Chr.5: 168568551-168568551 on Build GRCh38
rs780025307 Chr.5: 168568629-168568629 on Build GRCh38
rs771903715 Chr.5: 168568538-168568538 on Build GRCh38
rs562249889 Chr.5: 168568582-168568582 on Build GRCh38
rs952481069 Chr.5: 168568725-168568725 on Build GRCh38
rs144173342 Chr.5: 168568565-168568565 on Build GRCh38
rs745984042 Chr.5: 168568844-168568844 on Build GRCh38
rs931114654 Chr.5: 168568709-168568709 on Build GRCh38
rs759836686 Chr.5: 168568703-168568703 on Build GRCh38
rs150026739 Chr.5: 168568774-168568774 on Build GRCh38
rs756859202 Chr.5: 168568607-168568607 on Build GRCh38
rs374391109 Chr.5: 168568739-168568739 on Build GRCh38
rs764634057 Chr.5: 168568597-168568597 on Build GRCh38
rs531090717 Chr.5: 168568811-168568811 on Build GRCh38
rs749761171 Chr.5: 168568667-168568669 on Build GRCh38
rs149532439 Chr.5: 168568550-168568550 on Build GRCh38
rs749859432 Chr.5: 168568763-168568763 on Build GRCh38
rs748141409 Chr.5: 168568827-168568827 on Build GRCh38
rs779672597 Chr.5: 168568620-168568620 on Build GRCh38
rs765699854 Chr.5: 168568922-168568922 on Build GRCh38
rs914851368 Chr.5: 168568913-168568913 on Build GRCh38
rs752476992 Chr.5: 168568961-168568961 on Build GRCh38
rs754884394 Chr.5: 168568795-168568795 on Build GRCh38
rs771616770 Chr.5: 168568774-168568775 on Build GRCh38
rs746840803 Chr.5: 168568650-168568650 on Build GRCh38
rs772324108 Chr.5: 168568718-168568718 on Build GRCh38
rs928798207 Chr.5: 168568848-168568848 on Build GRCh38
rs936992004 Chr.5: 168568616-168568616 on Build GRCh38
rs777736705 Chr.5: 168568837-168568837 on Build GRCh38
rs761086843 Chr.5: 168568731-168568731 on Build GRCh38
rs1019474752 Chr.5: 168568588-168568588 on Build GRCh38
rs768545471 Chr.5: 168568658-168568658 on Build GRCh38
rs144506227 Chr.5: 168568791-168568791 on Build GRCh38
rs947356335 Chr.5: 168568932-168568932 on Build GRCh38
rs779896661 Chr.5: 168568780-168568780 on Build GRCh38
rs199884430 Chr.5: 168568949-168568949 on Build GRCh38
rs751015298 Chr.5: 168568724-168568724 on Build GRCh38
rs762171379 Chr.5: 168568747-168568747 on Build GRCh38
rs775759893 Chr.5: 168568853-168568853 on Build GRCh38
rs754618602 Chr.5: 168568950-168568950 on Build GRCh38
rs748027993 Chr.5: 168568694-168568694 on Build GRCh38
rs377051705 Chr.5: 168568598-168568598 on Build GRCh38
rs970466941 Chr.5: 168568843-168568843 on Build GRCh38
rs757936935 Chr.5: 168568621-168568621 on Build GRCh38
rs754690310 Chr.5: 168568881-168568881 on Build GRCh38
rs371166761 Chr.5: 168568904-168568904 on Build GRCh38
rs796390663 Chr.5: 168568566-168568566 on Build GRCh38
rs1049769899 Chr.5: 168568955-168568955 on Build GRCh38
rs376150396 Chr.5: 168568794-168568794 on Build GRCh38
rs750915592 Chr.5: 168568926-168568926 on Build GRCh38
rs748423483 Chr.5: 168568836-168568836 on Build GRCh38
rs764823569 Chr.5: 168568753-168568753 on Build GRCh38
rs754376364 Chr.5: 168568740-168568740 on Build GRCh38
rs934896272 Chr.5: 168568856-168568856 on Build GRCh38
rs914097183 Chr.5: 168568559-168568559 on Build GRCh38
rs781073428 Chr.5: 168568960-168568960 on Build GRCh38
rs762411357 Chr.5: 168568906-168568906 on Build GRCh38
rs780601525 Chr.5: 168568603-168568603 on Build GRCh38

View Full Product Details