Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 177434902-177435311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRK6
Entrez Gene ID
2870
Gene Name
G protein-coupled receptor kinase 6
Gene Aliases
GPRK6
UniGene
Hs.235116
Gene Chromosome Location
Chr.5: 177426496-177442849 on Build GRCh38
Transcripts
NM_001004105.2 NM_001004106.2 NM_002082.3 XM_006714858.3 XM_006714859.2 XM_011534537.2 XM_011534538.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1195888 c.1045G>A p.V349M Chr.5: 177435109-177435109 on Build GRCh38
7370322 c.1045G>A p.V349M Chr.5: 177435109-177435109 on Build GRCh38
1066380 c.981C>T p.I327I Chr.5: 177435045-177435045 on Build GRCh38
5789770 c.1044C>A p.T348T Chr.5: 177435108-177435108 on Build GRCh38
3661739 c.966C>T p.H322H Chr.5: 177434938-177434938 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1024118434 Chr.5: 177435182-177435182 on Build GRCh38
rs771486138 Chr.5: 177435001-177435001 on Build GRCh38
rs757778878 Chr.5: 177435169-177435169 on Build GRCh38
rs749635086 Chr.5: 177434958-177434958 on Build GRCh38
rs749724882 Chr.5: 177435088-177435088 on Build GRCh38
rs745447976 Chr.5: 177434995-177434995 on Build GRCh38
rs1050563848 Chr.5: 177435045-177435045 on Build GRCh38
rs778398556 Chr.5: 177435080-177435080 on Build GRCh38
rs560434892 Chr.5: 177435035-177435035 on Build GRCh38
rs150719729 Chr.5: 177435002-177435002 on Build GRCh38
rs200924933 Chr.5: 177435076-177435076 on Build GRCh38
rs774893545 Chr.5: 177435136-177435136 on Build GRCh38
rs768638203 Chr.5: 177435139-177435139 on Build GRCh38
rs773858585 Chr.5: 177434994-177434994 on Build GRCh38
rs867321732 Chr.5: 177435175-177435175 on Build GRCh38
rs1035172389 Chr.5: 177435256-177435256 on Build GRCh38
rs368737569 Chr.5: 177434950-177434950 on Build GRCh38
rs764205615 Chr.5: 177435159-177435160 on Build GRCh38
rs893765445 Chr.5: 177435138-177435138 on Build GRCh38
rs372214117 Chr.5: 177435009-177435009 on Build GRCh38
rs375225364 Chr.5: 177435041-177435041 on Build GRCh38
rs373013135 Chr.5: 177435078-177435078 on Build GRCh38
rs778316964 Chr.5: 177434952-177434952 on Build GRCh38
rs760627267 Chr.5: 177435003-177435003 on Build GRCh38
rs563835730 Chr.5: 177435288-177435288 on Build GRCh38
rs375782279 Chr.5: 177435150-177435150 on Build GRCh38
rs905459356 Chr.5: 177435025-177435025 on Build GRCh38
rs866525898 Chr.5: 177435218-177435218 on Build GRCh38
rs775666127 Chr.5: 177434967-177434971 on Build GRCh38
rs779166658 Chr.5: 177435172-177435172 on Build GRCh38
rs751736837 Chr.5: 177435058-177435058 on Build GRCh38
rs765231087 Chr.5: 177434956-177434957 on Build GRCh38
rs761583708 Chr.5: 177435019-177435019 on Build GRCh38
rs764957648 Chr.5: 177435144-177435144 on Build GRCh38
rs905301220 Chr.5: 177435200-177435200 on Build GRCh38
rs869164763 Chr.5: 177435080-177435080 on Build GRCh38
rs369754185 Chr.5: 177435124-177435124 on Build GRCh38
rs373368015 Chr.5: 177435135-177435135 on Build GRCh38
rs376452319 Chr.5: 177435109-177435109 on Build GRCh38
rs758264341 Chr.5: 177435004-177435004 on Build GRCh38
rs745518327 Chr.5: 177435128-177435128 on Build GRCh38
rs756246209 Chr.5: 177435154-177435154 on Build GRCh38
rs951761811 Chr.5: 177435290-177435290 on Build GRCh38
rs1033748230 Chr.5: 177435176-177435176 on Build GRCh38
rs920292266 Chr.5: 177434988-177434988 on Build GRCh38
rs754398191 Chr.5: 177435168-177435168 on Build GRCh38
rs962264842 Chr.5: 177435275-177435275 on Build GRCh38
rs971684526 Chr.5: 177434957-177434957 on Build GRCh38
rs773014143 Chr.5: 177435148-177435148 on Build GRCh38
rs761673596 Chr.5: 177435143-177435143 on Build GRCh38
rs752877386 Chr.5: 177435152-177435152 on Build GRCh38
rs199964997 Chr.5: 177434938-177434938 on Build GRCh38
rs1002762181 Chr.5: 177435042-177435042 on Build GRCh38
rs545424118 Chr.5: 177435251-177435251 on Build GRCh38
rs755009682 Chr.5: 177434927-177434927 on Build GRCh38
rs776459093 Chr.5: 177435012-177435012 on Build GRCh38
rs972757983 Chr.5: 177435285-177435285 on Build GRCh38
rs143935970 Chr.5: 177435064-177435064 on Build GRCh38
rs764761208 Chr.5: 177435163-177435163 on Build GRCh38
rs767520318 Chr.5: 177435075-177435075 on Build GRCh38
rs770753491 Chr.5: 177434989-177434989 on Build GRCh38
rs988555883 Chr.5: 177435201-177435201 on Build GRCh38
rs775005733 Chr.5: 177435002-177435002 on Build GRCh38
rs781268005 Chr.5: 177434929-177434929 on Build GRCh38

View Full Product Details