Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 179591480-179591918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RUFY1
Entrez Gene ID
80230
Gene Name
RUN and FYVE domain containing 1
Gene Aliases
RABIP4, ZFYVE12
UniGene
Hs.306769
Gene Chromosome Location
Chr.5: 179550551-179610026 on Build GRCh38
Transcripts
NM_001040451.2 NM_001040452.2 NM_025158.4 XM_005265993.4 XM_006714921.3 XM_006714922.3 XM_017009891.1 XM_017009892.1 XM_017009893.1 XM_017009894.1 XM_017009895.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3614757 c.903G>A p.E301E Chr.5: 179591723-179591723 on Build GRCh38
4777799 c.860_861insG p.L288fs*3 Chr.5: 179591680-179591681 on Build GRCh38
7402505 c.1153G>A p.E385K Chr.5: 179591649-179591649 on Build GRCh38
5403204 c.1243T>C p.L415L Chr.5: 179591739-179591739 on Build GRCh38
7402506 c.829G>A p.E277K Chr.5: 179591649-179591649 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771214954 Chr.5: 179591817-179591817 on Build GRCh38
rs779143872 Chr.5: 179591602-179591602 on Build GRCh38
rs750536760 Chr.5: 179591790-179591790 on Build GRCh38
rs768547016 Chr.5: 179591635-179591635 on Build GRCh38
rs544949239 Chr.5: 179591802-179591802 on Build GRCh38
rs752532292 Chr.5: 179591890-179591890 on Build GRCh38
rs761087357 Chr.5: 179591673-179591673 on Build GRCh38
rs199875147 Chr.5: 179591774-179591774 on Build GRCh38
rs142906534 Chr.5: 179591716-179591716 on Build GRCh38
rs776700678 Chr.5: 179591641-179591641 on Build GRCh38
rs750672564 Chr.5: 179591687-179591687 on Build GRCh38
rs904125990 Chr.5: 179591599-179591599 on Build GRCh38
rs760395789 Chr.5: 179591721-179591723 on Build GRCh38
rs533998425 Chr.5: 179591883-179591883 on Build GRCh38
rs779656343 Chr.5: 179591622-179591622 on Build GRCh38
rs981776047 Chr.5: 179591868-179591868 on Build GRCh38
rs1023350307 Chr.5: 179591825-179591825 on Build GRCh38
rs201900935 Chr.5: 179591610-179591610 on Build GRCh38
rs774031732 Chr.5: 179591780-179591780 on Build GRCh38
rs758541063 Chr.5: 179591553-179591553 on Build GRCh38
rs771914731 Chr.5: 179591683-179591683 on Build GRCh38
rs761910875 Chr.5: 179591683-179591683 on Build GRCh38
rs34184454 Chr.5: 179591876-179591877 on Build GRCh38
rs1037652192 Chr.5: 179591719-179591719 on Build GRCh38
rs1012401330 Chr.5: 179591569-179591569 on Build GRCh38
rs747712927 Chr.5: 179591643-179591643 on Build GRCh38
rs551011030 Chr.5: 179591787-179591787 on Build GRCh38
rs368351522 Chr.5: 179591758-179591758 on Build GRCh38
rs746972460 Chr.5: 179591633-179591633 on Build GRCh38
rs764633949 Chr.5: 179591674-179591674 on Build GRCh38
rs751372395 Chr.5: 179591705-179591705 on Build GRCh38
rs375449669 Chr.5: 179591785-179591785 on Build GRCh38
rs752842270 Chr.5: 179591779-179591779 on Build GRCh38
rs891377718 Chr.5: 179591874-179591874 on Build GRCh38
rs781321513 Chr.5: 179591736-179591736 on Build GRCh38
rs374747226 Chr.5: 179591892-179591892 on Build GRCh38
rs762413451 Chr.5: 179591531-179591531 on Build GRCh38
rs773017580 Chr.5: 179591648-179591648 on Build GRCh38
rs574366674 Chr.5: 179591580-179591580 on Build GRCh38
rs546955738 Chr.5: 179591511-179591511 on Build GRCh38
rs766148069 Chr.5: 179591759-179591759 on Build GRCh38
rs202117151 Chr.5: 179591772-179591772 on Build GRCh38
rs776401940 Chr.5: 179591776-179591777 on Build GRCh38
rs199499473 Chr.5: 179591868-179591869 on Build GRCh38
rs958860143 Chr.5: 179591811-179591811 on Build GRCh38
rs777350530 Chr.5: 179591762-179591762 on Build GRCh38
rs377338041 Chr.5: 179591782-179591782 on Build GRCh38
rs762453287 Chr.5: 179591649-179591649 on Build GRCh38
rs768967278 Chr.5: 179591783-179591783 on Build GRCh38
rs963756893 Chr.5: 179591747-179591747 on Build GRCh38
rs370666746 Chr.5: 179591820-179591820 on Build GRCh38
rs767925087 Chr.5: 179591667-179591667 on Build GRCh38
rs1007905989 Chr.5: 179591879-179591879 on Build GRCh38
rs75258376 Chr.5: 179591646-179591646 on Build GRCh38
rs1043915428 Chr.5: 179591858-179591858 on Build GRCh38
rs958890889 Chr.5: 179591585-179591585 on Build GRCh38
rs757303953 Chr.5: 179591600-179591600 on Build GRCh38
rs761397681 Chr.5: 179591625-179591626 on Build GRCh38
rs1009319031 Chr.5: 179591532-179591532 on Build GRCh38
rs758483065 Chr.5: 179591697-179591697 on Build GRCh38
rs745478094 Chr.5: 179591768-179591771 on Build GRCh38
rs982192406 Chr.5: 179591642-179591642 on Build GRCh38
rs573432318 Chr.5: 179591647-179591647 on Build GRCh38
rs564878803 Chr.5: 179591804-179591804 on Build GRCh38
rs967618154 Chr.5: 179591548-179591548 on Build GRCh38
rs949042359 Chr.5: 179591640-179591640 on Build GRCh38
rs758237659 Chr.5: 179591613-179591613 on Build GRCh38
rs191728471 Chr.5: 179591869-179591869 on Build GRCh38
rs903217283 Chr.5: 179591805-179591805 on Build GRCh38
rs990575143 Chr.5: 179591589-179591589 on Build GRCh38
rs773542482 Chr.5: 179591579-179591579 on Build GRCh38
rs111890912 Chr.5: 179591740-179591740 on Build GRCh38
rs368863668 Chr.5: 179591788-179591788 on Build GRCh38
rs769438894 Chr.5: 179591757-179591757 on Build GRCh38
rs754802602 Chr.5: 179591714-179591714 on Build GRCh38
rs148588709 Chr.5: 179591677-179591677 on Build GRCh38
rs753749975 Chr.5: 179591760-179591761 on Build GRCh38
rs557207431 Chr.5: 179591557-179591557 on Build GRCh38
rs772929671 Chr.5: 179591686-179591690 on Build GRCh38
rs575550007 Chr.5: 179591594-179591594 on Build GRCh38
rs186367492 Chr.5: 179591808-179591808 on Build GRCh38
rs765253999 Chr.5: 179591684-179591684 on Build GRCh38
rs898991577 Chr.5: 179591737-179591737 on Build GRCh38
rs753975092 Chr.5: 179591596-179591596 on Build GRCh38
rs748061274 Chr.5: 179591750-179591750 on Build GRCh38
rs780319201 Chr.5: 179591703-179591703 on Build GRCh38
rs970895965 Chr.5: 179591844-179591844 on Build GRCh38
rs916669669 Chr.5: 179591813-179591813 on Build GRCh38
rs1020503563 Chr.5: 179591539-179591539 on Build GRCh38
rs530460839 Chr.5: 179591807-179591807 on Build GRCh38

View Full Product Details