Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150492317-150492858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR6878
Entrez Gene ID
102465529
Gene Name
microRNA 6878
Gene Aliases
hsa-mir-6878
UniGene
Gene Chromosome Location
Chr.1: 150492345-150492410 on Build GRCh38
Transcripts
NR_106938.1 NM_001271895.1 NM_001271896.1 NM_025150.4 NR_073513.1 NR_073514.1 XM_006711555.1 XM_006711556.1 XM_017002394.1 XM_017002395.1

Gene Symbol
TARS2
Entrez Gene ID
80222
Gene Name
threonyl-tRNA synthetase 2, mitochondrial (putative)
Gene Aliases
COXPD21, TARSL1, thrRS
UniGene
Hs.288974
Gene Chromosome Location
Chr.1: 150487364-150507609 on Build GRCh38
Transcripts
NR_106938.1 NM_001271895.1 NM_001271896.1 NM_025150.4 NR_073513.1 NR_073514.1 XM_006711555.1 XM_006711556.1 XM_017002394.1 XM_017002395.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6227719 c.696-4C>A p.? Chr.1: 150492407-150492407 on Build GRCh38
6796887 c.738G>T p.R246R Chr.1: 150492453-150492453 on Build GRCh38
4388385 c.773C>T p.S258L Chr.1: 150492488-150492488 on Build GRCh38
5671936 c.705A>T p.T235T Chr.1: 150492420-150492420 on Build GRCh38
3473416 c.736C>T p.R246W Chr.1: 150492451-150492451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1046207397 Chr.1: 150492796-150492796 on Build GRCh38
rs113907888 Chr.1: 150492810-150492810 on Build GRCh38
rs988662212 Chr.1: 150492645-150492645 on Build GRCh38
rs777917633 Chr.1: 150492489-150492489 on Build GRCh38
rs985122583 Chr.1: 150492679-150492679 on Build GRCh38
rs587629898 Chr.1: 150492343-150492343 on Build GRCh38
rs184738664 Chr.1: 150492703-150492703 on Build GRCh38
rs375587671 Chr.1: 150492375-150492375 on Build GRCh38
rs796665206 Chr.1: 150492810-150492812 on Build GRCh38
rs779810930 Chr.1: 150492411-150492411 on Build GRCh38
rs766368590 Chr.1: 150492457-150492457 on Build GRCh38
rs80153934 Chr.1: 150492813-150492813 on Build GRCh38
rs953491566 Chr.1: 150492591-150492591 on Build GRCh38
rs771365307 Chr.1: 150492381-150492381 on Build GRCh38
rs905780049 Chr.1: 150492585-150492585 on Build GRCh38
rs978272185 Chr.1: 150492702-150492702 on Build GRCh38
rs752833046 Chr.1: 150492355-150492355 on Build GRCh38
rs145039072 Chr.1: 150492488-150492488 on Build GRCh38
rs1004951527 Chr.1: 150492831-150492831 on Build GRCh38
rs1035517576 Chr.1: 150492623-150492623 on Build GRCh38
rs781528342 Chr.1: 150492346-150492346 on Build GRCh38
rs910798250 Chr.1: 150492373-150492373 on Build GRCh38
rs751518235 Chr.1: 150492467-150492467 on Build GRCh38
rs368899093 Chr.1: 150492471-150492471 on Build GRCh38
rs1006142044 Chr.1: 150492532-150492532 on Build GRCh38
rs905890019 Chr.1: 150492770-150492770 on Build GRCh38
rs968489538 Chr.1: 150492660-150492660 on Build GRCh38
rs111351679 Chr.1: 150492809-150492809 on Build GRCh38
rs750302051 Chr.1: 150492809-150492810 on Build GRCh38
rs1022957990 Chr.1: 150492536-150492536 on Build GRCh38
rs372682193 Chr.1: 150492473-150492473 on Build GRCh38
rs779284538 Chr.1: 150492388-150492388 on Build GRCh38
rs375541864 Chr.1: 150492417-150492417 on Build GRCh38
rs916502709 Chr.1: 150492774-150492774 on Build GRCh38
rs987640186 Chr.1: 150492674-150492674 on Build GRCh38
rs587658021 Chr.1: 150492759-150492759 on Build GRCh38
rs963733614 Chr.1: 150492511-150492511 on Build GRCh38
rs762978927 Chr.1: 150492392-150492392 on Build GRCh38
rs749812543 Chr.1: 150492377-150492377 on Build GRCh38
rs587772810 Chr.1: 150492741-150492741 on Build GRCh38
rs767971079 Chr.1: 150492554-150492554 on Build GRCh38
rs1003227871 Chr.1: 150492616-150492616 on Build GRCh38
rs772543008 Chr.1: 150492534-150492534 on Build GRCh38
rs587646484 Chr.1: 150492801-150492801 on Build GRCh38
rs1047778755 Chr.1: 150492523-150492523 on Build GRCh38
rs780220587 Chr.1: 150492538-150492538 on Build GRCh38
rs923610175 Chr.1: 150492717-150492717 on Build GRCh38
rs1021345932 Chr.1: 150492646-150492646 on Build GRCh38
rs587720016 Chr.1: 150492833-150492833 on Build GRCh38
rs1053600341 Chr.1: 150492742-150492742 on Build GRCh38
rs910902241 Chr.1: 150492680-150492680 on Build GRCh38
rs113652065 Chr.1: 150492533-150492533 on Build GRCh38
rs887857228 Chr.1: 150492758-150492758 on Build GRCh38
rs757786376 Chr.1: 150492525-150492525 on Build GRCh38
rs887862316 Chr.1: 150492527-150492527 on Build GRCh38
rs375868769 Chr.1: 150492359-150492359 on Build GRCh38
rs111842933 Chr.1: 150492795-150492795 on Build GRCh38
rs763015481 Chr.1: 150492451-150492451 on Build GRCh38
rs943619909 Chr.1: 150492684-150492684 on Build GRCh38
rs373434114 Chr.1: 150492406-150492406 on Build GRCh38
rs587721411 Chr.1: 150492740-150492740 on Build GRCh38
rs753878517 Chr.1: 150492521-150492521 on Build GRCh38
rs772781562 Chr.1: 150492441-150492441 on Build GRCh38
rs923564200 Chr.1: 150492736-150492736 on Build GRCh38
rs956039476 Chr.1: 150492639-150492639 on Build GRCh38
rs148372463 Chr.1: 150492765-150492765 on Build GRCh38
rs182013142 Chr.1: 150492437-150492437 on Build GRCh38
rs746185664 Chr.1: 150492396-150492396 on Build GRCh38
rs587765665 Chr.1: 150492486-150492486 on Build GRCh38
rs189143746 Chr.1: 150492778-150492778 on Build GRCh38

View Full Product Details