Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150764549-150765048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSS
Entrez Gene ID
1520
Gene Name
cathepsin S
Gene Aliases
-
UniGene
Hs.181301
Gene Chromosome Location
Chr.1: 150730196-150765957 on Build GRCh38
Transcripts
NM_001199739.1 NM_004079.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4504237 c.65C>T p.P22L Chr.1: 150764699-150764699 on Build GRCh38
4651950 c.25T>G p.L9V Chr.1: 150764739-150764739 on Build GRCh38
222554 c.46G>A p.A16T Chr.1: 150764718-150764718 on Build GRCh38
1688502 c.121G>A p.E41K Chr.1: 150764643-150764643 on Build GRCh38
1333875 c.116A>G p.Y39C Chr.1: 150764648-150764648 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768934133 Chr.1: 150764708-150764708 on Build GRCh38
rs761829673 Chr.1: 150764683-150764683 on Build GRCh38
rs368866295 Chr.1: 150764713-150764713 on Build GRCh38
rs748138411 Chr.1: 150764750-150764750 on Build GRCh38
rs980241408 Chr.1: 150764843-150764843 on Build GRCh38
rs587652178 Chr.1: 150764892-150764892 on Build GRCh38
rs749411033 Chr.1: 150764622-150764622 on Build GRCh38
rs145551238 Chr.1: 150764680-150764680 on Build GRCh38
rs754982274 Chr.1: 150764793-150764793 on Build GRCh38
rs780325618 Chr.1: 150764698-150764698 on Build GRCh38
rs190922865 Chr.1: 150764866-150764866 on Build GRCh38
rs763627430 Chr.1: 150764654-150764654 on Build GRCh38
rs749728922 Chr.1: 150764778-150764778 on Build GRCh38
rs973136401 Chr.1: 150764966-150764966 on Build GRCh38
rs897795040 Chr.1: 150764790-150764790 on Build GRCh38
rs775268813 Chr.1: 150764630-150764630 on Build GRCh38
rs751778908 Chr.1: 150764721-150764721 on Build GRCh38
rs56231712 Chr.1: 150764615-150764615 on Build GRCh38
rs375149139 Chr.1: 150764592-150764592 on Build GRCh38
rs746649398 Chr.1: 150764813-150764813 on Build GRCh38
rs374352444 Chr.1: 150764890-150764890 on Build GRCh38
rs771192821 Chr.1: 150764684-150764684 on Build GRCh38
rs771417785 Chr.1: 150764785-150764785 on Build GRCh38
rs758127677 Chr.1: 150764697-150764697 on Build GRCh38
rs1012737632 Chr.1: 150764951-150764951 on Build GRCh38
rs747069060 Chr.1: 150764609-150764609 on Build GRCh38
rs763344176 Chr.1: 150764619-150764619 on Build GRCh38
rs199930336 Chr.1: 150764738-150764738 on Build GRCh38
rs369740966 Chr.1: 150764691-150764691 on Build GRCh38
rs202057733 Chr.1: 150764768-150764768 on Build GRCh38
rs1048511326 Chr.1: 150764574-150764574 on Build GRCh38
rs200747061 Chr.1: 150764612-150764612 on Build GRCh38
rs765117044 Chr.1: 150764688-150764688 on Build GRCh38
rs989988696 Chr.1: 150764693-150764693 on Build GRCh38
rs762156019 Chr.1: 150765007-150765007 on Build GRCh38
rs587595911 Chr.1: 150764666-150764666 on Build GRCh38
rs760168955 Chr.1: 150764634-150764634 on Build GRCh38
rs773745475 Chr.1: 150764618-150764618 on Build GRCh38
rs41271951 Chr.1: 150764744-150764744 on Build GRCh38
rs56404059 Chr.1: 150764614-150764614 on Build GRCh38
rs930559315 Chr.1: 150764888-150764888 on Build GRCh38
rs755064936 Chr.1: 150764742-150764742 on Build GRCh38
rs944738891 Chr.1: 150764645-150764645 on Build GRCh38
rs775044847 Chr.1: 150764796-150764796 on Build GRCh38
rs587754419 Chr.1: 150764889-150764889 on Build GRCh38
rs146441878 Chr.1: 150764757-150764757 on Build GRCh38
rs915425054 Chr.1: 150764582-150764582 on Build GRCh38

View Full Product Details