Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 10750991-10751505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM14B
Entrez Gene ID
81853
Gene Name
transmembrane protein 14B
Gene Aliases
-
UniGene
Hs.667371
Gene Chromosome Location
Chr.6: 10747759-10759774 on Build GRCh38
Transcripts
NM_001127711.2 NM_001286484.1 NM_001286488.1 NM_001286489.1 NM_030969.4 NR_104454.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1071821 c.105C>T p.S35S Chr.6: 10751137-10751137 on Build GRCh38
3619265 c.158C>T p.A53V Chr.6: 10751190-10751190 on Build GRCh38
3619266 c.175G>A p.D59N Chr.6: 10751207-10751207 on Build GRCh38
7394152 c.148G>A p.G50S Chr.6: 10751180-10751180 on Build GRCh38
3429827 c.154G>A p.G52S Chr.6: 10751186-10751186 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891804370 Chr.6: 10751460-10751460 on Build GRCh38
rs370771146 Chr.6: 10751117-10751117 on Build GRCh38
rs767381353 Chr.6: 10751184-10751184 on Build GRCh38
rs764921030 Chr.6: 10751183-10751183 on Build GRCh38
rs201494277 Chr.6: 10751270-10751270 on Build GRCh38
rs780895464 Chr.6: 10751177-10751177 on Build GRCh38
rs986465785 Chr.6: 10751157-10751157 on Build GRCh38
rs776144975 Chr.6: 10751179-10751179 on Build GRCh38
rs909299168 Chr.6: 10751045-10751046 on Build GRCh38
rs946084348 Chr.6: 10751471-10751471 on Build GRCh38
rs746243542 Chr.6: 10751086-10751086 on Build GRCh38
rs761807406 Chr.6: 10751199-10751199 on Build GRCh38
rs750544190 Chr.6: 10751269-10751269 on Build GRCh38
rs140364465 Chr.6: 10751116-10751116 on Build GRCh38
rs745694210 Chr.6: 10751177-10751180 on Build GRCh38
rs1019288812 Chr.6: 10751288-10751288 on Build GRCh38
rs1054028277 Chr.6: 10751300-10751300 on Build GRCh38
rs143782354 Chr.6: 10751208-10751208 on Build GRCh38
rs556847009 Chr.6: 10751265-10751265 on Build GRCh38
rs780805530 Chr.6: 10751225-10751225 on Build GRCh38
rs775853122 Chr.6: 10751091-10751091 on Build GRCh38
rs558104171 Chr.6: 10751343-10751343 on Build GRCh38
rs557127750 Chr.6: 10751042-10751042 on Build GRCh38
rs980294264 Chr.6: 10751340-10751340 on Build GRCh38
rs145552498 Chr.6: 10751204-10751204 on Build GRCh38
rs754222551 Chr.6: 10751218-10751218 on Build GRCh38
rs138792128 Chr.6: 10751143-10751143 on Build GRCh38
rs749850149 Chr.6: 10751092-10751092 on Build GRCh38
rs1015495345 Chr.6: 10751462-10751462 on Build GRCh38
rs539993092 Chr.6: 10751331-10751331 on Build GRCh38
rs548560408 Chr.6: 10751255-10751255 on Build GRCh38
rs149369273 Chr.6: 10751145-10751145 on Build GRCh38
rs772748063 Chr.6: 10751099-10751099 on Build GRCh38
rs770548943 Chr.6: 10751174-10751174 on Build GRCh38
rs767689842 Chr.6: 10751202-10751202 on Build GRCh38
rs527428434 Chr.6: 10751254-10751254 on Build GRCh38
rs142679222 Chr.6: 10751445-10751445 on Build GRCh38
rs764241832 Chr.6: 10751127-10751127 on Build GRCh38
rs891852568 Chr.6: 10751234-10751234 on Build GRCh38
rs1005502855 Chr.6: 10751461-10751461 on Build GRCh38
rs748535386 Chr.6: 10751112-10751112 on Build GRCh38
rs184489994 Chr.6: 10751262-10751262 on Build GRCh38
rs9689171 Chr.6: 10751047-10751047 on Build GRCh38
rs562421362 Chr.6: 10751295-10751295 on Build GRCh38
rs546089107 Chr.6: 10751078-10751078 on Build GRCh38
rs751636879 Chr.6: 10751198-10751198 on Build GRCh38
rs762859199 Chr.6: 10751251-10751251 on Build GRCh38
rs1033571964 Chr.6: 10751017-10751017 on Build GRCh38
rs898717592 Chr.6: 10751025-10751025 on Build GRCh38
rs17849403 Chr.6: 10751156-10751156 on Build GRCh38
rs763858850 Chr.6: 10751341-10751341 on Build GRCh38
rs1046584254 Chr.6: 10751373-10751373 on Build GRCh38
rs146042312 Chr.6: 10751137-10751137 on Build GRCh38
rs919707746 Chr.6: 10751306-10751306 on Build GRCh38
rs374886344 Chr.6: 10751421-10751421 on Build GRCh38
rs972169255 Chr.6: 10751071-10751071 on Build GRCh38
rs745480599 Chr.6: 10751226-10751226 on Build GRCh38
rs752670776 Chr.6: 10751122-10751122 on Build GRCh38
rs148467156 Chr.6: 10751357-10751357 on Build GRCh38
rs149683885 Chr.6: 10751224-10751224 on Build GRCh38
rs769604871 Chr.6: 10751237-10751237 on Build GRCh38
rs575575932 Chr.6: 10751038-10751038 on Build GRCh38
rs151307186 Chr.6: 10751219-10751219 on Build GRCh38
rs139370420 Chr.6: 10751221-10751221 on Build GRCh38
rs781744833 Chr.6: 10751088-10751088 on Build GRCh38
rs760279788 Chr.6: 10751107-10751107 on Build GRCh38
rs146152247 Chr.6: 10751180-10751180 on Build GRCh38
rs370938546 Chr.6: 10751223-10751223 on Build GRCh38
rs746535310 Chr.6: 10751171-10751171 on Build GRCh38
rs1036175721 Chr.6: 10751100-10751100 on Build GRCh38
rs961382617 Chr.6: 10751292-10751292 on Build GRCh38
rs1028190937 Chr.6: 10751257-10751257 on Build GRCh38
rs371666146 Chr.6: 10751093-10751093 on Build GRCh38
rs1021172600 Chr.6: 10751243-10751243 on Build GRCh38
rs1033382305 Chr.6: 10751450-10751450 on Build GRCh38
rs764151993 Chr.6: 10751194-10751194 on Build GRCh38
rs1046427 Chr.6: 10751146-10751146 on Build GRCh38
rs757456330 Chr.6: 10751138-10751138 on Build GRCh38
rs115801751 Chr.6: 10751167-10751167 on Build GRCh38
rs143628480 Chr.6: 10751131-10751131 on Build GRCh38
rs566539453 Chr.6: 10751250-10751250 on Build GRCh38
rs762946624 Chr.6: 10751192-10751192 on Build GRCh38
rs554783546 Chr.6: 10751142-10751142 on Build GRCh38
rs931460694 Chr.6: 10751310-10751310 on Build GRCh38
rs769181586 Chr.6: 10751162-10751162 on Build GRCh38
rs17856203 Chr.6: 10751141-10751141 on Build GRCh38
rs193155234 Chr.6: 10751158-10751158 on Build GRCh38
rs199717445 Chr.6: 10751205-10751205 on Build GRCh38
rs1057434600 Chr.6: 10751457-10751457 on Build GRCh38
rs952534993 Chr.6: 10751029-10751030 on Build GRCh38
rs775467044 Chr.6: 10751187-10751187 on Build GRCh38
rs765967223 Chr.6: 10751111-10751111 on Build GRCh38
rs758460139 Chr.6: 10751125-10751125 on Build GRCh38

View Full Product Details