Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 11576928-11577444 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM170B
Entrez Gene ID
100113407
Gene Name
transmembrane protein 170B
Gene Aliases
-
UniGene
Hs.146317
Gene Chromosome Location
Chr.6: 11538042-11583524 on Build GRCh38
Transcripts
NM_001100829.2

SNP Detail

SNP ID Chromosome Location
rs12212790 Chr.6: 11577233-11577233 on Build GRCh38
rs923935672 Chr.6: 11576989-11576989 on Build GRCh38
rs990333500 Chr.6: 11577097-11577097 on Build GRCh38
rs954438755 Chr.6: 11576962-11576962 on Build GRCh38
rs886737411 Chr.6: 11577251-11577251 on Build GRCh38
rs1017662496 Chr.6: 11577370-11577370 on Build GRCh38
rs1005242946 Chr.6: 11577280-11577280 on Build GRCh38
rs1040261774 Chr.6: 11577100-11577100 on Build GRCh38
rs893910358 Chr.6: 11577070-11577070 on Build GRCh38
rs556065033 Chr.6: 11577354-11577354 on Build GRCh38
rs998839791 Chr.6: 11577185-11577185 on Build GRCh38
rs1029963887 Chr.6: 11576954-11576954 on Build GRCh38
rs986180345 Chr.6: 11576967-11576967 on Build GRCh38
rs763930097 Chr.6: 11576956-11576956 on Build GRCh38
rs993285382 Chr.6: 11577243-11577244 on Build GRCh38
rs901363243 Chr.6: 11577179-11577179 on Build GRCh38
rs112555280 Chr.6: 11577001-11577001 on Build GRCh38
rs897631061 Chr.6: 11577240-11577240 on Build GRCh38
rs928707932 Chr.6: 11577160-11577160 on Build GRCh38
rs991280066 Chr.6: 11577402-11577402 on Build GRCh38
rs753659718 Chr.6: 11577188-11577188 on Build GRCh38
rs1041858354 Chr.6: 11577152-11577152 on Build GRCh38
rs1006071835 Chr.6: 11577231-11577231 on Build GRCh38
rs942852723 Chr.6: 11577094-11577094 on Build GRCh38
rs374317077 Chr.6: 11577414-11577414 on Build GRCh38
rs914703250 Chr.6: 11577099-11577099 on Build GRCh38
rs954597044 Chr.6: 11577327-11577327 on Build GRCh38
rs935341803 Chr.6: 11576998-11576998 on Build GRCh38
rs554341329 Chr.6: 11577008-11577008 on Build GRCh38
rs114100366 Chr.6: 11576970-11576970 on Build GRCh38
rs1053758379 Chr.6: 11577051-11577051 on Build GRCh38
rs911504534 Chr.6: 11576968-11576969 on Build GRCh38
rs946121196 Chr.6: 11577128-11577128 on Build GRCh38
rs556945247 Chr.6: 11577393-11577393 on Build GRCh38
rs756809667 Chr.6: 11577319-11577319 on Build GRCh38
rs938719405 Chr.6: 11577163-11577163 on Build GRCh38
rs12211376 Chr.6: 11577339-11577339 on Build GRCh38

View Full Product Details