Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 15248776-15249281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JARID2
Entrez Gene ID
3720
Gene Name
jumonji and AT-rich interaction domain containing 2
Gene Aliases
JMJ
UniGene
Hs.269059
Gene Chromosome Location
Chr.6: 15245975-15522042 on Build GRCh38
Transcripts
NM_001267040.1 NM_004973.3 XM_005249089.3 XM_011514579.2 XM_017010833.1 XM_017010834.1

SNP Detail

SNP ID Chromosome Location
rs575352125 Chr.6: 15248952-15248952 on Build GRCh38
rs897110527 Chr.6: 15248838-15248838 on Build GRCh38
rs991436660 Chr.6: 15249134-15249134 on Build GRCh38
rs928111389 Chr.6: 15248799-15248799 on Build GRCh38
rs989217400 Chr.6: 15248830-15248830 on Build GRCh38
rs903447849 Chr.6: 15248947-15248947 on Build GRCh38
rs942629060 Chr.6: 15249231-15249231 on Build GRCh38
rs762852982 Chr.6: 15249016-15249016 on Build GRCh38
rs999029837 Chr.6: 15248986-15248986 on Build GRCh38
rs1051327892 Chr.6: 15248924-15248924 on Build GRCh38
rs962466047 Chr.6: 15249166-15249166 on Build GRCh38
rs1055261713 Chr.6: 15248806-15248806 on Build GRCh38
rs1007655877 Chr.6: 15248848-15248848 on Build GRCh38
rs1020992105 Chr.6: 15249035-15249035 on Build GRCh38
rs998044279 Chr.6: 15248839-15248839 on Build GRCh38
rs986743650 Chr.6: 15249228-15249228 on Build GRCh38
rs572926824 Chr.6: 15248885-15248885 on Build GRCh38
rs552182148 Chr.6: 15249104-15249104 on Build GRCh38
rs767538836 Chr.6: 15249246-15249246 on Build GRCh38
rs1051424064 Chr.6: 15248957-15248957 on Build GRCh38
rs1036683651 Chr.6: 15248911-15248911 on Build GRCh38
rs1020960835 Chr.6: 15248866-15248866 on Build GRCh38
rs981940689 Chr.6: 15249010-15249010 on Build GRCh38
rs781568584 Chr.6: 15249237-15249237 on Build GRCh38
rs79327808 Chr.6: 15249009-15249009 on Build GRCh38
rs545791210 Chr.6: 15249197-15249197 on Build GRCh38
rs1036651202 Chr.6: 15248831-15248831 on Build GRCh38
rs1040225859 Chr.6: 15249090-15249090 on Build GRCh38
rs541548601 Chr.6: 15248912-15248912 on Build GRCh38
rs561962851 Chr.6: 15248923-15248923 on Build GRCh38
rs1050966645 Chr.6: 15248840-15248840 on Build GRCh38
rs933612582 Chr.6: 15249214-15249214 on Build GRCh38
rs187286191 Chr.6: 15249019-15249019 on Build GRCh38
rs918674917 Chr.6: 15248917-15248929 on Build GRCh38
rs568385611 Chr.6: 15248845-15248845 on Build GRCh38
rs958028088 Chr.6: 15248855-15248855 on Build GRCh38
rs1010951951 Chr.6: 15248861-15248861 on Build GRCh38
rs1047098928 Chr.6: 15248859-15248859 on Build GRCh38
rs972429313 Chr.6: 15249181-15249181 on Build GRCh38
rs951287841 Chr.6: 15249014-15249014 on Build GRCh38
rs941105249 Chr.6: 15248910-15248910 on Build GRCh38
rs532124831 Chr.6: 15249040-15249040 on Build GRCh38
rs1054312830 Chr.6: 15249233-15249233 on Build GRCh38
rs982337189 Chr.6: 15248796-15248796 on Build GRCh38
rs1007096413 Chr.6: 15248997-15248997 on Build GRCh38
rs190969949 Chr.6: 15249060-15249060 on Build GRCh38
rs374161632 Chr.6: 15249160-15249162 on Build GRCh38
rs928174872 Chr.6: 15248876-15248876 on Build GRCh38
rs889147176 Chr.6: 15248821-15248821 on Build GRCh38
rs867014290 Chr.6: 15249099-15249099 on Build GRCh38
rs879831082 Chr.6: 15248879-15248879 on Build GRCh38
rs893492105 Chr.6: 15249199-15249199 on Build GRCh38
rs990585008 Chr.6: 15248883-15248883 on Build GRCh38
rs1011096992 Chr.6: 15249001-15249001 on Build GRCh38
rs1019341071 Chr.6: 15248847-15248847 on Build GRCh38
rs966670945 Chr.6: 15249053-15249053 on Build GRCh38
rs937800835 Chr.6: 15248803-15248803 on Build GRCh38
rs1008883396 Chr.6: 15248844-15248844 on Build GRCh38
rs927744973 Chr.6: 15249011-15249011 on Build GRCh38
rs751515339 Chr.6: 15249202-15249202 on Build GRCh38
rs930001574 Chr.6: 15248858-15248858 on Build GRCh38
rs910037195 Chr.6: 15249141-15249141 on Build GRCh38
rs911162596 Chr.6: 15249229-15249229 on Build GRCh38
rs1031098235 Chr.6: 15248991-15248991 on Build GRCh38
rs1035638936 Chr.6: 15249091-15249091 on Build GRCh38
rs745917299 Chr.6: 15249248-15249248 on Build GRCh38
rs533028626 Chr.6: 15249065-15249065 on Build GRCh38
rs909565670 Chr.6: 15248817-15248817 on Build GRCh38
rs757700711 Chr.6: 15249171-15249171 on Build GRCh38
rs889635145 Chr.6: 15248970-15248970 on Build GRCh38
rs1051212378 Chr.6: 15249157-15249157 on Build GRCh38
rs1023373165 Chr.6: 15249002-15249002 on Build GRCh38
rs548926032 Chr.6: 15248805-15248805 on Build GRCh38
rs1054294004 Chr.6: 15249003-15249003 on Build GRCh38
rs958035992 Chr.6: 15248828-15248828 on Build GRCh38
rs982754441 Chr.6: 15248874-15248874 on Build GRCh38
rs757184356 Chr.6: 15249236-15249236 on Build GRCh38
rs528626947 Chr.6: 15249128-15249128 on Build GRCh38
rs180858549 Chr.6: 15249125-15249125 on Build GRCh38
rs921354742 Chr.6: 15249132-15249132 on Build GRCh38
rs1018070773 Chr.6: 15248854-15248854 on Build GRCh38
rs946794286 Chr.6: 15249242-15249242 on Build GRCh38
rs959778413 Chr.6: 15248882-15248882 on Build GRCh38
rs763933025 Chr.6: 15249083-15249083 on Build GRCh38
rs966982254 Chr.6: 15248869-15248869 on Build GRCh38
rs868421663 Chr.6: 15249044-15249044 on Build GRCh38
rs903291057 Chr.6: 15248851-15248851 on Build GRCh38

View Full Product Details