Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 38737454-38737987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH8
Entrez Gene ID
1769
Gene Name
dynein axonemal heavy chain 8
Gene Aliases
ATPase, hdhc9
UniGene
Hs.520106
Gene Chromosome Location
Chr.6: 38715329-39030798 on Build GRCh38
Transcripts
NM_001206927.1 XM_011514318.2 XM_011514319.2 XM_011514320.2 XM_011514321.1 XM_017010325.1 XM_017010326.1 XM_017010327.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3626505 c.974G>A p.G325E Chr.6: 38737830-38737830 on Build GRCh38
3430600 c.1069C>T p.H357Y Chr.6: 38737925-38737925 on Build GRCh38
7150132 c.1089G>C p.L363L Chr.6: 38737945-38737945 on Build GRCh38
7150130 c.438G>C p.L146L Chr.6: 38737945-38737945 on Build GRCh38
3430598 c.418C>T p.H140Y Chr.6: 38737925-38737925 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs137954757 Chr.6: 38737884-38737884 on Build GRCh38
rs1024254012 Chr.6: 38737560-38737560 on Build GRCh38
rs762501541 Chr.6: 38737907-38737907 on Build GRCh38
rs975534140 Chr.6: 38737737-38737737 on Build GRCh38
rs763828744 Chr.6: 38737826-38737826 on Build GRCh38
rs751210553 Chr.6: 38737912-38737912 on Build GRCh38
rs773448341 Chr.6: 38737874-38737874 on Build GRCh38
rs757946681 Chr.6: 38737814-38737814 on Build GRCh38
rs756876172 Chr.6: 38737923-38737923 on Build GRCh38
rs962690096 Chr.6: 38737688-38737688 on Build GRCh38
rs187527824 Chr.6: 38737614-38737614 on Build GRCh38
rs912166280 Chr.6: 38737753-38737753 on Build GRCh38
rs770743086 Chr.6: 38737880-38737880 on Build GRCh38
rs533910714 Chr.6: 38737765-38737765 on Build GRCh38
rs1008223012 Chr.6: 38737508-38737508 on Build GRCh38
rs972068548 Chr.6: 38737803-38737803 on Build GRCh38
rs764779469 Chr.6: 38737807-38737807 on Build GRCh38
rs548839228 Chr.6: 38737545-38737545 on Build GRCh38
rs920114494 Chr.6: 38737624-38737624 on Build GRCh38
rs930430078 Chr.6: 38737521-38737521 on Build GRCh38
rs143492695 Chr.6: 38737847-38737847 on Build GRCh38
rs897917214 Chr.6: 38737517-38737517 on Build GRCh38
rs376791268 Chr.6: 38737938-38737938 on Build GRCh38
rs755432086 Chr.6: 38737945-38737945 on Build GRCh38
rs747565030 Chr.6: 38737767-38737770 on Build GRCh38
rs571981683 Chr.6: 38737686-38737686 on Build GRCh38
rs528999541 Chr.6: 38737487-38737487 on Build GRCh38
rs1011492271 Chr.6: 38737565-38737565 on Build GRCh38
rs765294651 Chr.6: 38737900-38737900 on Build GRCh38
rs57772294 Chr.6: 38737610-38737610 on Build GRCh38
rs1013232065 Chr.6: 38737556-38737556 on Build GRCh38
rs908833654 Chr.6: 38737714-38737716 on Build GRCh38
rs9470924 Chr.6: 38737906-38737906 on Build GRCh38
rs759601727 Chr.6: 38737800-38737800 on Build GRCh38
rs200424041 Chr.6: 38737828-38737828 on Build GRCh38
rs748254078 Chr.6: 38737870-38737870 on Build GRCh38
rs1022880562 Chr.6: 38737683-38737683 on Build GRCh38
rs183395573 Chr.6: 38737481-38737481 on Build GRCh38
rs554151668 Chr.6: 38737772-38737772 on Build GRCh38
rs986761290 Chr.6: 38737721-38737721 on Build GRCh38
rs747257406 Chr.6: 38737877-38737877 on Build GRCh38
rs149280386 Chr.6: 38737837-38737837 on Build GRCh38
rs759859299 Chr.6: 38737940-38737940 on Build GRCh38
rs772095746 Chr.6: 38737872-38737872 on Build GRCh38
rs142065001 Chr.6: 38737951-38737951 on Build GRCh38
rs899809874 Chr.6: 38737824-38737824 on Build GRCh38
rs967278101 Chr.6: 38737575-38737575 on Build GRCh38
rs997322536 Chr.6: 38737549-38737549 on Build GRCh38
rs887113505 Chr.6: 38737503-38737503 on Build GRCh38
rs766123131 Chr.6: 38737790-38737790 on Build GRCh38
rs771138717 Chr.6: 38737791-38737792 on Build GRCh38
rs772838378 Chr.6: 38737768-38737768 on Build GRCh38
rs552095279 Chr.6: 38737662-38737662 on Build GRCh38
rs763579484 Chr.6: 38737911-38737911 on Build GRCh38
rs998494936 Chr.6: 38737573-38737573 on Build GRCh38
rs748882288 Chr.6: 38737933-38737933 on Build GRCh38
rs143368533 Chr.6: 38737892-38737892 on Build GRCh38
rs756498084 Chr.6: 38737830-38737830 on Build GRCh38
rs981131720 Chr.6: 38737578-38737578 on Build GRCh38
rs567584262 Chr.6: 38737809-38737809 on Build GRCh38
rs781671929 Chr.6: 38737856-38737857 on Build GRCh38
rs111566149 Chr.6: 38737863-38737863 on Build GRCh38
rs1048918972 Chr.6: 38737541-38737541 on Build GRCh38
rs374681790 Chr.6: 38737855-38737855 on Build GRCh38
rs373387718 Chr.6: 38737507-38737507 on Build GRCh38
rs1047727764 Chr.6: 38737484-38737484 on Build GRCh38
rs928265697 Chr.6: 38737599-38737599 on Build GRCh38
rs989891532 Chr.6: 38737628-38737628 on Build GRCh38
rs906833670 Chr.6: 38737566-38737566 on Build GRCh38
rs780454556 Chr.6: 38737842-38737842 on Build GRCh38
rs368108107 Chr.6: 38737792-38737792 on Build GRCh38
rs953593744 Chr.6: 38737690-38737690 on Build GRCh38

View Full Product Details