Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41159650-41160146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC105375056
Entrez Gene ID
105375056
Gene Name
uncharacterized LOC105375056
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.6: 41154028-41178568 on Build GRCh38
Transcripts
NM_001271821.1 NM_018965.3 XM_006715116.3

Gene Symbol
TREM2
Entrez Gene ID
54209
Gene Name
triggering receptor expressed on myeloid cells 2
Gene Aliases
TREM-2, Trem2a, Trem2b, Trem2c
UniGene
Hs.435295
Gene Chromosome Location
Chr.6: 41158506-41163200 on Build GRCh38
Transcripts
NM_001271821.1 NM_018965.3 XM_006715116.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4425552 c.397C>G p.L133V Chr.6: 41159877-41159877 on Build GRCh38
6807623 c.439T>C p.S147P Chr.6: 41159835-41159835 on Build GRCh38
5540763 c.406C>T p.R136W Chr.6: 41159868-41159868 on Build GRCh38
3078388 c.449T>A p.F150Y Chr.6: 41159825-41159825 on Build GRCh38
5486562 c.482+2T>C p.? Chr.6: 41159790-41159790 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546844731 Chr.6: 41159797-41159797 on Build GRCh38
rs867253281 Chr.6: 41159828-41159828 on Build GRCh38
rs766304941 Chr.6: 41159893-41159893 on Build GRCh38
rs764489173 Chr.6: 41159800-41159800 on Build GRCh38
rs768123358 Chr.6: 41159885-41159885 on Build GRCh38
rs778867308 Chr.6: 41159731-41159731 on Build GRCh38
rs897508772 Chr.6: 41160103-41160103 on Build GRCh38
rs994465122 Chr.6: 41160008-41160008 on Build GRCh38
rs1007861089 Chr.6: 41160027-41160027 on Build GRCh38
rs756126666 Chr.6: 41159839-41159839 on Build GRCh38
rs187905635 Chr.6: 41160091-41160091 on Build GRCh38
rs756430557 Chr.6: 41159748-41159748 on Build GRCh38
rs532907021 Chr.6: 41160110-41160110 on Build GRCh38
rs761969647 Chr.6: 41159857-41159858 on Build GRCh38
rs757420471 Chr.6: 41159742-41159742 on Build GRCh38
rs766647311 Chr.6: 41159841-41159841 on Build GRCh38
rs139397773 Chr.6: 41159845-41159845 on Build GRCh38
rs574310382 Chr.6: 41159959-41159959 on Build GRCh38
rs528648042 Chr.6: 41159676-41159676 on Build GRCh38
rs147769114 Chr.6: 41159925-41159925 on Build GRCh38
rs79011726 Chr.6: 41159823-41159823 on Build GRCh38
rs747924604 Chr.6: 41159857-41159857 on Build GRCh38
rs749132016 Chr.6: 41159763-41159763 on Build GRCh38
rs386834144 Chr.6: 41159790-41159790 on Build GRCh38
rs1026325039 Chr.6: 41160017-41160017 on Build GRCh38
rs773545966 Chr.6: 41159886-41159886 on Build GRCh38
rs758428150 Chr.6: 41159865-41159865 on Build GRCh38
rs577138928 Chr.6: 41159946-41159946 on Build GRCh38
rs554863014 Chr.6: 41159902-41159902 on Build GRCh38
rs117151816 Chr.6: 41160109-41160109 on Build GRCh38
rs950036221 Chr.6: 41159947-41159947 on Build GRCh38
rs139607688 Chr.6: 41159881-41159881 on Build GRCh38
rs754069674 Chr.6: 41159900-41159900 on Build GRCh38
rs530672802 Chr.6: 41159743-41159743 on Build GRCh38
rs772641807 Chr.6: 41159868-41159868 on Build GRCh38
rs762052858 Chr.6: 41159882-41159882 on Build GRCh38
rs774726836 Chr.6: 41159846-41159846 on Build GRCh38
rs2234254 Chr.6: 41159955-41159955 on Build GRCh38
rs555483777 Chr.6: 41159728-41159728 on Build GRCh38
rs780108939 Chr.6: 41159760-41159760 on Build GRCh38
rs963786962 Chr.6: 41160050-41160050 on Build GRCh38
rs934622149 Chr.6: 41160012-41160012 on Build GRCh38
rs182653531 Chr.6: 41160048-41160048 on Build GRCh38
rs890766240 Chr.6: 41160023-41160023 on Build GRCh38
rs985980430 Chr.6: 41159928-41159928 on Build GRCh38
rs778620014 Chr.6: 41159850-41159850 on Build GRCh38
rs1014380328 Chr.6: 41159732-41159732 on Build GRCh38
rs1053146510 Chr.6: 41159711-41159711 on Build GRCh38
rs1027858046 Chr.6: 41160108-41160108 on Build GRCh38
rs765674379 Chr.6: 41160074-41160074 on Build GRCh38
rs767909449 Chr.6: 41159824-41159824 on Build GRCh38
rs796448179 Chr.6: 41160075-41160075 on Build GRCh38
rs905993798 Chr.6: 41159738-41159738 on Build GRCh38
rs748010918 Chr.6: 41159776-41159776 on Build GRCh38
rs570462504 Chr.6: 41159784-41159784 on Build GRCh38
rs565823447 Chr.6: 41159949-41159949 on Build GRCh38
rs754022211 Chr.6: 41159842-41159842 on Build GRCh38
rs759809382 Chr.6: 41159921-41159921 on Build GRCh38
rs201273800 Chr.6: 41159924-41159924 on Build GRCh38
rs948937763 Chr.6: 41160066-41160066 on Build GRCh38
rs577715166 Chr.6: 41159717-41159718 on Build GRCh38
rs150598166 Chr.6: 41159764-41159764 on Build GRCh38
rs545890557 Chr.6: 41159954-41159954 on Build GRCh38
rs530608149 Chr.6: 41160090-41160090 on Build GRCh38
rs186056786 Chr.6: 41159733-41159733 on Build GRCh38
rs764276702 Chr.6: 41160101-41160101 on Build GRCh38
rs760843237 Chr.6: 41159888-41159888 on Build GRCh38
rs777536241 Chr.6: 41159866-41159866 on Build GRCh38
rs1056848093 Chr.6: 41159964-41159964 on Build GRCh38
rs144250872 Chr.6: 41159875-41159875 on Build GRCh38
rs149622783 Chr.6: 41159867-41159867 on Build GRCh38
rs546289076 Chr.6: 41160124-41160124 on Build GRCh38
rs956752148 Chr.6: 41160099-41160099 on Build GRCh38
rs750556369 Chr.6: 41159829-41159829 on Build GRCh38
rs375545806 Chr.6: 41159780-41159780 on Build GRCh38
rs562311103 Chr.6: 41160051-41160051 on Build GRCh38
rs898740410 Chr.6: 41159973-41159973 on Build GRCh38
rs777787951 Chr.6: 41159932-41159932 on Build GRCh38
rs539332069 Chr.6: 41159793-41159793 on Build GRCh38
rs28939079 Chr.6: 41159873-41159873 on Build GRCh38
rs746496916 Chr.6: 41159785-41159785 on Build GRCh38
rs2234255 Chr.6: 41159805-41159805 on Build GRCh38
rs751411176 Chr.6: 41159934-41159934 on Build GRCh38
rs749171917 Chr.6: 41159683-41159683 on Build GRCh38

View Full Product Details