Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41161523-41162031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC105375056
Entrez Gene ID
105375056
Gene Name
uncharacterized LOC105375056
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.6: 41154028-41178568 on Build GRCh38
Transcripts
NM_001271821.1 NM_018965.3 XM_006715116.3

Gene Symbol
TREM2
Entrez Gene ID
54209
Gene Name
triggering receptor expressed on myeloid cells 2
Gene Aliases
TREM-2, Trem2a, Trem2b, Trem2c
UniGene
Hs.435295
Gene Chromosome Location
Chr.6: 41158506-41163200 on Build GRCh38
Transcripts
NM_001271821.1 NM_018965.3 XM_006715116.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1079185 c.59A>G p.N20S Chr.6: 41161595-41161595 on Build GRCh38
4868232 c.59A>G p.N20S Chr.6: 41161595-41161595 on Build GRCh38
7275250 c.53C>T p.A18V Chr.6: 41161601-41161601 on Build GRCh38
4974149 c.48C>T p.S16S Chr.6: 41161606-41161606 on Build GRCh38
4974147 c.48C>T p.S16S Chr.6: 41161606-41161606 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549381797 Chr.6: 41161975-41161975 on Build GRCh38
rs746216516 Chr.6: 41161562-41161562 on Build GRCh38
rs576271749 Chr.6: 41161693-41161693 on Build GRCh38
rs2234251 Chr.6: 41161656-41161656 on Build GRCh38
rs377659415 Chr.6: 41161627-41161627 on Build GRCh38
rs578089722 Chr.6: 41161585-41161585 on Build GRCh38
rs772459581 Chr.6: 41161657-41161657 on Build GRCh38
rs1010247773 Chr.6: 41161909-41161909 on Build GRCh38
rs104894002 Chr.6: 41161557-41161557 on Build GRCh38
rs746818358 Chr.6: 41161663-41161663 on Build GRCh38
rs538188924 Chr.6: 41161570-41161570 on Build GRCh38
rs1050244756 Chr.6: 41161801-41161801 on Build GRCh38
rs534417680 Chr.6: 41161595-41161595 on Build GRCh38
rs772412391 Chr.6: 41161580-41161580 on Build GRCh38
rs968385256 Chr.6: 41161674-41161674 on Build GRCh38
rs376858300 Chr.6: 41161698-41161698 on Build GRCh38
rs777808487 Chr.6: 41161607-41161607 on Build GRCh38
rs927029313 Chr.6: 41161705-41161705 on Build GRCh38
rs769516398 Chr.6: 41161780-41161780 on Build GRCh38
rs565210171 Chr.6: 41161735-41161735 on Build GRCh38
rs748326594 Chr.6: 41161577-41161577 on Build GRCh38
rs34741596 Chr.6: 41161576-41161576 on Build GRCh38
rs920369859 Chr.6: 41161711-41161711 on Build GRCh38
rs1020388866 Chr.6: 41161667-41161667 on Build GRCh38
rs760511786 Chr.6: 41161589-41161589 on Build GRCh38
rs527461310 Chr.6: 41161744-41161744 on Build GRCh38
rs1035288861 Chr.6: 41162005-41162005 on Build GRCh38
rs2234252 Chr.6: 41161571-41161571 on Build GRCh38
rs755030265 Chr.6: 41161650-41161650 on Build GRCh38
rs146886435 Chr.6: 41161769-41161769 on Build GRCh38
rs764738032 Chr.6: 41161599-41161599 on Build GRCh38
rs745525344 Chr.6: 41161678-41161678 on Build GRCh38
rs930458671 Chr.6: 41161730-41161730 on Build GRCh38
rs750837494 Chr.6: 41161614-41161614 on Build GRCh38
rs553407586 Chr.6: 41161603-41161603 on Build GRCh38
rs538260161 Chr.6: 41162006-41162006 on Build GRCh38
rs1040276710 Chr.6: 41161771-41161771 on Build GRCh38
rs779843198 Chr.6: 41161784-41161784 on Build GRCh38
rs757622796 Chr.6: 41161551-41161551 on Build GRCh38
rs751429843 Chr.6: 41161655-41161655 on Build GRCh38
rs545005685 Chr.6: 41161696-41161696 on Build GRCh38
rs893921421 Chr.6: 41161862-41161862 on Build GRCh38
rs192843429 Chr.6: 41161948-41161948 on Build GRCh38
rs990952232 Chr.6: 41161704-41161704 on Build GRCh38
rs927577949 Chr.6: 41161666-41161666 on Build GRCh38
rs901144905 Chr.6: 41161777-41161777 on Build GRCh38
rs899915386 Chr.6: 41161815-41161815 on Build GRCh38
rs569184838 Chr.6: 41161999-41161999 on Build GRCh38
rs189667236 Chr.6: 41161664-41161664 on Build GRCh38
rs1051268914 Chr.6: 41161865-41161865 on Build GRCh38
rs758058447 Chr.6: 41161802-41161803 on Build GRCh38
rs1046950423 Chr.6: 41161753-41161753 on Build GRCh38
rs762650272 Chr.6: 41161606-41161606 on Build GRCh38
rs754308920 Chr.6: 41161649-41161649 on Build GRCh38
rs1003756223 Chr.6: 41161980-41161980 on Build GRCh38
rs778116618 Chr.6: 41161658-41161658 on Build GRCh38
rs573402783 Chr.6: 41161624-41161624 on Build GRCh38
rs61333415 Chr.6: 41161893-41161893 on Build GRCh38
rs367967703 Chr.6: 41161971-41161971 on Build GRCh38
rs907795315 Chr.6: 41161751-41161751 on Build GRCh38
rs1047512297 Chr.6: 41161573-41161573 on Build GRCh38
rs776578102 Chr.6: 41161590-41161590 on Build GRCh38
rs529457146 Chr.6: 41161967-41161967 on Build GRCh38
rs909885625 Chr.6: 41161763-41161763 on Build GRCh38
rs968815095 Chr.6: 41161958-41161958 on Build GRCh38
rs768745050 Chr.6: 41161575-41161575 on Build GRCh38

View Full Product Details