Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151994059-151994527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
S100A10
Entrez Gene ID
6281
Gene Name
S100 calcium binding protein A10
Gene Aliases
42C, ANX2L, ANX2LG, CAL1L, CLP11, Ca[1], GP11, P11, p10
UniGene
Hs.143873
Gene Chromosome Location
Chr.1: 151982910-151994238 on Build GRCh38
Transcripts
NM_002966.2

SNP Detail

SNP ID Chromosome Location
rs977200317 Chr.1: 151994200-151994200 on Build GRCh38
rs192289651 Chr.1: 151994132-151994132 on Build GRCh38
rs1037255127 Chr.1: 151994210-151994210 on Build GRCh38
rs897374772 Chr.1: 151994225-151994225 on Build GRCh38
rs934232890 Chr.1: 151994394-151994394 on Build GRCh38
rs1029611725 Chr.1: 151994248-151994248 on Build GRCh38
rs992521461 Chr.1: 151994327-151994327 on Build GRCh38
rs1051068882 Chr.1: 151994399-151994399 on Build GRCh38
rs917018287 Chr.1: 151994141-151994141 on Build GRCh38
rs998651875 Chr.1: 151994494-151994494 on Build GRCh38
rs1022380396 Chr.1: 151994460-151994460 on Build GRCh38
rs138034099 Chr.1: 151994187-151994187 on Build GRCh38
rs967069202 Chr.1: 151994477-151994477 on Build GRCh38
rs998180935 Chr.1: 151994241-151994241 on Build GRCh38
rs978550667 Chr.1: 151994311-151994311 on Build GRCh38
rs955930918 Chr.1: 151994325-151994325 on Build GRCh38
rs933402932 Chr.1: 151994277-151994277 on Build GRCh38
rs909591194 Chr.1: 151994372-151994372 on Build GRCh38
rs1019096657 Chr.1: 151994186-151994186 on Build GRCh38
rs968588614 Chr.1: 151994281-151994281 on Build GRCh38
rs1047807813 Chr.1: 151994360-151994360 on Build GRCh38
rs1028618215 Chr.1: 151994090-151994090 on Build GRCh38
rs1007268118 Chr.1: 151994437-151994437 on Build GRCh38
rs1006107444 Chr.1: 151994208-151994208 on Build GRCh38
rs1037160659 Chr.1: 151994380-151994380 on Build GRCh38
rs370243984 Chr.1: 151994416-151994416 on Build GRCh38
rs987271684 Chr.1: 151994175-151994175 on Build GRCh38
rs994513447 Chr.1: 151994085-151994085 on Build GRCh38
rs954300001 Chr.1: 151994261-151994261 on Build GRCh38
rs953159839 Chr.1: 151994144-151994144 on Build GRCh38
rs1038745 Chr.1: 151994269-151994269 on Build GRCh38
rs1000063220 Chr.1: 151994483-151994483 on Build GRCh38
rs979891537 Chr.1: 151994369-151994369 on Build GRCh38
rs941536358 Chr.1: 151994378-151994378 on Build GRCh38
rs529498840 Chr.1: 151994222-151994222 on Build GRCh38
rs916900906 Chr.1: 151994329-151994329 on Build GRCh38
rs184159918 Chr.1: 151994409-151994409 on Build GRCh38
rs903932550 Chr.1: 151994468-151994468 on Build GRCh38
rs906588192 Chr.1: 151994276-151994276 on Build GRCh38
rs1032848870 Chr.1: 151994500-151994500 on Build GRCh38
rs948364105 Chr.1: 151994365-151994365 on Build GRCh38
rs1044979548 Chr.1: 151994168-151994168 on Build GRCh38
rs995126478 Chr.1: 151994346-151994346 on Build GRCh38
rs897446754 Chr.1: 151994384-151994384 on Build GRCh38
rs948482778 Chr.1: 151994143-151994143 on Build GRCh38
rs537837934 Chr.1: 151994294-151994294 on Build GRCh38
rs924562047 Chr.1: 151994324-151994324 on Build GRCh38
rs534322422 Chr.1: 151994469-151994469 on Build GRCh38
rs948037658 Chr.1: 151994272-151994272 on Build GRCh38
rs142526671 Chr.1: 151994273-151994273 on Build GRCh38

View Full Product Details