Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 45900403-45900911 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIC5
Entrez Gene ID
53405
Gene Name
chloride intracellular channel 5
Gene Aliases
DFNB102, DFNB103, MST130, MSTP130
UniGene
Hs.485489
Gene Chromosome Location
Chr.6: 45898451-46129809 on Build GRCh38
Transcripts
NM_001114086.1 NM_016929.4 NR_045672.1 NR_045673.1 XM_017010953.1

SNP Detail

SNP ID Chromosome Location
rs1040999278 Chr.6: 45900861-45900861 on Build GRCh38
rs573736988 Chr.6: 45900734-45900734 on Build GRCh38
rs904109807 Chr.6: 45900505-45900505 on Build GRCh38
rs1044336618 Chr.6: 45900503-45900503 on Build GRCh38
rs184534662 Chr.6: 45900716-45900716 on Build GRCh38
rs889612557 Chr.6: 45900838-45900838 on Build GRCh38
rs558015808 Chr.6: 45900560-45900560 on Build GRCh38
rs1009748294 Chr.6: 45900471-45900471 on Build GRCh38
rs1051097576 Chr.6: 45900793-45900793 on Build GRCh38
rs779732451 Chr.6: 45900782-45900782 on Build GRCh38
rs200444578 Chr.6: 45900527-45900527 on Build GRCh38
rs3829841 Chr.6: 45900531-45900531 on Build GRCh38
rs962690900 Chr.6: 45900706-45900706 on Build GRCh38
rs11969051 Chr.6: 45900639-45900639 on Build GRCh38
rs568093188 Chr.6: 45900525-45900525 on Build GRCh38
rs945499669 Chr.6: 45900806-45900806 on Build GRCh38
rs565637530 Chr.6: 45900631-45900631 on Build GRCh38
rs1008033845 Chr.6: 45900850-45900850 on Build GRCh38
rs555624839 Chr.6: 45900718-45900718 on Build GRCh38
rs746396833 Chr.6: 45900527-45900528 on Build GRCh38
rs372268194 Chr.6: 45900462-45900462 on Build GRCh38
rs372460592 Chr.6: 45900731-45900731 on Build GRCh38
rs397957956 Chr.6: 45900542-45900543 on Build GRCh38
rs553874567 Chr.6: 45900620-45900620 on Build GRCh38
rs189059064 Chr.6: 45900743-45900743 on Build GRCh38
rs913995616 Chr.6: 45900801-45900801 on Build GRCh38
rs1028152044 Chr.6: 45900477-45900478 on Build GRCh38
rs892609915 Chr.6: 45900469-45900469 on Build GRCh38
rs577864635 Chr.6: 45900827-45900827 on Build GRCh38
rs934010371 Chr.6: 45900770-45900770 on Build GRCh38
rs1000268194 Chr.6: 45900532-45900532 on Build GRCh38
rs889748576 Chr.6: 45900479-45900479 on Build GRCh38
rs146413220 Chr.6: 45900565-45900565 on Build GRCh38
rs904097663 Chr.6: 45900869-45900869 on Build GRCh38
rs76883893 Chr.6: 45900784-45900784 on Build GRCh38
rs866437994 Chr.6: 45900543-45900543 on Build GRCh38
rs990757645 Chr.6: 45900625-45900625 on Build GRCh38
rs957441069 Chr.6: 45900607-45900607 on Build GRCh38
rs928868258 Chr.6: 45900675-45900675 on Build GRCh38
rs1014038112 Chr.6: 45900570-45900570 on Build GRCh38
rs755814338 Chr.6: 45900744-45900744 on Build GRCh38
rs556330145 Chr.6: 45900674-45900674 on Build GRCh38
rs866664997 Chr.6: 45900544-45900544 on Build GRCh38
rs193059735 Chr.6: 45900444-45900444 on Build GRCh38
rs60854847 Chr.6: 45900651-45900651 on Build GRCh38
rs951261873 Chr.6: 45900644-45900644 on Build GRCh38
rs778527231 Chr.6: 45900781-45900781 on Build GRCh38
rs939693089 Chr.6: 45900717-45900717 on Build GRCh38
rs751320832 Chr.6: 45900531-45900532 on Build GRCh38
rs1043941176 Chr.6: 45900864-45900864 on Build GRCh38

View Full Product Details