Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 52500056-52500594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAM2
Entrez Gene ID
9697
Gene Name
translocation associated membrane protein 2
Gene Aliases
-
UniGene
Hs.520182
Gene Chromosome Location
Chr.6: 52497402-52577064 on Build GRCh38
Transcripts
NM_012288.3 XM_011515005.2

SNP Detail

SNP ID Chromosome Location
rs995426964 Chr.6: 52500220-52500220 on Build GRCh38
rs907159318 Chr.6: 52500539-52500539 on Build GRCh38
rs370458340 Chr.6: 52500523-52500523 on Build GRCh38
rs1008312479 Chr.6: 52500186-52500186 on Build GRCh38
rs994162954 Chr.6: 52500344-52500344 on Build GRCh38
rs752612489 Chr.6: 52500529-52500530 on Build GRCh38
rs368947355 Chr.6: 52500519-52500522 on Build GRCh38
rs543448491 Chr.6: 52500537-52500537 on Build GRCh38
rs971665364 Chr.6: 52500388-52500388 on Build GRCh38
rs9395798 Chr.6: 52500519-52500519 on Build GRCh38
rs547555190 Chr.6: 52500191-52500191 on Build GRCh38
rs951631645 Chr.6: 52500226-52500226 on Build GRCh38
rs73740390 Chr.6: 52500538-52500538 on Build GRCh38
rs531890719 Chr.6: 52500474-52500475 on Build GRCh38
rs191667108 Chr.6: 52500322-52500322 on Build GRCh38
rs1039516439 Chr.6: 52500456-52500456 on Build GRCh38
rs899903417 Chr.6: 52500458-52500458 on Build GRCh38
rs996268733 Chr.6: 52500532-52500532 on Build GRCh38
rs897225299 Chr.6: 52500535-52500536 on Build GRCh38
rs943837006 Chr.6: 52500421-52500421 on Build GRCh38
rs979064573 Chr.6: 52500336-52500336 on Build GRCh38
rs12195210 Chr.6: 52500524-52500524 on Build GRCh38
rs770312726 Chr.6: 52500525-52500526 on Build GRCh38
rs527528871 Chr.6: 52500571-52500571 on Build GRCh38
rs781200143 Chr.6: 52500524-52500528 on Build GRCh38
rs13217180 Chr.6: 52500094-52500094 on Build GRCh38
rs999862496 Chr.6: 52500118-52500118 on Build GRCh38
rs532984806 Chr.6: 52500422-52500422 on Build GRCh38
rs527388957 Chr.6: 52500160-52500160 on Build GRCh38
rs112967280 Chr.6: 52500533-52500534 on Build GRCh38
rs983465137 Chr.6: 52500251-52500251 on Build GRCh38
rs61590094 Chr.6: 52500531-52500531 on Build GRCh38
rs11752918 Chr.6: 52500534-52500534 on Build GRCh38
rs567979112 Chr.6: 52500096-52500096 on Build GRCh38
rs550190291 Chr.6: 52500459-52500459 on Build GRCh38
rs960524893 Chr.6: 52500278-52500278 on Build GRCh38
rs879228961 Chr.6: 52500529-52500529 on Build GRCh38
rs867154990 Chr.6: 52500525-52500525 on Build GRCh38
rs1005646749 Chr.6: 52500178-52500178 on Build GRCh38
rs535645790 Chr.6: 52500463-52500463 on Build GRCh38
rs774648725 Chr.6: 52500148-52500148 on Build GRCh38
rs866084655 Chr.6: 52500132-52500132 on Build GRCh38
rs541618008 Chr.6: 52500549-52500549 on Build GRCh38
rs13217723 Chr.6: 52500528-52500528 on Build GRCh38
rs76545285 Chr.6: 52500462-52500462 on Build GRCh38
rs887622049 Chr.6: 52500520-52500520 on Build GRCh38
rs74741335 Chr.6: 52500567-52500567 on Build GRCh38
rs536931963 Chr.6: 52500117-52500117 on Build GRCh38
rs935245448 Chr.6: 52500374-52500374 on Build GRCh38
rs183392583 Chr.6: 52500087-52500087 on Build GRCh38
rs751940820 Chr.6: 52500526-52500528 on Build GRCh38
rs901744373 Chr.6: 52500147-52500147 on Build GRCh38
rs746116356 Chr.6: 52500527-52500528 on Build GRCh38
rs994609613 Chr.6: 52500536-52500536 on Build GRCh38
rs767683174 Chr.6: 52500306-52500306 on Build GRCh38
rs1026959066 Chr.6: 52500222-52500222 on Build GRCh38
rs887569554 Chr.6: 52500174-52500174 on Build GRCh38
rs753296528 Chr.6: 52500393-52500393 on Build GRCh38
rs1050246999 Chr.6: 52500173-52500173 on Build GRCh38
rs912959861 Chr.6: 52500440-52500440 on Build GRCh38
rs1035844160 Chr.6: 52500252-52500252 on Build GRCh38
rs887129158 Chr.6: 52500502-52500502 on Build GRCh38
rs59714938 Chr.6: 52500544-52500545 on Build GRCh38
rs912899498 Chr.6: 52500107-52500107 on Build GRCh38
rs1017000648 Chr.6: 52500206-52500206 on Build GRCh38
rs947804838 Chr.6: 52500307-52500307 on Build GRCh38
rs991914472 Chr.6: 52500304-52500304 on Build GRCh38
rs924970548 Chr.6: 52500347-52500347 on Build GRCh38
rs368703713 Chr.6: 52500463-52500463 on Build GRCh38
rs369554880 Chr.6: 52500523-52500524 on Build GRCh38
rs989873590 Chr.6: 52500435-52500435 on Build GRCh38
rs576285967 Chr.6: 52500540-52500540 on Build GRCh38
rs368707931 Chr.6: 52500518-52500519 on Build GRCh38
rs890956884 Chr.6: 52500418-52500418 on Build GRCh38
rs899936403 Chr.6: 52500210-52500210 on Build GRCh38
rs1018349298 Chr.6: 52500208-52500208 on Build GRCh38
rs927469842 Chr.6: 52500397-52500397 on Build GRCh38
rs923218795 Chr.6: 52500518-52500518 on Build GRCh38
rs1050299027 Chr.6: 52500520-52500528 on Build GRCh38
rs752489489 Chr.6: 52500516-52500516 on Build GRCh38
rs200545924 Chr.6: 52500529-52500529 on Build GRCh38
rs557879933 Chr.6: 52500533-52500533 on Build GRCh38
rs868780111 Chr.6: 52500105-52500105 on Build GRCh38
rs890979306 Chr.6: 52500156-52500156 on Build GRCh38
rs759719856 Chr.6: 52500212-52500212 on Build GRCh38

View Full Product Details