Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 55541736-55542239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMGCLL1
Entrez Gene ID
54511
Gene Name
3-hydroxymethyl-3-methylglutaryl-CoA lyase like 1
Gene Aliases
bA418P12.1, er-cHL
UniGene
Hs.147054
Gene Chromosome Location
Chr.6: 55434373-55579214 on Build GRCh38
Transcripts
NM_001042406.1 NM_001287741.1 NM_001287746.1 NM_001287753.1 NM_019036.2 NR_109867.1 NR_109869.1 XM_011514700.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5870948 c.277A>G p.K93E Chr.6: 55542062-55542062 on Build GRCh38
3366567 c.218G>T p.G73V Chr.6: 55542121-55542121 on Build GRCh38
3874878 c.246A>T p.E82D Chr.6: 55542093-55542093 on Build GRCh38
3874877 c.331A>G p.T111A Chr.6: 55541785-55541785 on Build GRCh38
3874879 c.246A>C p.E82D Chr.6: 55542093-55542093 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368235647 Chr.6: 55542149-55542149 on Build GRCh38
rs770276313 Chr.6: 55542099-55542099 on Build GRCh38
rs1034735119 Chr.6: 55542207-55542207 on Build GRCh38
rs924221492 Chr.6: 55542201-55542201 on Build GRCh38
rs754175137 Chr.6: 55542041-55542041 on Build GRCh38
rs779054521 Chr.6: 55542055-55542055 on Build GRCh38
rs756362676 Chr.6: 55542025-55542025 on Build GRCh38
rs537632205 Chr.6: 55541887-55541887 on Build GRCh38
rs758246894 Chr.6: 55542115-55542115 on Build GRCh38
rs373008744 Chr.6: 55541774-55541774 on Build GRCh38
rs752734099 Chr.6: 55542136-55542136 on Build GRCh38
rs1043703504 Chr.6: 55541876-55541876 on Build GRCh38
rs924198161 Chr.6: 55541955-55541955 on Build GRCh38
rs766194501 Chr.6: 55541777-55541777 on Build GRCh38
rs780702325 Chr.6: 55542095-55542097 on Build GRCh38
rs755330325 Chr.6: 55542192-55542192 on Build GRCh38
rs978776923 Chr.6: 55541913-55541913 on Build GRCh38
rs369398868 Chr.6: 55541870-55541870 on Build GRCh38
rs746458408 Chr.6: 55541822-55541822 on Build GRCh38
rs765971485 Chr.6: 55541873-55541873 on Build GRCh38
rs889923126 Chr.6: 55542000-55542000 on Build GRCh38
rs990823465 Chr.6: 55541839-55541839 on Build GRCh38
rs377549470 Chr.6: 55541817-55541817 on Build GRCh38
rs936914324 Chr.6: 55542210-55542210 on Build GRCh38
rs201294383 Chr.6: 55542171-55542171 on Build GRCh38
rs773557662 Chr.6: 55542089-55542089 on Build GRCh38
rs576879078 Chr.6: 55542155-55542155 on Build GRCh38
rs377158973 Chr.6: 55542045-55542045 on Build GRCh38
rs748496553 Chr.6: 55541796-55541796 on Build GRCh38
rs775847582 Chr.6: 55541831-55541831 on Build GRCh38
rs557736389 Chr.6: 55542133-55542133 on Build GRCh38
rs752562682 Chr.6: 55541994-55541994 on Build GRCh38
rs561064194 Chr.6: 55541922-55541922 on Build GRCh38
rs772657643 Chr.6: 55542060-55542060 on Build GRCh38
rs757284956 Chr.6: 55542050-55542050 on Build GRCh38
rs773922270 Chr.6: 55542103-55542103 on Build GRCh38
rs143872373 Chr.6: 55542170-55542170 on Build GRCh38
rs574784760 Chr.6: 55542006-55542006 on Build GRCh38
rs376191221 Chr.6: 55541767-55541767 on Build GRCh38
rs761407411 Chr.6: 55541852-55541852 on Build GRCh38
rs749960854 Chr.6: 55541770-55541770 on Build GRCh38
rs772889179 Chr.6: 55541858-55541858 on Build GRCh38
rs777948133 Chr.6: 55541799-55541799 on Build GRCh38
rs1051237561 Chr.6: 55541990-55541990 on Build GRCh38
rs780358456 Chr.6: 55542067-55542067 on Build GRCh38
rs900848729 Chr.6: 55542042-55542042 on Build GRCh38
rs891301244 Chr.6: 55542020-55542020 on Build GRCh38
rs776261820 Chr.6: 55541762-55541762 on Build GRCh38
rs1016343481 Chr.6: 55541810-55541810 on Build GRCh38
rs761460532 Chr.6: 55541764-55541764 on Build GRCh38
rs747341972 Chr.6: 55542070-55542070 on Build GRCh38
rs760536172 Chr.6: 55542147-55542147 on Build GRCh38
rs199901825 Chr.6: 55542179-55542179 on Build GRCh38
rs949383419 Chr.6: 55541894-55541894 on Build GRCh38
rs767359888 Chr.6: 55542022-55542022 on Build GRCh38
rs377493761 Chr.6: 55542023-55542023 on Build GRCh38
rs769056962 Chr.6: 55542081-55542081 on Build GRCh38
rs772259012 Chr.6: 55541827-55541827 on Build GRCh38
rs188759930 Chr.6: 55541779-55541779 on Build GRCh38
rs764051638 Chr.6: 55542162-55542162 on Build GRCh38
rs759518220 Chr.6: 55542019-55542019 on Build GRCh38
rs936873904 Chr.6: 55541965-55541965 on Build GRCh38
rs201603961 Chr.6: 55542083-55542083 on Build GRCh38
rs780515537 Chr.6: 55542184-55542184 on Build GRCh38
rs746304590 Chr.6: 55542059-55542059 on Build GRCh38
rs1027306088 Chr.6: 55542095-55542095 on Build GRCh38
rs201567160 Chr.6: 55541780-55541780 on Build GRCh38
rs958507384 Chr.6: 55542131-55542131 on Build GRCh38
rs12196868 Chr.6: 55541872-55541872 on Build GRCh38
rs762734248 Chr.6: 55542091-55542091 on Build GRCh38
rs553817629 Chr.6: 55542126-55542126 on Build GRCh38
rs373355530 Chr.6: 55541797-55541797 on Build GRCh38
rs200883858 Chr.6: 55542130-55542130 on Build GRCh38
rs371252434 Chr.6: 55542186-55542186 on Build GRCh38
rs781103945 Chr.6: 55541784-55541784 on Build GRCh38
rs778122632 Chr.6: 55542031-55542031 on Build GRCh38
rs370628283 Chr.6: 55542073-55542073 on Build GRCh38
rs369668244 Chr.6: 55541795-55541795 on Build GRCh38
rs753945050 Chr.6: 55542165-55542165 on Build GRCh38
rs751446215 Chr.6: 55541888-55541888 on Build GRCh38
rs370090783 Chr.6: 55541855-55541855 on Build GRCh38
rs749350015 Chr.6: 55541811-55541811 on Build GRCh38
rs371077384 Chr.6: 55542166-55542166 on Build GRCh38
rs756624309 Chr.6: 55541857-55541858 on Build GRCh38
rs771094530 Chr.6: 55541813-55541813 on Build GRCh38

View Full Product Details