Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 96521741-96522242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UFL1
Entrez Gene ID
23376
Gene Name
UFM1 specific ligase 1
Gene Aliases
KIAA0776, Maxer, NLBP, RCAD
UniGene
Hs.149367
Gene Chromosome Location
Chr.6: 96521826-96555276 on Build GRCh38
Transcripts
NM_015323.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5595532 c.5C>G p.A2G Chr.6: 96521878-96521878 on Build GRCh38
1697656 c.19G>A p.E7K Chr.6: 96521892-96521892 on Build GRCh38
7139003 c.19G>C p.E7Q Chr.6: 96521892-96521892 on Build GRCh38
4739030 c.71C>T p.T24M Chr.6: 96521944-96521944 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749960877 Chr.6: 96521881-96521881 on Build GRCh38
rs921228694 Chr.6: 96521844-96521844 on Build GRCh38
rs1021941870 Chr.6: 96522100-96522100 on Build GRCh38
rs917312980 Chr.6: 96522184-96522184 on Build GRCh38
rs755427839 Chr.6: 96521899-96521899 on Build GRCh38
rs779862144 Chr.6: 96521890-96521891 on Build GRCh38
rs530579178 Chr.6: 96522002-96522002 on Build GRCh38
rs1033233087 Chr.6: 96521794-96521794 on Build GRCh38
rs77958363 Chr.6: 96522041-96522041 on Build GRCh38
rs936076801 Chr.6: 96522070-96522070 on Build GRCh38
rs746919999 Chr.6: 96521930-96521930 on Build GRCh38
rs745846634 Chr.6: 96521858-96521858 on Build GRCh38
rs371351936 Chr.6: 96521841-96521841 on Build GRCh38
rs944812791 Chr.6: 96521882-96521882 on Build GRCh38
rs1057233046 Chr.6: 96521761-96521761 on Build GRCh38
rs369308301 Chr.6: 96521991-96521991 on Build GRCh38
rs889579550 Chr.6: 96522091-96522091 on Build GRCh38
rs748439570 Chr.6: 96521900-96521900 on Build GRCh38
rs377568383 Chr.6: 96521901-96521902 on Build GRCh38
rs572822034 Chr.6: 96521970-96521970 on Build GRCh38
rs150419821 Chr.6: 96521771-96521771 on Build GRCh38
rs763244566 Chr.6: 96522198-96522198 on Build GRCh38
rs888687593 Chr.6: 96522025-96522025 on Build GRCh38
rs953804467 Chr.6: 96521840-96521840 on Build GRCh38
rs13210597 Chr.6: 96521818-96521818 on Build GRCh38
rs1028559844 Chr.6: 96522033-96522033 on Build GRCh38
rs1045442800 Chr.6: 96521945-96521945 on Build GRCh38
rs761621268 Chr.6: 96521950-96521950 on Build GRCh38
rs775504062 Chr.6: 96521860-96521860 on Build GRCh38
rs568905143 Chr.6: 96522024-96522024 on Build GRCh38
rs749407211 Chr.6: 96521908-96521908 on Build GRCh38
rs768536494 Chr.6: 96521944-96521944 on Build GRCh38
rs909855891 Chr.6: 96522210-96522210 on Build GRCh38
rs541972244 Chr.6: 96521984-96521984 on Build GRCh38
rs756378970 Chr.6: 96521902-96521902 on Build GRCh38
rs1015798827 Chr.6: 96521803-96521803 on Build GRCh38
rs41292432 Chr.6: 96521842-96521842 on Build GRCh38
rs778831056 Chr.6: 96521992-96521992 on Build GRCh38
rs143837187 Chr.6: 96521874-96521874 on Build GRCh38
rs148081377 Chr.6: 96521893-96521893 on Build GRCh38
rs576876120 Chr.6: 96521863-96521863 on Build GRCh38
rs948726085 Chr.6: 96521998-96521998 on Build GRCh38
rs772153994 Chr.6: 96521859-96521859 on Build GRCh38
rs773390018 Chr.6: 96521852-96521852 on Build GRCh38
rs903338487 Chr.6: 96521789-96521789 on Build GRCh38
rs971165416 Chr.6: 96521822-96521822 on Build GRCh38
rs764776545 Chr.6: 96521879-96521879 on Build GRCh38
rs960780877 Chr.6: 96522141-96522141 on Build GRCh38
rs1006501815 Chr.6: 96522093-96522093 on Build GRCh38
rs1045980691 Chr.6: 96522108-96522108 on Build GRCh38
rs545684749 Chr.6: 96521956-96521956 on Build GRCh38
rs998367931 Chr.6: 96522061-96522061 on Build GRCh38
rs911908712 Chr.6: 96522054-96522054 on Build GRCh38
rs778090067 Chr.6: 96521904-96521904 on Build GRCh38
rs924758963 Chr.6: 96522067-96522067 on Build GRCh38
rs995644928 Chr.6: 96522156-96522157 on Build GRCh38
rs965985966 Chr.6: 96522055-96522055 on Build GRCh38
rs906870360 Chr.6: 96522109-96522109 on Build GRCh38
rs578093742 Chr.6: 96522076-96522077 on Build GRCh38
rs748303900 Chr.6: 96521849-96521849 on Build GRCh38
rs998314778 Chr.6: 96521808-96521808 on Build GRCh38
rs559384235 Chr.6: 96522107-96522107 on Build GRCh38
rs755404414 Chr.6: 96521965-96521965 on Build GRCh38
rs566260417 Chr.6: 96521791-96521791 on Build GRCh38
rs941571087 Chr.6: 96522031-96522031 on Build GRCh38
rs755884785 Chr.6: 96521886-96521888 on Build GRCh38
rs781489845 Chr.6: 96521848-96521848 on Build GRCh38
rs993286272 Chr.6: 96521964-96521964 on Build GRCh38
rs768835105 Chr.6: 96521845-96521845 on Build GRCh38
rs764901798 Chr.6: 96521954-96521954 on Build GRCh38
rs771080473 Chr.6: 96521922-96521922 on Build GRCh38
rs775558153 Chr.6: 96521924-96521924 on Build GRCh38
rs753378572 Chr.6: 96521831-96521831 on Build GRCh38
rs992043533 Chr.6: 96522157-96522157 on Build GRCh38
rs138149174 Chr.6: 96522122-96522122 on Build GRCh38
rs769980249 Chr.6: 96521850-96521850 on Build GRCh38
rs1008108230 Chr.6: 96522204-96522204 on Build GRCh38
rs776268284 Chr.6: 96521947-96521947 on Build GRCh38
rs568507071 Chr.6: 96521829-96521829 on Build GRCh38
rs747146758 Chr.6: 96522196-96522196 on Build GRCh38
rs111828576 Chr.6: 96522074-96522074 on Build GRCh38
rs537159862 Chr.6: 96521837-96521837 on Build GRCh38
rs552886092 Chr.6: 96521957-96521957 on Build GRCh38
rs944778419 Chr.6: 96521774-96521774 on Build GRCh38
rs757866071 Chr.6: 96521889-96521889 on Build GRCh38
rs564062066 Chr.6: 96522039-96522039 on Build GRCh38
rs897340042 Chr.6: 96521804-96521804 on Build GRCh38
rs977947614 Chr.6: 96522133-96522133 on Build GRCh38
rs752175111 Chr.6: 96521960-96521960 on Build GRCh38
rs763788123 Chr.6: 96521870-96521870 on Build GRCh38
rs767970078 Chr.6: 96521967-96521967 on Build GRCh38
rs774434584 Chr.6: 96521796-96521796 on Build GRCh38
rs890572283 Chr.6: 96521768-96521768 on Build GRCh38
rs751951771 Chr.6: 96521894-96521894 on Build GRCh38
rs866430810 Chr.6: 96521872-96521872 on Build GRCh38
rs948755226 Chr.6: 96521775-96521775 on Build GRCh38
rs749511196 Chr.6: 96521990-96521990 on Build GRCh38
rs1028174782 Chr.6: 96522187-96522187 on Build GRCh38
rs149561858 Chr.6: 96522209-96522209 on Build GRCh38
rs1030924257 Chr.6: 96522135-96522135 on Build GRCh38
rs778391128 Chr.6: 96521835-96521835 on Build GRCh38
rs953985286 Chr.6: 96522190-96522190 on Build GRCh38
rs1029825714 Chr.6: 96522083-96522083 on Build GRCh38
rs749008816 Chr.6: 96521896-96521897 on Build GRCh38
rs756853002 Chr.6: 96521834-96521834 on Build GRCh38
rs756503902 Chr.6: 96521979-96521979 on Build GRCh38
rs9486460 Chr.6: 96521792-96521792 on Build GRCh38
rs1055029795 Chr.6: 96521909-96521909 on Build GRCh38

View Full Product Details