Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 98834466-98834989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POU3F2
Entrez Gene ID
5454
Gene Name
POU class 3 homeobox 2
Gene Aliases
BRN2, N-Oct3, OCT7, OTF-7, OTF7, POUF3, brn-2, oct-7
UniGene
Hs.182505
Gene Chromosome Location
Chr.6: 98834704-98838790 on Build GRCh38
Transcripts
NM_005604.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6752949 c.46G>A p.A16T Chr.6: 98834919-98834919 on Build GRCh38
3179250 c.82C>T p.Q28* Chr.6: 98834955-98834955 on Build GRCh38
4643112 c.68C>T p.P23L Chr.6: 98834941-98834941 on Build GRCh38
6752950 c.116G>T p.S39I Chr.6: 98834989-98834989 on Build GRCh38
743755 c.106G>C p.E36Q Chr.6: 98834979-98834979 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1046745991 Chr.6: 98834620-98834620 on Build GRCh38
rs781334524 Chr.6: 98834828-98834828 on Build GRCh38
rs1017881066 Chr.6: 98834752-98834752 on Build GRCh38
rs904815201 Chr.6: 98834630-98834630 on Build GRCh38
rs956129747 Chr.6: 98834598-98834598 on Build GRCh38
rs756228158 Chr.6: 98834885-98834885 on Build GRCh38
rs557369736 Chr.6: 98834649-98834649 on Build GRCh38
rs1000206538 Chr.6: 98834546-98834546 on Build GRCh38
rs879670097 Chr.6: 98834738-98834739 on Build GRCh38
rs949242597 Chr.6: 98834613-98834613 on Build GRCh38
rs771011381 Chr.6: 98834834-98834834 on Build GRCh38
rs763367137 Chr.6: 98834865-98834865 on Build GRCh38
rs781220601 Chr.6: 98834882-98834882 on Build GRCh38
rs756316047 Chr.6: 98834972-98834972 on Build GRCh38
rs367580619 Chr.6: 98834860-98834860 on Build GRCh38
rs746735872 Chr.6: 98834918-98834918 on Build GRCh38
rs371461013 Chr.6: 98834926-98834926 on Build GRCh38
rs552686016 Chr.6: 98834756-98834756 on Build GRCh38
rs913238716 Chr.6: 98834949-98834949 on Build GRCh38
rs775075666 Chr.6: 98834852-98834852 on Build GRCh38
rs913756348 Chr.6: 98834608-98834608 on Build GRCh38
rs573432263 Chr.6: 98834694-98834694 on Build GRCh38
rs774389211 Chr.6: 98834836-98834836 on Build GRCh38
rs956056697 Chr.6: 98834822-98834822 on Build GRCh38
rs935987796 Chr.6: 98834633-98834633 on Build GRCh38
rs1031706285 Chr.6: 98834549-98834549 on Build GRCh38
rs761363151 Chr.6: 98834925-98834925 on Build GRCh38
rs1009597027 Chr.6: 98834564-98834564 on Build GRCh38
rs926042200 Chr.6: 98834583-98834583 on Build GRCh38
rs1009030406 Chr.6: 98834747-98834747 on Build GRCh38
rs569128447 Chr.6: 98834671-98834671 on Build GRCh38
rs893050563 Chr.6: 98834525-98834525 on Build GRCh38
rs926507959 Chr.6: 98834635-98834635 on Build GRCh38
rs1026923678 Chr.6: 98834539-98834539 on Build GRCh38
rs763454533 Chr.6: 98834936-98834936 on Build GRCh38
rs767818457 Chr.6: 98834966-98834966 on Build GRCh38
rs978725644 Chr.6: 98834572-98834572 on Build GRCh38
rs908288674 Chr.6: 98834594-98834594 on Build GRCh38
rs1008945786 Chr.6: 98834669-98834669 on Build GRCh38
rs937690404 Chr.6: 98834642-98834642 on Build GRCh38
rs1054129235 Chr.6: 98834519-98834519 on Build GRCh38
rs201781902 Chr.6: 98834883-98834883 on Build GRCh38
rs970974590 Chr.6: 98834947-98834947 on Build GRCh38
rs773775751 Chr.6: 98834927-98834927 on Build GRCh38
rs917393922 Chr.6: 98834791-98834791 on Build GRCh38
rs961075291 Chr.6: 98834787-98834787 on Build GRCh38
rs1033037887 Chr.6: 98834807-98834807 on Build GRCh38
rs779917134 Chr.6: 98834907-98834907 on Build GRCh38
rs755239626 Chr.6: 98834872-98834872 on Build GRCh38
rs1045053184 Chr.6: 98834622-98834622 on Build GRCh38
rs779038428 Chr.6: 98834886-98834886 on Build GRCh38
rs766738537 Chr.6: 98834945-98834945 on Build GRCh38
rs891875185 Chr.6: 98834661-98834661 on Build GRCh38
rs772071914 Chr.6: 98834903-98834903 on Build GRCh38
rs948885654 Chr.6: 98834803-98834803 on Build GRCh38
rs751804272 Chr.6: 98834870-98834870 on Build GRCh38
rs891906540 Chr.6: 98834553-98834553 on Build GRCh38
rs768389252 Chr.6: 98834919-98834919 on Build GRCh38
rs766725738 Chr.6: 98834866-98834866 on Build GRCh38
rs1001159830 Chr.6: 98834516-98834516 on Build GRCh38
rs983334689 Chr.6: 98834562-98834563 on Build GRCh38
rs993698609 Chr.6: 98834537-98834537 on Build GRCh38
rs992975842 Chr.6: 98834600-98834600 on Build GRCh38
rs536962188 Chr.6: 98834677-98834677 on Build GRCh38
rs371518292 Chr.6: 98834646-98834646 on Build GRCh38
rs751975215 Chr.6: 98834954-98834954 on Build GRCh38
rs539068250 Chr.6: 98834543-98834543 on Build GRCh38
rs1017869753 Chr.6: 98834569-98834569 on Build GRCh38
rs983779231 Chr.6: 98834780-98834780 on Build GRCh38
rs186667950 Chr.6: 98834704-98834704 on Build GRCh38
rs745693384 Chr.6: 98834847-98834847 on Build GRCh38
rs554918739 Chr.6: 98834693-98834693 on Build GRCh38
rs759775933 Chr.6: 98834960-98834960 on Build GRCh38
rs988896907 Chr.6: 98834599-98834599 on Build GRCh38
rs571759235 Chr.6: 98834542-98834542 on Build GRCh38
rs965290942 Chr.6: 98834563-98834563 on Build GRCh38
rs1010192802 Chr.6: 98834528-98834528 on Build GRCh38
rs577299542 Chr.6: 98834790-98834790 on Build GRCh38
rs200105737 Chr.6: 98834862-98834862 on Build GRCh38
rs1000217603 Chr.6: 98834762-98834762 on Build GRCh38
rs1015275439 Chr.6: 98834780-98834780 on Build GRCh38
rs752836778 Chr.6: 98834970-98834970 on Build GRCh38
rs898817967 Chr.6: 98834682-98834682 on Build GRCh38
rs745783228 Chr.6: 98834902-98834902 on Build GRCh38
rs948992683 Chr.6: 98834513-98834513 on Build GRCh38
rs1053016703 Chr.6: 98834894-98834894 on Build GRCh38
rs748117807 Chr.6: 98834831-98834831 on Build GRCh38
rs750908730 Chr.6: 98834863-98834863 on Build GRCh38
rs866778761 Chr.6: 98834571-98834571 on Build GRCh38

View Full Product Details