Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 98838223-98838719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POU3F2
Entrez Gene ID
5454
Gene Name
POU class 3 homeobox 2
Gene Aliases
BRN2, N-Oct3, OCT7, OTF-7, OTF7, POUF3, brn-2, oct-7
UniGene
Hs.182505
Gene Chromosome Location
Chr.6: 98834704-98838790 on Build GRCh38
Transcripts
NM_005604.3

SNP Detail

SNP ID Chromosome Location
rs894130549 Chr.6: 98838448-98838448 on Build GRCh38
rs550416789 Chr.6: 98838421-98838421 on Build GRCh38
rs1042432353 Chr.6: 98838447-98838447 on Build GRCh38
rs185412225 Chr.6: 98838257-98838257 on Build GRCh38
rs190040379 Chr.6: 98838290-98838290 on Build GRCh38
rs991047406 Chr.6: 98838327-98838327 on Build GRCh38
rs998420710 Chr.6: 98838683-98838683 on Build GRCh38
rs924520490 Chr.6: 98838320-98838320 on Build GRCh38
rs915524715 Chr.6: 98838374-98838374 on Build GRCh38
rs540232403 Chr.6: 98838281-98838281 on Build GRCh38
rs1038950151 Chr.6: 98838594-98838594 on Build GRCh38
rs934099184 Chr.6: 98838497-98838498 on Build GRCh38
rs566556863 Chr.6: 98838546-98838546 on Build GRCh38
rs924013783 Chr.6: 98838470-98838470 on Build GRCh38
rs1016273431 Chr.6: 98838651-98838651 on Build GRCh38
rs1051995423 Chr.6: 98838498-98838498 on Build GRCh38
rs568570734 Chr.6: 98838427-98838427 on Build GRCh38
rs545778736 Chr.6: 98838265-98838265 on Build GRCh38
rs529442854 Chr.6: 98838487-98838487 on Build GRCh38
rs1012653136 Chr.6: 98838491-98838491 on Build GRCh38
rs901402159 Chr.6: 98838663-98838663 on Build GRCh38
rs1048139553 Chr.6: 98838545-98838545 on Build GRCh38
rs959591190 Chr.6: 98838306-98838306 on Build GRCh38
rs193052910 Chr.6: 98838526-98838526 on Build GRCh38
rs1056747568 Chr.6: 98838434-98838434 on Build GRCh38
rs576553030 Chr.6: 98838622-98838622 on Build GRCh38
rs533797850 Chr.6: 98838587-98838587 on Build GRCh38

View Full Product Details