Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 101818256-101818800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRIK2
Entrez Gene ID
2898
Gene Name
glutamate ionotropic receptor kainate type subunit 2
Gene Aliases
EAA4, GLR6, GLUK6, GLUR6, GluK2, MRT6
UniGene
Hs.98262
Gene Chromosome Location
Chr.6: 101393708-102070083 on Build GRCh38
Transcripts
NM_001166247.1 NM_021956.4 NM_175768.3 XM_005266945.2 XM_005266946.3 XM_011535777.2 XM_011535778.2 XM_017010781.1 XM_017010782.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3429810 c.1266G>A p.A422A Chr.6: 101818432-101818432 on Build GRCh38
353397 c.1265C>G p.A422G Chr.6: 101818431-101818431 on Build GRCh38
3429809 c.1266G>A p.A422A Chr.6: 101818432-101818432 on Build GRCh38
6047568 c.1210A>G p.T404A Chr.6: 101818376-101818376 on Build GRCh38
7268561 c.1234A>C p.N412H Chr.6: 101818400-101818400 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371470629 Chr.6: 101818335-101818335 on Build GRCh38
rs1047759346 Chr.6: 101818705-101818705 on Build GRCh38
rs747460784 Chr.6: 101818429-101818429 on Build GRCh38
rs761841038 Chr.6: 101818459-101818459 on Build GRCh38
rs772890192 Chr.6: 101818438-101818438 on Build GRCh38
rs766247996 Chr.6: 101818352-101818352 on Build GRCh38
rs201947494 Chr.6: 101818327-101818327 on Build GRCh38
rs769463301 Chr.6: 101818435-101818435 on Build GRCh38
rs567486874 Chr.6: 101818697-101818697 on Build GRCh38
rs529327586 Chr.6: 101818698-101818698 on Build GRCh38
rs527427379 Chr.6: 101818552-101818552 on Build GRCh38
rs372157937 Chr.6: 101818378-101818378 on Build GRCh38
rs561404499 Chr.6: 101818524-101818524 on Build GRCh38
rs752702455 Chr.6: 101818453-101818453 on Build GRCh38
rs750388978 Chr.6: 101818643-101818643 on Build GRCh38
rs376836256 Chr.6: 101818432-101818432 on Build GRCh38
rs969084333 Chr.6: 101818664-101818664 on Build GRCh38
rs753793494 Chr.6: 101818372-101818372 on Build GRCh38
rs568417501 Chr.6: 101818676-101818676 on Build GRCh38
rs368928564 Chr.6: 101818708-101818708 on Build GRCh38
rs780386611 Chr.6: 101818505-101818505 on Build GRCh38
rs1002691544 Chr.6: 101818484-101818484 on Build GRCh38
rs746116443 Chr.6: 101818416-101818416 on Build GRCh38
rs924776084 Chr.6: 101818301-101818301 on Build GRCh38
rs878993708 Chr.6: 101818298-101818298 on Build GRCh38
rs1001999953 Chr.6: 101818357-101818357 on Build GRCh38
rs772446281 Chr.6: 101818319-101818319 on Build GRCh38
rs781515341 Chr.6: 101818532-101818532 on Build GRCh38
rs755394921 Chr.6: 101818527-101818527 on Build GRCh38
rs767297777 Chr.6: 101818451-101818451 on Build GRCh38
rs1057350941 Chr.6: 101818582-101818583 on Build GRCh38
rs765266376 Chr.6: 101818463-101818463 on Build GRCh38
rs987659978 Chr.6: 101818764-101818764 on Build GRCh38
rs762797067 Chr.6: 101818351-101818351 on Build GRCh38
rs34747916 Chr.6: 101818447-101818447 on Build GRCh38
rs758603527 Chr.6: 101818499-101818499 on Build GRCh38
rs758656803 Chr.6: 101818418-101818418 on Build GRCh38
rs775987807 Chr.6: 101818330-101818330 on Build GRCh38
rs750547464 Chr.6: 101818490-101818490 on Build GRCh38
rs1013411269 Chr.6: 101818622-101818622 on Build GRCh38
rs199786854 Chr.6: 101818439-101818439 on Build GRCh38
rs950998037 Chr.6: 101818411-101818411 on Build GRCh38
rs925157238 Chr.6: 101818638-101818638 on Build GRCh38
rs937872627 Chr.6: 101818668-101818668 on Build GRCh38
rs751419152 Chr.6: 101818356-101818356 on Build GRCh38
rs773023664 Chr.6: 101818346-101818346 on Build GRCh38
rs769251596 Chr.6: 101818329-101818329 on Build GRCh38
rs779057205 Chr.6: 101818386-101818386 on Build GRCh38
rs142741212 Chr.6: 101818326-101818326 on Build GRCh38
rs751662930 Chr.6: 101818431-101818431 on Build GRCh38
rs1033777066 Chr.6: 101818361-101818361 on Build GRCh38
rs767556780 Chr.6: 101818371-101818371 on Build GRCh38
rs548076508 Chr.6: 101818710-101818710 on Build GRCh38
rs886441994 Chr.6: 101818707-101818707 on Build GRCh38
rs945313739 Chr.6: 101818569-101818569 on Build GRCh38
rs34006430 Chr.6: 101818532-101818533 on Build GRCh38
rs55749135 Chr.6: 101818341-101818341 on Build GRCh38
rs1026717121 Chr.6: 101818390-101818390 on Build GRCh38
rs1021635621 Chr.6: 101818626-101818626 on Build GRCh38
rs531702161 Chr.6: 101818535-101818535 on Build GRCh38
rs896593544 Chr.6: 101818677-101818677 on Build GRCh38
rs550307464 Chr.6: 101818601-101818601 on Build GRCh38
rs1006423305 Chr.6: 101818422-101818422 on Build GRCh38
rs780255159 Chr.6: 101818420-101818420 on Build GRCh38
rs566362621 Chr.6: 101818712-101818712 on Build GRCh38
rs763391260 Chr.6: 101818366-101818366 on Build GRCh38

View Full Product Details