Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 125781069-125781574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCOA7
Entrez Gene ID
135112
Gene Name
nuclear receptor coactivator 7
Gene Aliases
ERAP140, ESNA1, NCOA7-AS, Nbla00052, Nbla10993, TLDC4, dJ187J11.3
UniGene
Hs.171426
Gene Chromosome Location
Chr.6: 125780948-125932030 on Build GRCh38
Transcripts
NM_001199619.1 NM_001199620.1 XM_006715340.3 XM_017010270.1 XM_017010274.1

SNP Detail

SNP ID Chromosome Location
rs1003290049 Chr.6: 125781462-125781462 on Build GRCh38
rs375726266 Chr.6: 125781330-125781330 on Build GRCh38
rs963953169 Chr.6: 125781502-125781502 on Build GRCh38
rs3832433 Chr.6: 125781405-125781405 on Build GRCh38
rs893688366 Chr.6: 125781388-125781388 on Build GRCh38
rs757196614 Chr.6: 125781122-125781122 on Build GRCh38
rs990224382 Chr.6: 125781292-125781292 on Build GRCh38
rs138286667 Chr.6: 125781480-125781480 on Build GRCh38
rs977637251 Chr.6: 125781358-125781358 on Build GRCh38
rs886797870 Chr.6: 125781152-125781152 on Build GRCh38
rs972122332 Chr.6: 125781178-125781178 on Build GRCh38
rs41285252 Chr.6: 125781256-125781256 on Build GRCh38
rs958829584 Chr.6: 125781258-125781258 on Build GRCh38
rs1002415305 Chr.6: 125781547-125781547 on Build GRCh38
rs931032365 Chr.6: 125781463-125781463 on Build GRCh38
rs909442645 Chr.6: 125781135-125781135 on Build GRCh38
rs781029762 Chr.6: 125781139-125781139 on Build GRCh38
rs1026637891 Chr.6: 125781144-125781144 on Build GRCh38
rs1053631220 Chr.6: 125781384-125781384 on Build GRCh38
rs377027685 Chr.6: 125781391-125781391 on Build GRCh38
rs914558116 Chr.6: 125781303-125781303 on Build GRCh38
rs899583424 Chr.6: 125781113-125781113 on Build GRCh38
rs547972432 Chr.6: 125781229-125781229 on Build GRCh38
rs755634949 Chr.6: 125781214-125781214 on Build GRCh38
rs773665963 Chr.6: 125781478-125781478 on Build GRCh38
rs3734635 Chr.6: 125781340-125781340 on Build GRCh38
rs571541845 Chr.6: 125781398-125781398 on Build GRCh38
rs1020689722 Chr.6: 125781396-125781396 on Build GRCh38
rs572721651 Chr.6: 125781285-125781288 on Build GRCh38
rs918958984 Chr.6: 125781189-125781189 on Build GRCh38
rs918916933 Chr.6: 125781521-125781521 on Build GRCh38
rs745378160 Chr.6: 125781165-125781165 on Build GRCh38
rs958797105 Chr.6: 125781477-125781477 on Build GRCh38
rs933058419 Chr.6: 125781194-125781194 on Build GRCh38
rs970930663 Chr.6: 125781198-125781198 on Build GRCh38
rs1004129236 Chr.6: 125781184-125781184 on Build GRCh38
rs768450203 Chr.6: 125781390-125781390 on Build GRCh38
rs539167220 Chr.6: 125781104-125781104 on Build GRCh38
rs149196829 Chr.6: 125781492-125781492 on Build GRCh38
rs555847136 Chr.6: 125781515-125781515 on Build GRCh38
rs912822263 Chr.6: 125781366-125781366 on Build GRCh38
rs991628208 Chr.6: 125781101-125781101 on Build GRCh38
rs958827366 Chr.6: 125781096-125781096 on Build GRCh38
rs906468221 Chr.6: 125781527-125781527 on Build GRCh38
rs150451452 Chr.6: 125781121-125781121 on Build GRCh38
rs942316708 Chr.6: 125781176-125781176 on Build GRCh38
rs558473659 Chr.6: 125781470-125781470 on Build GRCh38
rs903216151 Chr.6: 125781432-125781432 on Build GRCh38

View Full Product Details