Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154552297-154552795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBE2Q1
Entrez Gene ID
55585
Gene Name
ubiquitin conjugating enzyme E2 Q1
Gene Aliases
GTAP, NICE-5, PRO3094, UBE2Q
UniGene
Hs.607928
Gene Chromosome Location
Chr.1: 154548575-154558644 on Build GRCh38
Transcripts
NM_017582.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1600876 c.875+7G>T p.? Chr.1: 154552397-154552397 on Build GRCh38
7286001 c.796T>C p.S266P Chr.1: 154552754-154552754 on Build GRCh38
6815154 c.794G>A p.R265Q Chr.1: 154552756-154552756 on Build GRCh38
6815155 c.763C>T p.R255W Chr.1: 154552787-154552787 on Build GRCh38
206573 c.829G>A p.E277K Chr.1: 154552450-154552450 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977110190 Chr.1: 154552675-154552675 on Build GRCh38
rs546229703 Chr.1: 154552623-154552623 on Build GRCh38
rs201001765 Chr.1: 154552737-154552737 on Build GRCh38
rs751863612 Chr.1: 154552540-154552540 on Build GRCh38
rs540223445 Chr.1: 154552389-154552389 on Build GRCh38
rs779443130 Chr.1: 154552721-154552721 on Build GRCh38
rs976817697 Chr.1: 154552586-154552586 on Build GRCh38
rs992414346 Chr.1: 154552561-154552561 on Build GRCh38
rs765440474 Chr.1: 154552377-154552377 on Build GRCh38
rs921100972 Chr.1: 154552683-154552684 on Build GRCh38
rs556868100 Chr.1: 154552719-154552719 on Build GRCh38
rs12144146 Chr.1: 154552497-154552497 on Build GRCh38
rs760690764 Chr.1: 154552511-154552511 on Build GRCh38
rs373461148 Chr.1: 154552547-154552547 on Build GRCh38
rs77412635 Chr.1: 154552555-154552555 on Build GRCh38
rs894316227 Chr.1: 154552475-154552475 on Build GRCh38
rs374287641 Chr.1: 154552482-154552482 on Build GRCh38
rs113687056 Chr.1: 154552332-154552332 on Build GRCh38
rs965462586 Chr.1: 154552559-154552559 on Build GRCh38
rs370466044 Chr.1: 154552445-154552445 on Build GRCh38
rs894189614 Chr.1: 154552765-154552765 on Build GRCh38
rs753664930 Chr.1: 154552514-154552515 on Build GRCh38
rs938302137 Chr.1: 154552717-154552717 on Build GRCh38
rs752610086 Chr.1: 154552391-154552391 on Build GRCh38
rs763990597 Chr.1: 154552444-154552444 on Build GRCh38
rs1045144991 Chr.1: 154552594-154552594 on Build GRCh38
rs756075808 Chr.1: 154552467-154552467 on Build GRCh38
rs752154185 Chr.1: 154552434-154552435 on Build GRCh38
rs377680876 Chr.1: 154552603-154552603 on Build GRCh38
rs945610728 Chr.1: 154552661-154552661 on Build GRCh38
rs201201500 Chr.1: 154552502-154552502 on Build GRCh38
rs950698403 Chr.1: 154552584-154552584 on Build GRCh38
rs974335212 Chr.1: 154552680-154552680 on Build GRCh38
rs909256799 Chr.1: 154552349-154552349 on Build GRCh38
rs761675191 Chr.1: 154552699-154552699 on Build GRCh38
rs375941844 Chr.1: 154552510-154552510 on Build GRCh38
rs571264795 Chr.1: 154552651-154552651 on Build GRCh38
rs758020772 Chr.1: 154552720-154552720 on Build GRCh38
rs567213683 Chr.1: 154552592-154552592 on Build GRCh38
rs190883331 Chr.1: 154552342-154552342 on Build GRCh38
rs753740632 Chr.1: 154552515-154552515 on Build GRCh38
rs554217579 Chr.1: 154552711-154552711 on Build GRCh38
rs905675555 Chr.1: 154552608-154552608 on Build GRCh38
rs755537885 Chr.1: 154552738-154552738 on Build GRCh38
rs1001460027 Chr.1: 154552538-154552538 on Build GRCh38
rs778094115 Chr.1: 154552520-154552520 on Build GRCh38
rs538532747 Chr.1: 154552665-154552665 on Build GRCh38
rs765020687 Chr.1: 154552710-154552710 on Build GRCh38
rs563057401 Chr.1: 154552523-154552523 on Build GRCh38
rs769040502 Chr.1: 154552367-154552367 on Build GRCh38
rs775836652 Chr.1: 154552363-154552363 on Build GRCh38
rs781649844 Chr.1: 154552756-154552756 on Build GRCh38
rs924263073 Chr.1: 154552706-154552706 on Build GRCh38
rs917911667 Chr.1: 154552327-154552327 on Build GRCh38
rs201993216 Chr.1: 154552468-154552468 on Build GRCh38
rs749997323 Chr.1: 154552472-154552472 on Build GRCh38
rs773340231 Chr.1: 154552379-154552379 on Build GRCh38
rs781604647 Chr.1: 154552494-154552494 on Build GRCh38
rs776931058 Chr.1: 154552368-154552368 on Build GRCh38
rs1025434749 Chr.1: 154552575-154552575 on Build GRCh38
rs769085788 Chr.1: 154552487-154552487 on Build GRCh38
rs764151960 Chr.1: 154552473-154552474 on Build GRCh38
rs762921384 Chr.1: 154552380-154552380 on Build GRCh38
rs760615541 Chr.1: 154552424-154552424 on Build GRCh38
rs989631119 Chr.1: 154552613-154552613 on Build GRCh38
rs374677299 Chr.1: 154552451-154552451 on Build GRCh38
rs747438755 Chr.1: 154552364-154552364 on Build GRCh38
rs915514158 Chr.1: 154552614-154552614 on Build GRCh38
rs757814009 Chr.1: 154552476-154552476 on Build GRCh38
rs770942689 Chr.1: 154552509-154552509 on Build GRCh38
rs763196683 Chr.1: 154552505-154552505 on Build GRCh38
rs747492338 Chr.1: 154552486-154552486 on Build GRCh38
rs368490109 Chr.1: 154552490-154552490 on Build GRCh38
rs573303733 Chr.1: 154552373-154552373 on Build GRCh38
rs764119144 Chr.1: 154552514-154552514 on Build GRCh38

View Full Product Details