Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155178324-155178819 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM46
Entrez Gene ID
80128
Gene Name
tripartite motif containing 46
Gene Aliases
GENEY, TRIFIC
UniGene
Hs.287735
Gene Chromosome Location
Chr.1: 155173714-155184971 on Build GRCh38
Transcripts
NM_001256599.1 NM_001256600.1 NM_001256601.1 NM_001282378.1 NM_001282379.1 NM_025058.4 NR_046327.1 NR_046329.1 NR_104150.1 XM_011510002.1 XM_017002393.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6808285 c.1171C>T p.R391* Chr.1: 155178499-155178499 on Build GRCh38
1334564 c.1282C>T p.R428* Chr.1: 155178610-155178610 on Build GRCh38
5651211 c.1240G>A p.V414M Chr.1: 155178568-155178568 on Build GRCh38
7206856 c.1226A>G p.H409R Chr.1: 155178554-155178554 on Build GRCh38
424032 c.1184delC p.L396fs*23 Chr.1: 155178512-155178512 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043986975 Chr.1: 155178673-155178673 on Build GRCh38
rs759270899 Chr.1: 155178624-155178624 on Build GRCh38
rs781105980 Chr.1: 155178527-155178527 on Build GRCh38
rs12737132 Chr.1: 155178478-155178478 on Build GRCh38
rs780249356 Chr.1: 155178774-155178774 on Build GRCh38
rs535055009 Chr.1: 155178765-155178765 on Build GRCh38
rs1002694669 Chr.1: 155178432-155178432 on Build GRCh38
rs748001343 Chr.1: 155178539-155178539 on Build GRCh38
rs914224883 Chr.1: 155178725-155178725 on Build GRCh38
rs762208041 Chr.1: 155178575-155178575 on Build GRCh38
rs762450576 Chr.1: 155178496-155178496 on Build GRCh38
rs1056897741 Chr.1: 155178434-155178434 on Build GRCh38
rs902734305 Chr.1: 155178697-155178697 on Build GRCh38
rs751306680 Chr.1: 155178511-155178511 on Build GRCh38
rs747314139 Chr.1: 155178568-155178568 on Build GRCh38
rs567301894 Chr.1: 155178650-155178650 on Build GRCh38
rs960746929 Chr.1: 155178721-155178721 on Build GRCh38
rs756004751 Chr.1: 155178652-155178652 on Build GRCh38
rs368783002 Chr.1: 155178611-155178611 on Build GRCh38
rs777095970 Chr.1: 155178574-155178574 on Build GRCh38
rs772215425 Chr.1: 155178565-155178565 on Build GRCh38
rs778419282 Chr.1: 155178402-155178402 on Build GRCh38
rs902673990 Chr.1: 155178486-155178486 on Build GRCh38
rs553621506 Chr.1: 155178773-155178773 on Build GRCh38
rs150155187 Chr.1: 155178572-155178572 on Build GRCh38
rs772812574 Chr.1: 155178598-155178598 on Build GRCh38
rs994059248 Chr.1: 155178489-155178489 on Build GRCh38
rs1032486707 Chr.1: 155178743-155178743 on Build GRCh38
rs989486183 Chr.1: 155178718-155178718 on Build GRCh38
rs923438757 Chr.1: 155178642-155178642 on Build GRCh38
rs12750342 Chr.1: 155178359-155178359 on Build GRCh38
rs745978771 Chr.1: 155178551-155178551 on Build GRCh38
rs865802058 Chr.1: 155178752-155178752 on Build GRCh38
rs770449098 Chr.1: 155178579-155178579 on Build GRCh38
rs999801362 Chr.1: 155178703-155178703 on Build GRCh38
rs199614812 Chr.1: 155178667-155178667 on Build GRCh38
rs754712280 Chr.1: 155178526-155178526 on Build GRCh38
rs12736969 Chr.1: 155178362-155178362 on Build GRCh38
rs767193360 Chr.1: 155178640-155178640 on Build GRCh38
rs555376790 Chr.1: 155178620-155178620 on Build GRCh38
rs766165172 Chr.1: 155178502-155178502 on Build GRCh38
rs769057226 Chr.1: 155178763-155178763 on Build GRCh38
rs562410050 Chr.1: 155178500-155178500 on Build GRCh38
rs373216490 Chr.1: 155178643-155178643 on Build GRCh38
rs984002287 Chr.1: 155178403-155178403 on Build GRCh38
rs897045040 Chr.1: 155178456-155178456 on Build GRCh38
rs780154098 Chr.1: 155178567-155178567 on Build GRCh38
rs762560404 Chr.1: 155178608-155178608 on Build GRCh38
rs905681836 Chr.1: 155178428-155178428 on Build GRCh38
rs569836274 Chr.1: 155178357-155178357 on Build GRCh38
rs778912457 Chr.1: 155178550-155178550 on Build GRCh38
rs909815271 Chr.1: 155178433-155178433 on Build GRCh38
rs1026291227 Chr.1: 155178494-155178494 on Build GRCh38
rs779217295 Chr.1: 155178764-155178764 on Build GRCh38
rs1044562201 Chr.1: 155178415-155178415 on Build GRCh38
rs1044142775 Chr.1: 155178775-155178775 on Build GRCh38
rs1035394434 Chr.1: 155178720-155178720 on Build GRCh38
rs777691029 Chr.1: 155178657-155178657 on Build GRCh38
rs756091188 Chr.1: 155178545-155178545 on Build GRCh38
rs947133573 Chr.1: 155178373-155178373 on Build GRCh38
rs764812271 Chr.1: 155178499-155178499 on Build GRCh38
rs376790392 Chr.1: 155178564-155178564 on Build GRCh38

View Full Product Details