Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 127440659-127441176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0408
Entrez Gene ID
9729
Gene Name
KIAA0408
Gene Aliases
-
UniGene
Hs.319247
Gene Chromosome Location
Chr.6: 127438406-127459390 on Build GRCh38
Transcripts
NM_014702.4 NM_001012279.2

Gene Symbol
SOGA3
Entrez Gene ID
387104
Gene Name
SOGA family member 3
Gene Aliases
C6orf174, dJ403A15.3
UniGene
Hs.319247
Gene Chromosome Location
Chr.6: 127438406-127519355 on Build GRCh38
Transcripts
NM_014702.4 NM_001012279.2

SNP Detail

SNP ID Chromosome Location
rs1048300157 Chr.6: 127440853-127440853 on Build GRCh38
rs1004014086 Chr.6: 127440865-127440865 on Build GRCh38
rs959362891 Chr.6: 127440918-127440918 on Build GRCh38
rs11450965 Chr.6: 127440697-127440698 on Build GRCh38
rs754832632 Chr.6: 127440830-127440830 on Build GRCh38
rs397816106 Chr.6: 127440704-127440705 on Build GRCh38
rs989707777 Chr.6: 127440929-127440929 on Build GRCh38
rs6935096 Chr.6: 127440723-127440723 on Build GRCh38
rs894044483 Chr.6: 127440911-127440911 on Build GRCh38
rs568872310 Chr.6: 127440921-127440921 on Build GRCh38
rs909789468 Chr.6: 127441143-127441143 on Build GRCh38
rs552209744 Chr.6: 127441022-127441022 on Build GRCh38
rs1004683676 Chr.6: 127441142-127441142 on Build GRCh38
rs911987550 Chr.6: 127440845-127440845 on Build GRCh38
rs569788586 Chr.6: 127440724-127440724 on Build GRCh38
rs1022567961 Chr.6: 127440930-127440930 on Build GRCh38
rs912087382 Chr.6: 127441021-127441021 on Build GRCh38
rs978268133 Chr.6: 127440971-127440971 on Build GRCh38
rs1001215369 Chr.6: 127440991-127440991 on Build GRCh38
rs932335722 Chr.6: 127441139-127441139 on Build GRCh38
rs1035415125 Chr.6: 127441019-127441019 on Build GRCh38
rs973802560 Chr.6: 127440751-127440751 on Build GRCh38
rs906599255 Chr.6: 127440900-127440900 on Build GRCh38
rs920898303 Chr.6: 127441136-127441141 on Build GRCh38
rs905557500 Chr.6: 127440961-127440961 on Build GRCh38
rs1052562996 Chr.6: 127440907-127440907 on Build GRCh38
rs1011198209 Chr.6: 127440937-127440937 on Build GRCh38
rs888255178 Chr.6: 127440860-127440860 on Build GRCh38
rs1033652492 Chr.6: 127440917-127440917 on Build GRCh38
rs935475922 Chr.6: 127440850-127440850 on Build GRCh38
rs185331694 Chr.6: 127440835-127440835 on Build GRCh38
rs1043099564 Chr.6: 127440945-127440945 on Build GRCh38
rs35540131 Chr.6: 127441122-127441123 on Build GRCh38
rs976913308 Chr.6: 127441088-127441088 on Build GRCh38
rs1015870724 Chr.6: 127440868-127440868 on Build GRCh38
rs1004075051 Chr.6: 127440917-127440917 on Build GRCh38
rs866146456 Chr.6: 127440888-127440888 on Build GRCh38
rs921995116 Chr.6: 127440816-127440816 on Build GRCh38
rs966982226 Chr.6: 127440939-127440939 on Build GRCh38
rs778415904 Chr.6: 127440990-127440990 on Build GRCh38
rs1040715949 Chr.6: 127440754-127440754 on Build GRCh38
rs571326163 Chr.6: 127440721-127440721 on Build GRCh38

View Full Product Details