Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 127446682-127447183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0408
Entrez Gene ID
9729
Gene Name
KIAA0408
Gene Aliases
-
UniGene
Hs.319247
Gene Chromosome Location
Chr.6: 127438406-127459390 on Build GRCh38
Transcripts
NM_014702.4 NM_001012279.2

Gene Symbol
SOGA3
Entrez Gene ID
387104
Gene Name
SOGA family member 3
Gene Aliases
C6orf174, dJ403A15.3
UniGene
Hs.319247
Gene Chromosome Location
Chr.6: 127438406-127519355 on Build GRCh38
Transcripts
NM_014702.4 NM_001012279.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
312265 c.1153C>A p.P385T Chr.6: 127447166-127447166 on Build GRCh38
6514367 c.1631C>T p.P544L Chr.6: 127446688-127446688 on Build GRCh38
6514368 c.1389C>T p.H463H Chr.6: 127446930-127446930 on Build GRCh38
7310941 c.1239C>T p.S413S Chr.6: 127447080-127447080 on Build GRCh38
3171917 c.1523T>G p.V508G Chr.6: 127446796-127446796 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772115810 Chr.6: 127447086-127447086 on Build GRCh38
rs751803238 Chr.6: 127446828-127446828 on Build GRCh38
rs765282504 Chr.6: 127447124-127447124 on Build GRCh38
rs748677689 Chr.6: 127446859-127446859 on Build GRCh38
rs765203412 Chr.6: 127446764-127446764 on Build GRCh38
rs775807602 Chr.6: 127447087-127447087 on Build GRCh38
rs749886586 Chr.6: 127447128-127447128 on Build GRCh38
rs763922338 Chr.6: 127447097-127447097 on Build GRCh38
rs762881412 Chr.6: 127447100-127447101 on Build GRCh38
rs146403375 Chr.6: 127446763-127446763 on Build GRCh38
rs373650400 Chr.6: 127446964-127446964 on Build GRCh38
rs17054687 Chr.6: 127446759-127446759 on Build GRCh38
rs781112554 Chr.6: 127446725-127446725 on Build GRCh38
rs61735562 Chr.6: 127446831-127446831 on Build GRCh38
rs751289424 Chr.6: 127446776-127446776 on Build GRCh38
rs79160411 Chr.6: 127447031-127447031 on Build GRCh38
rs774453923 Chr.6: 127446965-127446965 on Build GRCh38
rs768525449 Chr.6: 127446719-127446719 on Build GRCh38
rs753756483 Chr.6: 127447100-127447100 on Build GRCh38
rs998982732 Chr.6: 127446747-127446747 on Build GRCh38
rs756114209 Chr.6: 127446792-127446792 on Build GRCh38
rs376092824 Chr.6: 127446805-127446805 on Build GRCh38
rs181271629 Chr.6: 127447101-127447101 on Build GRCh38
rs748013416 Chr.6: 127446789-127446789 on Build GRCh38
rs188314558 Chr.6: 127446720-127446720 on Build GRCh38
rs761129644 Chr.6: 127446758-127446758 on Build GRCh38
rs538723456 Chr.6: 127447007-127447007 on Build GRCh38
rs757302813 Chr.6: 127447013-127447013 on Build GRCh38
rs975105667 Chr.6: 127447083-127447083 on Build GRCh38
rs150637496 Chr.6: 127446801-127446801 on Build GRCh38
rs749710325 Chr.6: 127447160-127447160 on Build GRCh38
rs79339979 Chr.6: 127446794-127446794 on Build GRCh38
rs778049995 Chr.6: 127447038-127447038 on Build GRCh38
rs778104905 Chr.6: 127447151-127447151 on Build GRCh38
rs1026943266 Chr.6: 127446881-127446881 on Build GRCh38
rs372590924 Chr.6: 127446766-127446766 on Build GRCh38
rs773109864 Chr.6: 127446822-127446822 on Build GRCh38
rs776849728 Chr.6: 127446906-127446906 on Build GRCh38
rs759748148 Chr.6: 127446975-127446975 on Build GRCh38
rs771492160 Chr.6: 127447057-127447057 on Build GRCh38
rs368538507 Chr.6: 127446752-127446752 on Build GRCh38
rs748560373 Chr.6: 127446912-127446912 on Build GRCh38
rs750015390 Chr.6: 127447021-127447021 on Build GRCh38
rs751204090 Chr.6: 127447140-127447140 on Build GRCh38
rs371074447 Chr.6: 127447026-127447026 on Build GRCh38
rs754799569 Chr.6: 127446777-127446777 on Build GRCh38
rs1019643988 Chr.6: 127446864-127446864 on Build GRCh38
rs762224628 Chr.6: 127446812-127446812 on Build GRCh38
rs777777621 Chr.6: 127446852-127446852 on Build GRCh38
rs78892092 Chr.6: 127446739-127446739 on Build GRCh38
rs758537917 Chr.6: 127446768-127446768 on Build GRCh38
rs373351607 Chr.6: 127447041-127447041 on Build GRCh38
rs575039191 Chr.6: 127446840-127446840 on Build GRCh38
rs541088934 Chr.6: 127446748-127446748 on Build GRCh38
rs766548239 Chr.6: 127446827-127446827 on Build GRCh38
rs770772164 Chr.6: 127446744-127446744 on Build GRCh38
rs535064718 Chr.6: 127446867-127446867 on Build GRCh38
rs770219441 Chr.6: 127447035-127447035 on Build GRCh38
rs779461041 Chr.6: 127446888-127446888 on Build GRCh38
rs778564256 Chr.6: 127447019-127447019 on Build GRCh38
rs200967734 Chr.6: 127446995-127446995 on Build GRCh38
rs779822528 Chr.6: 127446718-127446718 on Build GRCh38
rs549658342 Chr.6: 127446823-127446823 on Build GRCh38
rs764723520 Chr.6: 127446933-127446934 on Build GRCh38
rs375751864 Chr.6: 127447146-127447146 on Build GRCh38
rs762971806 Chr.6: 127446958-127446958 on Build GRCh38
rs745488624 Chr.6: 127446880-127446880 on Build GRCh38
rs760336244 Chr.6: 127447090-127447090 on Build GRCh38
rs758069275 Chr.6: 127447022-127447022 on Build GRCh38
rs199642157 Chr.6: 127446876-127446876 on Build GRCh38
rs758690216 Chr.6: 127446712-127446712 on Build GRCh38
rs759625356 Chr.6: 127447085-127447085 on Build GRCh38
rs779762074 Chr.6: 127447025-127447025 on Build GRCh38
rs1044568314 Chr.6: 127446868-127446868 on Build GRCh38
rs770135090 Chr.6: 127446938-127446938 on Build GRCh38
rs757957661 Chr.6: 127447129-127447129 on Build GRCh38
rs773799835 Chr.6: 127446954-127446954 on Build GRCh38
rs571746345 Chr.6: 127446983-127446983 on Build GRCh38
rs893051006 Chr.6: 127446829-127446829 on Build GRCh38
rs752602386 Chr.6: 127446843-127446843 on Build GRCh38
rs771918935 Chr.6: 127446885-127446885 on Build GRCh38
rs775465481 Chr.6: 127447100-127447100 on Build GRCh38
rs780270394 Chr.6: 127446771-127446771 on Build GRCh38
rs1004284105 Chr.6: 127447009-127447009 on Build GRCh38
rs907335755 Chr.6: 127446875-127446875 on Build GRCh38
rs970899872 Chr.6: 127447070-127447070 on Build GRCh38
rs975380090 Chr.6: 127446904-127446904 on Build GRCh38
rs147193772 Chr.6: 127447136-127447136 on Build GRCh38
rs747655022 Chr.6: 127446727-127446727 on Build GRCh38
rs765834146 Chr.6: 127446820-127446820 on Build GRCh38
rs941153948 Chr.6: 127446705-127446705 on Build GRCh38
rs755991192 Chr.6: 127446850-127446850 on Build GRCh38
rs767837088 Chr.6: 127446976-127446976 on Build GRCh38
rs200170150 Chr.6: 127446927-127446927 on Build GRCh38
rs767185394 Chr.6: 127446842-127446842 on Build GRCh38
rs774400628 Chr.6: 127447068-127447068 on Build GRCh38
rs267600792 Chr.6: 127446979-127446979 on Build GRCh38
rs763895159 Chr.6: 127447000-127447000 on Build GRCh38
rs759009069 Chr.6: 127446818-127446819 on Build GRCh38
rs755937723 Chr.6: 127446996-127446996 on Build GRCh38
rs772449495 Chr.6: 127447117-127447117 on Build GRCh38
rs772827319 Chr.6: 127446728-127446728 on Build GRCh38
rs139745911 Chr.6: 127446809-127446809 on Build GRCh38
rs201499487 Chr.6: 127446783-127446783 on Build GRCh38
rs376131588 Chr.6: 127447004-127447004 on Build GRCh38

View Full Product Details