Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 137875547-137876061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFAIP3
Entrez Gene ID
7128
Gene Name
TNF alpha induced protein 3
Gene Aliases
A20, AISBL, OTUD7C, TNFA1P2
UniGene
Hs.211600
Gene Chromosome Location
Chr.6: 137866317-137883314 on Build GRCh38
Transcripts
NM_001270507.1 NM_001270508.1 NM_006290.3 XM_005267119.1 XM_006715555.1 XM_011536095.1 XM_011536096.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5404454 c.595C>T p.L199F Chr.6: 137875796-137875796 on Build GRCh38
1581846 c.615G>T p.R205S Chr.6: 137875816-137875816 on Build GRCh38
1132027 c.635A>G p.D212G Chr.6: 137875996-137875996 on Build GRCh38
35938 c.491_492insA p.W164fs*1 Chr.6: 137875692-137875693 on Build GRCh38
5869403 c.539C>T p.P180L Chr.6: 137875740-137875740 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752174190 Chr.6: 137875662-137875663 on Build GRCh38
rs768949689 Chr.6: 137875965-137875965 on Build GRCh38
rs765193387 Chr.6: 137875826-137875826 on Build GRCh38
rs375378882 Chr.6: 137875749-137875749 on Build GRCh38
rs145706326 Chr.6: 137876002-137876002 on Build GRCh38
rs377402222 Chr.6: 137875951-137875951 on Build GRCh38
rs748806273 Chr.6: 137875867-137875867 on Build GRCh38
rs370381369 Chr.6: 137876033-137876033 on Build GRCh38
rs5029947 Chr.6: 137875680-137875680 on Build GRCh38
rs756258288 Chr.6: 137875661-137875661 on Build GRCh38
rs547067487 Chr.6: 137875966-137875966 on Build GRCh38
rs864321682 Chr.6: 137876032-137876032 on Build GRCh38
rs746800027 Chr.6: 137875834-137875834 on Build GRCh38
rs950049665 Chr.6: 137875938-137875938 on Build GRCh38
rs768012213 Chr.6: 137875818-137875818 on Build GRCh38
rs776217388 Chr.6: 137875746-137875746 on Build GRCh38
rs780878996 Chr.6: 137875837-137875837 on Build GRCh38
rs200007148 Chr.6: 137875638-137875638 on Build GRCh38
rs779928915 Chr.6: 137875716-137875716 on Build GRCh38
rs373331557 Chr.6: 137875710-137875710 on Build GRCh38
rs758353383 Chr.6: 137875829-137875829 on Build GRCh38
rs772637841 Chr.6: 137875858-137875858 on Build GRCh38
rs761876876 Chr.6: 137875969-137875969 on Build GRCh38
rs974398639 Chr.6: 137875952-137875952 on Build GRCh38
rs776043863 Chr.6: 137875883-137875883 on Build GRCh38
rs761953299 Chr.6: 137875855-137875855 on Build GRCh38
rs747702591 Chr.6: 137875845-137875845 on Build GRCh38
rs772553813 Chr.6: 137875881-137875881 on Build GRCh38
rs531456508 Chr.6: 137875850-137875850 on Build GRCh38
rs780201198 Chr.6: 137875833-137875833 on Build GRCh38
rs370174617 Chr.6: 137875753-137875753 on Build GRCh38
rs554058322 Chr.6: 137875612-137875612 on Build GRCh38
rs750518190 Chr.6: 137876007-137876007 on Build GRCh38
rs899059690 Chr.6: 137875807-137875807 on Build GRCh38
rs973575962 Chr.6: 137875799-137875799 on Build GRCh38
rs537289140 Chr.6: 137875654-137875654 on Build GRCh38
rs751503246 Chr.6: 137876034-137876034 on Build GRCh38
rs750536183 Chr.6: 137875828-137875828 on Build GRCh38
rs753940311 Chr.6: 137875677-137875677 on Build GRCh38
rs1002015155 Chr.6: 137875579-137875579 on Build GRCh38
rs893707432 Chr.6: 137875651-137875651 on Build GRCh38
rs201505674 Chr.6: 137876017-137876017 on Build GRCh38
rs754771774 Chr.6: 137875836-137875836 on Build GRCh38
rs754045894 Chr.6: 137875823-137875823 on Build GRCh38
rs1018691124 Chr.6: 137875922-137875922 on Build GRCh38
rs372145581 Chr.6: 137875676-137875676 on Build GRCh38
rs147932545 Chr.6: 137875752-137875752 on Build GRCh38
rs141807543 Chr.6: 137875820-137875820 on Build GRCh38
rs753087450 Chr.6: 137875645-137875645 on Build GRCh38
rs779262522 Chr.6: 137875890-137875890 on Build GRCh38
rs768346040 Chr.6: 137875743-137875743 on Build GRCh38
rs781171267 Chr.6: 137875979-137875982 on Build GRCh38
rs561335450 Chr.6: 137875931-137875931 on Build GRCh38
rs746814438 Chr.6: 137875739-137875739 on Build GRCh38
rs764373111 Chr.6: 137875822-137875822 on Build GRCh38
rs775746637 Chr.6: 137875581-137875581 on Build GRCh38
rs34935799 Chr.6: 137875759-137875760 on Build GRCh38
rs201927262 Chr.6: 137875856-137875856 on Build GRCh38
rs199833786 Chr.6: 137875644-137875644 on Build GRCh38
rs757390153 Chr.6: 137875815-137875816 on Build GRCh38
rs765570875 Chr.6: 137875973-137875973 on Build GRCh38
rs759993608 Chr.6: 137875767-137875767 on Build GRCh38
rs34143521 Chr.6: 137875670-137875670 on Build GRCh38
rs745752476 Chr.6: 137875685-137875685 on Build GRCh38

View Full Product Details