Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 149512033-149512547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPIL4
Entrez Gene ID
85313
Gene Name
peptidylprolyl isomerase like 4
Gene Aliases
HDCME13P
UniGene
Hs.744238
Gene Chromosome Location
Chr.6: 149504495-149546102 on Build GRCh38
Transcripts
NM_139126.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6260189 c.1082C>A p.T361K Chr.6: 149512300-149512300 on Build GRCh38
1683174 c.1096_1101delATATTA p.I366_L367delIL Chr.6: 149512281-149512286 on Build GRCh38
1496285 c.1146T>A p.S382R Chr.6: 149512236-149512236 on Build GRCh38
4716947 c.1155_1156delCA p.H385fs*9 Chr.6: 149512226-149512227 on Build GRCh38
1074419 c.1116A>C p.E372D Chr.6: 149512266-149512266 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149512214 Chr.6: 149512182-149512182 on Build GRCh38
rs374144563 Chr.6: 149512288-149512288 on Build GRCh38
rs535267469 Chr.6: 149512281-149512286 on Build GRCh38
rs758767193 Chr.6: 149512168-149512168 on Build GRCh38
rs554507594 Chr.6: 149512312-149512312 on Build GRCh38
rs200526214 Chr.6: 149512192-149512192 on Build GRCh38
rs749243736 Chr.6: 149512331-149512334 on Build GRCh38
rs530165275 Chr.6: 149512070-149512070 on Build GRCh38
rs755428882 Chr.6: 149512285-149512285 on Build GRCh38
rs183200782 Chr.6: 149512229-149512229 on Build GRCh38
rs201100357 Chr.6: 149512144-149512144 on Build GRCh38
rs759764488 Chr.6: 149512082-149512082 on Build GRCh38
rs996060186 Chr.6: 149512162-149512162 on Build GRCh38
rs1035201890 Chr.6: 149512497-149512497 on Build GRCh38
rs892992336 Chr.6: 149512421-149512421 on Build GRCh38
rs747835467 Chr.6: 149512214-149512214 on Build GRCh38
rs760656623 Chr.6: 149512227-149512227 on Build GRCh38
rs765310002 Chr.6: 149512268-149512268 on Build GRCh38
rs945773491 Chr.6: 149512424-149512424 on Build GRCh38
rs770988898 Chr.6: 149512352-149512352 on Build GRCh38
rs144132135 Chr.6: 149512185-149512185 on Build GRCh38
rs1054170736 Chr.6: 149512064-149512064 on Build GRCh38
rs867100116 Chr.6: 149512240-149512240 on Build GRCh38
rs1041501509 Chr.6: 149512434-149512434 on Build GRCh38
rs150616132 Chr.6: 149512342-149512342 on Build GRCh38
rs769644035 Chr.6: 149512148-149512156 on Build GRCh38
rs1054713287 Chr.6: 149512369-149512369 on Build GRCh38
rs377351697 Chr.6: 149512215-149512215 on Build GRCh38
rs779511178 Chr.6: 149512186-149512186 on Build GRCh38
rs944363209 Chr.6: 149512119-149512119 on Build GRCh38
rs771891901 Chr.6: 149512337-149512337 on Build GRCh38
rs368515515 Chr.6: 149512293-149512293 on Build GRCh38
rs758717817 Chr.6: 149512283-149512283 on Build GRCh38
rs369166335 Chr.6: 149512118-149512118 on Build GRCh38
rs781754938 Chr.6: 149512303-149512303 on Build GRCh38
rs114868889 Chr.6: 149512130-149512130 on Build GRCh38
rs774053905 Chr.6: 149512213-149512213 on Build GRCh38
rs777686567 Chr.6: 149512341-149512341 on Build GRCh38
rs773291927 Chr.6: 149512104-149512104 on Build GRCh38
rs201670613 Chr.6: 149512217-149512217 on Build GRCh38
rs912797530 Chr.6: 149512471-149512471 on Build GRCh38
rs1002496780 Chr.6: 149512469-149512469 on Build GRCh38
rs141874064 Chr.6: 149512255-149512255 on Build GRCh38
rs371642016 Chr.6: 149512269-149512269 on Build GRCh38
rs748883421 Chr.6: 149512188-149512188 on Build GRCh38
rs778198794 Chr.6: 149512180-149512180 on Build GRCh38
rs148702522 Chr.6: 149512218-149512218 on Build GRCh38
rs765555651 Chr.6: 149512150-149512150 on Build GRCh38
rs900260005 Chr.6: 149512161-149512161 on Build GRCh38
rs776561241 Chr.6: 149512235-149512235 on Build GRCh38
rs9386252 Chr.6: 149512107-149512107 on Build GRCh38
rs778377257 Chr.6: 149512322-149512322 on Build GRCh38
rs866875509 Chr.6: 149512435-149512435 on Build GRCh38
rs906875719 Chr.6: 149512460-149512460 on Build GRCh38
rs747864994 Chr.6: 149512327-149512327 on Build GRCh38
rs975956068 Chr.6: 149512340-149512340 on Build GRCh38
rs768187193 Chr.6: 149512206-149512206 on Build GRCh38
rs969452261 Chr.6: 149512320-149512320 on Build GRCh38
rs530426424 Chr.6: 149512160-149512160 on Build GRCh38
rs925403375 Chr.6: 149512348-149512348 on Build GRCh38
rs775573890 Chr.6: 149512368-149512368 on Build GRCh38
rs370213593 Chr.6: 149512243-149512243 on Build GRCh38

View Full Product Details