Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 166934008-166934515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNASET2
Entrez Gene ID
8635
Gene Name
ribonuclease T2
Gene Aliases
RNASE6PL, bA514O12.3
UniGene
Hs.529989
Gene Chromosome Location
Chr.6: 166929509-166956589 on Build GRCh38
Transcripts
NM_003730.4 XM_017011397.1 XM_017011398.1 XM_017011399.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5780631 c.481A>G p.N161D Chr.6: 166934102-166934102 on Build GRCh38
741163 c.450G>A p.V150V Chr.6: 166934133-166934133 on Build GRCh38
3697669 c.447-1G>A p.? Chr.6: 166934137-166934137 on Build GRCh38
3697668 c.482A>G p.N161S Chr.6: 166934101-166934101 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs113477891 Chr.6: 166934396-166934396 on Build GRCh38
rs769475041 Chr.6: 166934048-166934048 on Build GRCh38
rs1004281681 Chr.6: 166934256-166934256 on Build GRCh38
rs1018730572 Chr.6: 166934089-166934089 on Build GRCh38
rs376512986 Chr.6: 166934471-166934471 on Build GRCh38
rs755860620 Chr.6: 166934064-166934064 on Build GRCh38
rs998561561 Chr.6: 166934203-166934203 on Build GRCh38
rs905749598 Chr.6: 166934269-166934269 on Build GRCh38
rs914390094 Chr.6: 166934087-166934087 on Build GRCh38
rs569263361 Chr.6: 166934320-166934320 on Build GRCh38
rs1013450644 Chr.6: 166934451-166934451 on Build GRCh38
rs962843041 Chr.6: 166934268-166934268 on Build GRCh38
rs774457054 Chr.6: 166934059-166934059 on Build GRCh38
rs749067371 Chr.6: 166934150-166934150 on Build GRCh38
rs1003111171 Chr.6: 166934479-166934479 on Build GRCh38
rs771231451 Chr.6: 166934135-166934135 on Build GRCh38
rs774372972 Chr.6: 166934162-166934162 on Build GRCh38
rs766632251 Chr.6: 166934071-166934071 on Build GRCh38
rs917369698 Chr.6: 166934254-166934254 on Build GRCh38
rs748671487 Chr.6: 166934109-166934109 on Build GRCh38
rs761898019 Chr.6: 166934167-166934167 on Build GRCh38
rs35419028 Chr.6: 166934279-166934280 on Build GRCh38
rs371620473 Chr.6: 166934243-166934244 on Build GRCh38
rs898588276 Chr.6: 166934444-166934444 on Build GRCh38
rs3823208 Chr.6: 166934358-166934358 on Build GRCh38
rs200299596 Chr.6: 166934051-166934051 on Build GRCh38
rs767129334 Chr.6: 166934170-166934170 on Build GRCh38
rs917294804 Chr.6: 166934440-166934440 on Build GRCh38
rs956230506 Chr.6: 166934415-166934415 on Build GRCh38
rs982139597 Chr.6: 166934346-166934346 on Build GRCh38
rs550915754 Chr.6: 166934313-166934313 on Build GRCh38
rs115770362 Chr.6: 166934384-166934384 on Build GRCh38
rs989282013 Chr.6: 166934218-166934218 on Build GRCh38
rs142218956 Chr.6: 166934157-166934157 on Build GRCh38
rs533016872 Chr.6: 166934411-166934411 on Build GRCh38
rs1023603501 Chr.6: 166934322-166934322 on Build GRCh38
rs772829306 Chr.6: 166934171-166934171 on Build GRCh38
rs151005716 Chr.6: 166934032-166934032 on Build GRCh38
rs887146781 Chr.6: 166934233-166934233 on Build GRCh38
rs536551271 Chr.6: 166934442-166934442 on Build GRCh38
rs924280036 Chr.6: 166934348-166934348 on Build GRCh38
rs758461719 Chr.6: 166934111-166934111 on Build GRCh38
rs575291808 Chr.6: 166934412-166934412 on Build GRCh38
rs965534779 Chr.6: 166934127-166934127 on Build GRCh38
rs762824295 Chr.6: 166934166-166934167 on Build GRCh38
rs754184305 Chr.6: 166934073-166934073 on Build GRCh38
rs201087711 Chr.6: 166934063-166934063 on Build GRCh38
rs769412781 Chr.6: 166934187-166934187 on Build GRCh38
rs768949865 Chr.6: 166934210-166934210 on Build GRCh38
rs114661525 Chr.6: 166934383-166934383 on Build GRCh38
rs145934320 Chr.6: 166934468-166934468 on Build GRCh38
rs761018044 Chr.6: 166934047-166934047 on Build GRCh38
rs766015176 Chr.6: 166934179-166934179 on Build GRCh38
rs773368914 Chr.6: 166934044-166934044 on Build GRCh38
rs747165370 Chr.6: 166934133-166934133 on Build GRCh38
rs1038439689 Chr.6: 166934443-166934443 on Build GRCh38
rs564700188 Chr.6: 166934128-166934128 on Build GRCh38
rs779325837 Chr.6: 166934106-166934106 on Build GRCh38
rs77834464 Chr.6: 166934175-166934175 on Build GRCh38
rs41269597 Chr.6: 166934185-166934185 on Build GRCh38
rs539510219 Chr.6: 166934419-166934419 on Build GRCh38
rs760127843 Chr.6: 166934172-166934172 on Build GRCh38
rs777049117 Chr.6: 166934072-166934072 on Build GRCh38
rs895638648 Chr.6: 166934031-166934031 on Build GRCh38
rs199674368 Chr.6: 166934101-166934101 on Build GRCh38
rs41269599 Chr.6: 166934391-166934391 on Build GRCh38
rs887094362 Chr.6: 166934439-166934439 on Build GRCh38
rs189958566 Chr.6: 166934376-166934376 on Build GRCh38
rs775392413 Chr.6: 166934049-166934049 on Build GRCh38
rs753408417 Chr.6: 166934394-166934394 on Build GRCh38
rs575265045 Chr.6: 166934062-166934062 on Build GRCh38
rs981139021 Chr.6: 166934038-166934038 on Build GRCh38
rs58163761 Chr.6: 166934330-166934330 on Build GRCh38
rs185397741 Chr.6: 166934257-166934257 on Build GRCh38
rs372788481 Chr.6: 166934052-166934052 on Build GRCh38
rs886086861 Chr.6: 166934355-166934355 on Build GRCh38
rs994479828 Chr.6: 166934050-166934050 on Build GRCh38
rs752734332 Chr.6: 166934478-166934478 on Build GRCh38
rs1051096148 Chr.6: 166934208-166934208 on Build GRCh38
rs556868642 Chr.6: 166934401-166934401 on Build GRCh38
rs988804215 Chr.6: 166934435-166934435 on Build GRCh38
rs1044214376 Chr.6: 166934260-166934260 on Build GRCh38

View Full Product Details