Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 36154851-36155362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEPD1
Entrez Gene ID
80820
Gene Name
endonuclease/exonuclease/phosphatase family domain containing 1
Gene Aliases
HSPC107
UniGene
Hs.487994
Gene Chromosome Location
Chr.7: 36153149-36301543 on Build GRCh38
Transcripts
NM_030636.2 XM_017012657.1 XM_017012658.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6243387 c.612C>T p.S204S Chr.7: 36154936-36154936 on Build GRCh38
6177541 c.537G>T p.R179S Chr.7: 36154861-36154861 on Build GRCh38
3228771 c.705_706insG p.P238fs*67 Chr.7: 36155029-36155030 on Build GRCh38
5018519 c.706G>A p.G236R Chr.7: 36155030-36155030 on Build GRCh38
1089366 c.862A>G p.T288A Chr.7: 36155186-36155186 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1052206716 Chr.7: 36154932-36154932 on Build GRCh38
rs376724368 Chr.7: 36154945-36154945 on Build GRCh38
rs774203788 Chr.7: 36154992-36154992 on Build GRCh38
rs759264914 Chr.7: 36155154-36155154 on Build GRCh38
rs913741079 Chr.7: 36155320-36155320 on Build GRCh38
rs942346132 Chr.7: 36154876-36154876 on Build GRCh38
rs746792385 Chr.7: 36155244-36155244 on Build GRCh38
rs758943146 Chr.7: 36154934-36154934 on Build GRCh38
rs202070535 Chr.7: 36154878-36154878 on Build GRCh38
rs748690801 Chr.7: 36155216-36155217 on Build GRCh38
rs996172553 Chr.7: 36155270-36155270 on Build GRCh38
rs772990253 Chr.7: 36155118-36155118 on Build GRCh38
rs773010785 Chr.7: 36155027-36155027 on Build GRCh38
rs780290633 Chr.7: 36155191-36155191 on Build GRCh38
rs762345552 Chr.7: 36155169-36155169 on Build GRCh38
rs540375594 Chr.7: 36155141-36155141 on Build GRCh38
rs764632704 Chr.7: 36155061-36155061 on Build GRCh38
rs1014462102 Chr.7: 36155073-36155073 on Build GRCh38
rs778991786 Chr.7: 36155102-36155102 on Build GRCh38
rs753296627 Chr.7: 36154931-36154931 on Build GRCh38
rs369820285 Chr.7: 36155188-36155188 on Build GRCh38
rs752270387 Chr.7: 36155168-36155168 on Build GRCh38
rs748227863 Chr.7: 36155215-36155215 on Build GRCh38
rs759021383 Chr.7: 36155060-36155060 on Build GRCh38
rs563058717 Chr.7: 36155034-36155034 on Build GRCh38
rs904475964 Chr.7: 36155314-36155314 on Build GRCh38
rs753963666 Chr.7: 36155192-36155192 on Build GRCh38
rs774191235 Chr.7: 36154890-36154890 on Build GRCh38
rs191385066 Chr.7: 36155336-36155336 on Build GRCh38
rs1034229596 Chr.7: 36155290-36155290 on Build GRCh38
rs143445715 Chr.7: 36155161-36155161 on Build GRCh38
rs1021625853 Chr.7: 36154958-36154958 on Build GRCh38
rs775572213 Chr.7: 36154967-36154967 on Build GRCh38
rs753853609 Chr.7: 36155263-36155263 on Build GRCh38
rs146751299 Chr.7: 36155181-36155181 on Build GRCh38
rs922538697 Chr.7: 36155284-36155284 on Build GRCh38
rs960301890 Chr.7: 36155057-36155057 on Build GRCh38
rs749365066 Chr.7: 36155098-36155098 on Build GRCh38
rs776508106 Chr.7: 36155249-36155249 on Build GRCh38
rs373110272 Chr.7: 36155090-36155090 on Build GRCh38
rs377390131 Chr.7: 36155111-36155111 on Build GRCh38
rs763941759 Chr.7: 36154880-36154880 on Build GRCh38
rs767255473 Chr.7: 36155013-36155013 on Build GRCh38
rs778904152 Chr.7: 36155213-36155213 on Build GRCh38
rs760257117 Chr.7: 36155031-36155031 on Build GRCh38
rs144236439 Chr.7: 36154930-36154930 on Build GRCh38
rs778205698 Chr.7: 36154944-36154944 on Build GRCh38
rs762608785 Chr.7: 36155252-36155252 on Build GRCh38
rs202088491 Chr.7: 36155251-36155251 on Build GRCh38
rs770599770 Chr.7: 36155140-36155140 on Build GRCh38
rs200749099 Chr.7: 36154875-36154875 on Build GRCh38
rs34444080 Chr.7: 36154974-36154974 on Build GRCh38
rs746040709 Chr.7: 36154960-36154960 on Build GRCh38
rs768204660 Chr.7: 36155170-36155170 on Build GRCh38
rs771986858 Chr.7: 36155232-36155232 on Build GRCh38
rs901085217 Chr.7: 36154898-36154898 on Build GRCh38
rs1052280178 Chr.7: 36155302-36155302 on Build GRCh38
rs187691058 Chr.7: 36155200-36155200 on Build GRCh38
rs757505911 Chr.7: 36155292-36155292 on Build GRCh38
rs141136607 Chr.7: 36154966-36154966 on Build GRCh38
rs572952080 Chr.7: 36155066-36155066 on Build GRCh38
rs781642725 Chr.7: 36155071-36155071 on Build GRCh38
rs776101800 Chr.7: 36155026-36155026 on Build GRCh38
rs1010296170 Chr.7: 36154937-36154937 on Build GRCh38
rs1025833028 Chr.7: 36155074-36155074 on Build GRCh38
rs779402455 Chr.7: 36155029-36155030 on Build GRCh38
rs1001536085 Chr.7: 36155001-36155001 on Build GRCh38
rs375151691 Chr.7: 36155038-36155038 on Build GRCh38
rs757737845 Chr.7: 36154949-36154949 on Build GRCh38
rs373414903 Chr.7: 36154879-36154879 on Build GRCh38
rs761653350 Chr.7: 36154906-36154906 on Build GRCh38
rs368502060 Chr.7: 36155333-36155333 on Build GRCh38
rs756353085 Chr.7: 36155089-36155089 on Build GRCh38
rs755437236 Chr.7: 36155029-36155029 on Build GRCh38
rs111452772 Chr.7: 36155186-36155186 on Build GRCh38
rs749392477 Chr.7: 36154980-36154980 on Build GRCh38
rs200105421 Chr.7: 36155125-36155125 on Build GRCh38
rs760448582 Chr.7: 36155129-36155129 on Build GRCh38
rs527674970 Chr.7: 36155101-36155101 on Build GRCh38
rs373823956 Chr.7: 36155248-36155248 on Build GRCh38
rs753379189 Chr.7: 36155041-36155041 on Build GRCh38
rs755793330 Chr.7: 36154927-36154927 on Build GRCh38
rs200979746 Chr.7: 36155065-36155065 on Build GRCh38
rs750112692 Chr.7: 36154916-36154916 on Build GRCh38
rs759054332 Chr.7: 36155250-36155250 on Build GRCh38
rs761162670 Chr.7: 36155063-36155063 on Build GRCh38
rs777740200 Chr.7: 36155243-36155243 on Build GRCh38
rs769957243 Chr.7: 36154877-36154877 on Build GRCh38
rs768624440 Chr.7: 36154981-36154981 on Build GRCh38
rs371689321 Chr.7: 36154915-36154915 on Build GRCh38
rs200242641 Chr.7: 36154993-36154993 on Build GRCh38

View Full Product Details