Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44516625-44517124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPC1L1
Entrez Gene ID
29881
Gene Name
NPC1 like intracellular cholesterol transporter 1
Gene Aliases
NPC11L1
UniGene
Hs.567486
Gene Chromosome Location
Chr.7: 44512440-44541650 on Build GRCh38
Transcripts
NM_001101648.1 NM_013389.2 XM_011515326.2 XM_011515328.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6498882 c.3499C>G p.L1167V Chr.7: 44516804-44516804 on Build GRCh38
3262739 c.3393G>C p.Q1131H Chr.7: 44516910-44516910 on Build GRCh38
4961176 c.3531G>A p.V1177V Chr.7: 44516772-44516772 on Build GRCh38
1580187 c.3547A>G p.M1183V Chr.7: 44516756-44516756 on Build GRCh38
6548221 c.3446C>A p.P1149H Chr.7: 44516857-44516857 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371019585 Chr.7: 44517070-44517070 on Build GRCh38
rs751149157 Chr.7: 44516683-44516683 on Build GRCh38
rs747036704 Chr.7: 44517095-44517095 on Build GRCh38
rs781483583 Chr.7: 44516986-44516986 on Build GRCh38
rs773678788 Chr.7: 44516783-44516783 on Build GRCh38
rs145423806 Chr.7: 44516816-44516816 on Build GRCh38
rs201578123 Chr.7: 44516775-44516775 on Build GRCh38
rs1019436713 Chr.7: 44516999-44516999 on Build GRCh38
rs528352114 Chr.7: 44516704-44516704 on Build GRCh38
rs371006859 Chr.7: 44516679-44516679 on Build GRCh38
rs200207059 Chr.7: 44516810-44516810 on Build GRCh38
rs192247595 Chr.7: 44516946-44516946 on Build GRCh38
rs777480616 Chr.7: 44516820-44516820 on Build GRCh38
rs540523301 Chr.7: 44516754-44516754 on Build GRCh38
rs747201148 Chr.7: 44516870-44516870 on Build GRCh38
rs150978599 Chr.7: 44516832-44516832 on Build GRCh38
rs748562310 Chr.7: 44516874-44516874 on Build GRCh38
rs902225009 Chr.7: 44516830-44516830 on Build GRCh38
rs760440118 Chr.7: 44516824-44516824 on Build GRCh38
rs552498840 Chr.7: 44516876-44516876 on Build GRCh38
rs1054543879 Chr.7: 44517007-44517007 on Build GRCh38
rs894640704 Chr.7: 44516742-44516742 on Build GRCh38
rs759071254 Chr.7: 44516895-44516895 on Build GRCh38
rs746069810 Chr.7: 44516672-44516672 on Build GRCh38
rs753684816 Chr.7: 44516706-44516706 on Build GRCh38
rs570421239 Chr.7: 44517017-44517017 on Build GRCh38
rs371226658 Chr.7: 44516811-44516811 on Build GRCh38
rs377695153 Chr.7: 44516897-44516897 on Build GRCh38
rs558524472 Chr.7: 44516763-44516763 on Build GRCh38
rs948869565 Chr.7: 44517043-44517043 on Build GRCh38
rs377675595 Chr.7: 44516744-44516744 on Build GRCh38
rs545686241 Chr.7: 44517049-44517049 on Build GRCh38
rs775202840 Chr.7: 44516904-44516904 on Build GRCh38
rs767294400 Chr.7: 44516692-44516692 on Build GRCh38
rs1025298045 Chr.7: 44517093-44517093 on Build GRCh38
rs753507585 Chr.7: 44516922-44516922 on Build GRCh38
rs750353030 Chr.7: 44516952-44516952 on Build GRCh38
rs954118253 Chr.7: 44516855-44516855 on Build GRCh38
rs764180139 Chr.7: 44516708-44516708 on Build GRCh38
rs781398788 Chr.7: 44516872-44516872 on Build GRCh38
rs766445118 Chr.7: 44516975-44516975 on Build GRCh38
rs757763983 Chr.7: 44516950-44516950 on Build GRCh38
rs866076761 Chr.7: 44516691-44516691 on Build GRCh38
rs564228764 Chr.7: 44516675-44516675 on Build GRCh38
rs767241752 Chr.7: 44516815-44516815 on Build GRCh38
rs111597821 Chr.7: 44516940-44516940 on Build GRCh38
rs116716160 Chr.7: 44516910-44516910 on Build GRCh38
rs972780034 Chr.7: 44516662-44516662 on Build GRCh38
rs556804455 Chr.7: 44517011-44517011 on Build GRCh38
rs931313148 Chr.7: 44516684-44516684 on Build GRCh38
rs965459192 Chr.7: 44516658-44516658 on Build GRCh38
rs535032460 Chr.7: 44516911-44516911 on Build GRCh38
rs757082934 Chr.7: 44516840-44516840 on Build GRCh38
rs115797477 Chr.7: 44516850-44516850 on Build GRCh38
rs763627653 Chr.7: 44516697-44516697 on Build GRCh38
rs758372877 Chr.7: 44516971-44516971 on Build GRCh38
rs184631951 Chr.7: 44517041-44517041 on Build GRCh38
rs922483180 Chr.7: 44516945-44516945 on Build GRCh38
rs770135270 Chr.7: 44516883-44516883 on Build GRCh38
rs765034232 Chr.7: 44516948-44516948 on Build GRCh38
rs771673560 Chr.7: 44516901-44516901 on Build GRCh38
rs758494260 Chr.7: 44516863-44516863 on Build GRCh38
rs777145768 Chr.7: 44516678-44516678 on Build GRCh38
rs773803950 Chr.7: 44516894-44516894 on Build GRCh38
rs1003595534 Chr.7: 44516789-44516789 on Build GRCh38
rs374640043 Chr.7: 44516687-44516687 on Build GRCh38
rs778913010 Chr.7: 44516849-44516849 on Build GRCh38
rs772394644 Chr.7: 44516674-44516674 on Build GRCh38
rs770891484 Chr.7: 44516663-44516663 on Build GRCh38
rs752476916 Chr.7: 44516703-44516703 on Build GRCh38
rs765777410 Chr.7: 44516681-44516681 on Build GRCh38
rs761698767 Chr.7: 44516926-44516926 on Build GRCh38
rs751696999 Chr.7: 44516980-44516980 on Build GRCh38
rs764543971 Chr.7: 44516676-44516676 on Build GRCh38
rs780157168 Chr.7: 44516869-44516869 on Build GRCh38
rs571171154 Chr.7: 44516871-44516871 on Build GRCh38
rs567755472 Chr.7: 44516988-44516988 on Build GRCh38
rs370523161 Chr.7: 44516762-44516762 on Build GRCh38
rs535397564 Chr.7: 44516989-44516989 on Build GRCh38
rs993751185 Chr.7: 44517091-44517091 on Build GRCh38
rs895973476 Chr.7: 44516768-44516768 on Build GRCh38
rs756854433 Chr.7: 44516921-44516921 on Build GRCh38
rs749337526 Chr.7: 44516654-44516654 on Build GRCh38
rs183439593 Chr.7: 44516774-44516774 on Build GRCh38
rs188329482 Chr.7: 44516886-44516886 on Build GRCh38
rs755138020 Chr.7: 44516983-44516983 on Build GRCh38
rs757282762 Chr.7: 44516709-44516709 on Build GRCh38
rs867876807 Chr.7: 44516865-44516865 on Build GRCh38
rs570099521 Chr.7: 44516838-44516838 on Build GRCh38
rs79744511 Chr.7: 44517079-44517079 on Build GRCh38

View Full Product Details