Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 57469628-57470134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF716
Entrez Gene ID
441234
Gene Name
zinc finger protein 716
Gene Aliases
-
UniGene
Hs.533121
Gene Chromosome Location
Chr.7: 57450177-57473559 on Build GRCh38
Transcripts
NM_001159279.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5825092 c.1383G>C p.E461D Chr.7: 57469844-57469844 on Build GRCh38
3923868 c.1381G>T p.E461* Chr.7: 57469842-57469842 on Build GRCh38
5407761 c.1306T>C p.Y436H Chr.7: 57469767-57469767 on Build GRCh38
7145069 c.1351C>G p.L451V Chr.7: 57469812-57469812 on Build GRCh38
3084568 c.1388C>A p.P463H Chr.7: 57469849-57469849 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200193410 Chr.7: 57469738-57469738 on Build GRCh38
rs782167999 Chr.7: 57469702-57469702 on Build GRCh38
rs782581861 Chr.7: 57469768-57469768 on Build GRCh38
rs10276219 Chr.7: 57469898-57469898 on Build GRCh38
rs192253742 Chr.7: 57469780-57469780 on Build GRCh38
rs782755792 Chr.7: 57469996-57469996 on Build GRCh38
rs781814325 Chr.7: 57469707-57469707 on Build GRCh38
rs782351042 Chr.7: 57469887-57469887 on Build GRCh38
rs782407347 Chr.7: 57469765-57469765 on Build GRCh38
rs781842699 Chr.7: 57469814-57469814 on Build GRCh38
rs782103405 Chr.7: 57469914-57469914 on Build GRCh38
rs782537773 Chr.7: 57469822-57469822 on Build GRCh38
rs782368687 Chr.7: 57469680-57469680 on Build GRCh38
rs782706178 Chr.7: 57469802-57469802 on Build GRCh38
rs782261122 Chr.7: 57469829-57469829 on Build GRCh38
rs782382436 Chr.7: 57469833-57469833 on Build GRCh38
rs782816521 Chr.7: 57469915-57469915 on Build GRCh38
rs186936913 Chr.7: 57469742-57469742 on Build GRCh38
rs781950128 Chr.7: 57469669-57469669 on Build GRCh38
rs782766103 Chr.7: 57469819-57469819 on Build GRCh38
rs13235676 Chr.7: 57469686-57469686 on Build GRCh38
rs782803759 Chr.7: 57469704-57469704 on Build GRCh38
rs551632870 Chr.7: 57469752-57469752 on Build GRCh38
rs781907719 Chr.7: 57469998-57469998 on Build GRCh38
rs73347709 Chr.7: 57470007-57470007 on Build GRCh38
rs782675600 Chr.7: 57469841-57469841 on Build GRCh38
rs782436451 Chr.7: 57469827-57469827 on Build GRCh38
rs538387754 Chr.7: 57469798-57469798 on Build GRCh38
rs782111532 Chr.7: 57469790-57469790 on Build GRCh38
rs782008578 Chr.7: 57469922-57469922 on Build GRCh38
rs200260740 Chr.7: 57469720-57469720 on Build GRCh38
rs782401889 Chr.7: 57469704-57469704 on Build GRCh38
rs535726423 Chr.7: 57469874-57469874 on Build GRCh38
rs782421238 Chr.7: 57469792-57469792 on Build GRCh38
rs782333381 Chr.7: 57469861-57469861 on Build GRCh38
rs137967121 Chr.7: 57469817-57469817 on Build GRCh38
rs534008198 Chr.7: 57469661-57469662 on Build GRCh38
rs782065934 Chr.7: 57469929-57469929 on Build GRCh38
rs782370269 Chr.7: 57469687-57469687 on Build GRCh38
rs782040291 Chr.7: 57470014-57470014 on Build GRCh38
rs781837359 Chr.7: 57469946-57469946 on Build GRCh38
rs572515468 Chr.7: 57469978-57469978 on Build GRCh38
rs184317577 Chr.7: 57469823-57469823 on Build GRCh38
rs782256356 Chr.7: 57469674-57469674 on Build GRCh38
rs201939109 Chr.7: 57470010-57470010 on Build GRCh38
rs782614855 Chr.7: 57469828-57469828 on Build GRCh38
rs782774321 Chr.7: 57469690-57469690 on Build GRCh38
rs782291425 Chr.7: 57469763-57469763 on Build GRCh38
rs781860162 Chr.7: 57469695-57469695 on Build GRCh38
rs557960333 Chr.7: 57470057-57470057 on Build GRCh38
rs782075340 Chr.7: 57469795-57469795 on Build GRCh38
rs559949160 Chr.7: 57470104-57470104 on Build GRCh38
rs782084946 Chr.7: 57469688-57469688 on Build GRCh38
rs782144211 Chr.7: 57469682-57469682 on Build GRCh38
rs781848267 Chr.7: 57469821-57469821 on Build GRCh38
rs782276048 Chr.7: 57469858-57469858 on Build GRCh38
rs782695242 Chr.7: 57469938-57469938 on Build GRCh38
rs782108582 Chr.7: 57469885-57469885 on Build GRCh38
rs782173657 Chr.7: 57469772-57469772 on Build GRCh38
rs782631225 Chr.7: 57469718-57469718 on Build GRCh38
rs782437065 Chr.7: 57469709-57469709 on Build GRCh38

View Full Product Details