Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 57470949-57471478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF716
Entrez Gene ID
441234
Gene Name
zinc finger protein 716
Gene Aliases
-
UniGene
Hs.533121
Gene Chromosome Location
Chr.7: 57450177-57473559 on Build GRCh38
Transcripts
NM_001159279.1

SNP Detail

SNP ID Chromosome Location
rs71527709 Chr.7: 57471025-57471028 on Build GRCh38
rs759164082 Chr.7: 57471323-57471323 on Build GRCh38
rs782296418 Chr.7: 57471051-57471051 on Build GRCh38
rs572779191 Chr.7: 57471274-57471274 on Build GRCh38
rs181098746 Chr.7: 57471365-57471365 on Build GRCh38
rs369706930 Chr.7: 57471272-57471272 on Build GRCh38
rs372986396 Chr.7: 57471178-57471178 on Build GRCh38
rs6977435 Chr.7: 57471027-57471027 on Build GRCh38
rs6950361 Chr.7: 57471080-57471080 on Build GRCh38
rs568045439 Chr.7: 57471112-57471112 on Build GRCh38
rs190237103 Chr.7: 57471242-57471242 on Build GRCh38
rs546319903 Chr.7: 57471013-57471013 on Build GRCh38
rs6965676 Chr.7: 57471314-57471314 on Build GRCh38
rs796259062 Chr.7: 57471428-57471429 on Build GRCh38
rs112358502 Chr.7: 57471026-57471026 on Build GRCh38
rs187015262 Chr.7: 57471432-57471432 on Build GRCh38
rs529007227 Chr.7: 57471310-57471310 on Build GRCh38
rs558035901 Chr.7: 57471214-57471214 on Build GRCh38
rs574755869 Chr.7: 57471416-57471416 on Build GRCh38
rs35530073 Chr.7: 57471370-57471371 on Build GRCh38
rs539707710 Chr.7: 57471154-57471154 on Build GRCh38
rs6965227 Chr.7: 57471025-57471025 on Build GRCh38
rs547769443 Chr.7: 57471121-57471121 on Build GRCh38
rs565778700 Chr.7: 57471126-57471126 on Build GRCh38
rs78192466 Chr.7: 57471428-57471428 on Build GRCh38
rs534458648 Chr.7: 57471431-57471431 on Build GRCh38
rs141563161 Chr.7: 57471264-57471264 on Build GRCh38
rs6965218 Chr.7: 57471009-57471009 on Build GRCh38
rs5884478 Chr.7: 57471415-57471415 on Build GRCh38

View Full Product Details