Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 72926710-72927210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POM121
Entrez Gene ID
9883
Gene Name
POM121 transmembrane nucleoporin
Gene Aliases
P145, POM121A
UniGene
Hs.655217
Gene Chromosome Location
Chr.7: 72879335-72951440 on Build GRCh38
Transcripts
NM_001257190.2 NM_172020.4 XM_005250726.3 XM_005250727.3 XM_006716196.3 XM_011516730.1 XM_017012853.1 XM_017012854.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5407837 c.150C>T p.A50A Chr.7: 72926886-72926886 on Build GRCh38
5884584 c.1019G>C p.R340T Chr.7: 72926960-72926960 on Build GRCh38
5648086 c.101C>T p.P34L Chr.7: 72926837-72926837 on Build GRCh38
5919411 c.997C>T p.L333F Chr.7: 72926938-72926938 on Build GRCh38
5407839 c.150C>T p.A50A Chr.7: 72926886-72926886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782460426 Chr.7: 72927139-72927139 on Build GRCh38
rs782756414 Chr.7: 72926998-72926998 on Build GRCh38
rs782611651 Chr.7: 72926975-72926976 on Build GRCh38
rs781843820 Chr.7: 72927144-72927144 on Build GRCh38
rs781866865 Chr.7: 72926943-72926943 on Build GRCh38
rs551118607 Chr.7: 72926860-72926860 on Build GRCh38
rs782144951 Chr.7: 72926771-72926771 on Build GRCh38
rs117799166 Chr.7: 72927039-72927039 on Build GRCh38
rs549135243 Chr.7: 72927007-72927007 on Build GRCh38
rs781895105 Chr.7: 72927130-72927130 on Build GRCh38
rs563215467 Chr.7: 72926847-72926847 on Build GRCh38
rs183582090 Chr.7: 72927156-72927156 on Build GRCh38
rs782796768 Chr.7: 72926773-72926773 on Build GRCh38
rs537709798 Chr.7: 72927037-72927037 on Build GRCh38
rs782118735 Chr.7: 72926985-72926985 on Build GRCh38
rs562165164 Chr.7: 72926827-72926827 on Build GRCh38
rs782750987 Chr.7: 72926788-72926788 on Build GRCh38
rs202119231 Chr.7: 72926997-72926997 on Build GRCh38
rs782760932 Chr.7: 72926887-72926887 on Build GRCh38
rs782590787 Chr.7: 72926798-72926798 on Build GRCh38
rs782421689 Chr.7: 72926972-72926972 on Build GRCh38
rs201332972 Chr.7: 72926837-72926837 on Build GRCh38
rs781962306 Chr.7: 72926780-72926780 on Build GRCh38
rs573288391 Chr.7: 72926764-72926764 on Build GRCh38
rs201082459 Chr.7: 72926937-72926937 on Build GRCh38
rs571232633 Chr.7: 72927049-72927049 on Build GRCh38
rs781789475 Chr.7: 72926889-72926889 on Build GRCh38
rs782520501 Chr.7: 72926885-72926885 on Build GRCh38
rs782714906 Chr.7: 72926936-72926936 on Build GRCh38
rs782577901 Chr.7: 72926834-72926834 on Build GRCh38
rs782094540 Chr.7: 72926825-72926825 on Build GRCh38
rs540372795 Chr.7: 72927185-72927185 on Build GRCh38
rs781848321 Chr.7: 72926781-72926781 on Build GRCh38
rs782464120 Chr.7: 72926897-72926897 on Build GRCh38
rs782490608 Chr.7: 72927082-72927082 on Build GRCh38
rs781894774 Chr.7: 72926799-72926799 on Build GRCh38
rs567354147 Chr.7: 72927015-72927015 on Build GRCh38
rs782318264 Chr.7: 72927058-72927058 on Build GRCh38
rs140010950 Chr.7: 72926875-72926875 on Build GRCh38
rs782442506 Chr.7: 72926792-72926792 on Build GRCh38
rs781924385 Chr.7: 72926944-72926944 on Build GRCh38
rs140853733 Chr.7: 72926952-72926952 on Build GRCh38
rs199996427 Chr.7: 72926832-72926832 on Build GRCh38
rs782575037 Chr.7: 72927142-72927142 on Build GRCh38
rs782293431 Chr.7: 72926914-72926914 on Build GRCh38
rs782820514 Chr.7: 72927103-72927103 on Build GRCh38
rs782303819 Chr.7: 72926999-72926999 on Build GRCh38
rs781919362 Chr.7: 72926775-72926775 on Build GRCh38
rs368167847 Chr.7: 72926990-72926990 on Build GRCh38
rs782038455 Chr.7: 72926983-72926983 on Build GRCh38
rs782554582 Chr.7: 72926819-72926819 on Build GRCh38
rs782654317 Chr.7: 72926901-72926901 on Build GRCh38
rs782135911 Chr.7: 72926782-72926782 on Build GRCh38
rs539429883 Chr.7: 72927073-72927073 on Build GRCh38
rs151261529 Chr.7: 72926890-72926890 on Build GRCh38
rs782242056 Chr.7: 72926848-72926848 on Build GRCh38
rs782404585 Chr.7: 72926845-72926845 on Build GRCh38
rs377167035 Chr.7: 72926991-72926991 on Build GRCh38
rs540785922 Chr.7: 72926770-72926770 on Build GRCh38
rs781842778 Chr.7: 72926790-72926790 on Build GRCh38
rs375816974 Chr.7: 72926871-72926871 on Build GRCh38
rs532954210 Chr.7: 72926852-72926852 on Build GRCh38
rs199869678 Chr.7: 72926996-72926996 on Build GRCh38
rs782388409 Chr.7: 72926849-72926849 on Build GRCh38
rs749366550 Chr.7: 72927000-72927000 on Build GRCh38
rs187699228 Chr.7: 72927177-72927177 on Build GRCh38
rs116164279 Chr.7: 72926882-72926882 on Build GRCh38
rs782780223 Chr.7: 72926986-72926986 on Build GRCh38
rs781958495 Chr.7: 72926850-72926850 on Build GRCh38

View Full Product Details