Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73442638-73443136 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAZ1B
Entrez Gene ID
9031
Gene Name
bromodomain adjacent to zinc finger domain 1B
Gene Aliases
WBSCR10, WBSCR9, WSTF
UniGene
Hs.743372
Gene Chromosome Location
Chr.7: 73440398-73522298 on Build GRCh38
Transcripts
NM_032408.3 XM_017012773.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5813040 c.4045G>T p.E1349* Chr.7: 73442774-73442774 on Build GRCh38
4665209 c.4066delG p.V1356fs*1 Chr.7: 73442753-73442753 on Build GRCh38
6387157 c.4093A>G p.R1365G Chr.7: 73442726-73442726 on Build GRCh38
6504623 c.4000A>G p.T1334A Chr.7: 73442819-73442819 on Build GRCh38
6434891 c.4030G>A p.E1344K Chr.7: 73442789-73442789 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782721967 Chr.7: 73442710-73442710 on Build GRCh38
rs140686642 Chr.7: 73442818-73442818 on Build GRCh38
rs1178977 Chr.7: 73442719-73442719 on Build GRCh38
rs563917825 Chr.7: 73442986-73442986 on Build GRCh38
rs781892292 Chr.7: 73442809-73442809 on Build GRCh38
rs782294803 Chr.7: 73442793-73442793 on Build GRCh38
rs574615363 Chr.7: 73442925-73442925 on Build GRCh38
rs781803573 Chr.7: 73442749-73442749 on Build GRCh38
rs781801612 Chr.7: 73442727-73442727 on Build GRCh38
rs188367547 Chr.7: 73443081-73443081 on Build GRCh38
rs150115317 Chr.7: 73442800-73442800 on Build GRCh38
rs782560377 Chr.7: 73442830-73442830 on Build GRCh38
rs782697900 Chr.7: 73442711-73442711 on Build GRCh38
rs375363923 Chr.7: 73442686-73442686 on Build GRCh38
rs138650548 Chr.7: 73442803-73442803 on Build GRCh38
rs782326072 Chr.7: 73442786-73442786 on Build GRCh38
rs781848597 Chr.7: 73442727-73442727 on Build GRCh38
rs201581330 Chr.7: 73442736-73442736 on Build GRCh38
rs528054867 Chr.7: 73442991-73442991 on Build GRCh38
rs552883348 Chr.7: 73442974-73442974 on Build GRCh38
rs782465081 Chr.7: 73442820-73442820 on Build GRCh38
rs782783122 Chr.7: 73442879-73442879 on Build GRCh38
rs577212970 Chr.7: 73442843-73442843 on Build GRCh38
rs180802912 Chr.7: 73442934-73442934 on Build GRCh38
rs1005480068 Chr.7: 73442951-73442951 on Build GRCh38
rs782665302 Chr.7: 73442831-73442831 on Build GRCh38
rs782094823 Chr.7: 73442703-73442704 on Build GRCh38
rs916090762 Chr.7: 73443075-73443075 on Build GRCh38
rs782431200 Chr.7: 73442840-73442841 on Build GRCh38
rs140665563 Chr.7: 73442754-73442754 on Build GRCh38
rs781912972 Chr.7: 73442848-73442848 on Build GRCh38
rs998907367 Chr.7: 73442670-73442670 on Build GRCh38
rs782808446 Chr.7: 73442808-73442808 on Build GRCh38
rs534706747 Chr.7: 73442672-73442672 on Build GRCh38
rs782528958 Chr.7: 73442812-73442812 on Build GRCh38
rs1043005763 Chr.7: 73442846-73442846 on Build GRCh38
rs563943059 Chr.7: 73442945-73442945 on Build GRCh38
rs782150287 Chr.7: 73442851-73442851 on Build GRCh38
rs782210877 Chr.7: 73442782-73442782 on Build GRCh38
rs782272920 Chr.7: 73442760-73442760 on Build GRCh38
rs74803013 Chr.7: 73442887-73442887 on Build GRCh38
rs531090143 Chr.7: 73442959-73442959 on Build GRCh38
rs200219180 Chr.7: 73442802-73442802 on Build GRCh38
rs782262449 Chr.7: 73442834-73442834 on Build GRCh38
rs781904943 Chr.7: 73442716-73442716 on Build GRCh38
rs782232040 Chr.7: 73443022-73443022 on Build GRCh38
rs983764562 Chr.7: 73442748-73442748 on Build GRCh38
rs960465784 Chr.7: 73443031-73443031 on Build GRCh38
rs535596302 Chr.7: 73442730-73442730 on Build GRCh38
rs781820703 Chr.7: 73442690-73442690 on Build GRCh38
rs782340112 Chr.7: 73442674-73442674 on Build GRCh38
rs782519319 Chr.7: 73442763-73442763 on Build GRCh38
rs782301902 Chr.7: 73442685-73442685 on Build GRCh38
rs782113439 Chr.7: 73442694-73442694 on Build GRCh38
rs140578726 Chr.7: 73442842-73442844 on Build GRCh38
rs570458251 Chr.7: 73442657-73442657 on Build GRCh38
rs782359795 Chr.7: 73442703-73442703 on Build GRCh38
rs541118115 Chr.7: 73442849-73442849 on Build GRCh38
rs782605102 Chr.7: 73442739-73442739 on Build GRCh38
rs782768713 Chr.7: 73442817-73442817 on Build GRCh38
rs782109581 Chr.7: 73442801-73442801 on Build GRCh38
rs782610530 Chr.7: 73442772-73442772 on Build GRCh38
rs553146938 Chr.7: 73442718-73442718 on Build GRCh38
rs183658382 Chr.7: 73443039-73443039 on Build GRCh38
rs782575589 Chr.7: 73442678-73442678 on Build GRCh38
rs781925912 Chr.7: 73442792-73442792 on Build GRCh38
rs924609090 Chr.7: 73443111-73443111 on Build GRCh38
rs1028595687 Chr.7: 73442944-73442944 on Build GRCh38
rs145782828 Chr.7: 73442826-73442826 on Build GRCh38
rs782559078 Chr.7: 73442753-73442753 on Build GRCh38
rs749567870 Chr.7: 73442990-73442990 on Build GRCh38
rs782403273 Chr.7: 73442799-73442799 on Build GRCh38
rs535557853 Chr.7: 73443077-73443077 on Build GRCh38
rs782061342 Chr.7: 73442806-73442806 on Build GRCh38
rs558656491 Chr.7: 73442825-73442825 on Build GRCh38
rs916187939 Chr.7: 73442836-73442836 on Build GRCh38

View Full Product Details