Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99705347-99705858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP3A7
Entrez Gene ID
1551
Gene Name
cytochrome P450 family 3 subfamily A member 7
Gene Aliases
CP37, CYPIIIA7, P-450(HFL33), P-450111A7, P450-HFLA
UniGene
Hs.111944
Gene Chromosome Location
Chr.7: 99705037-99735200 on Build GRCh38
Transcripts
NM_000765.4 NM_001256497.2

Gene Symbol
CYP3A7-CYP3A51P
Entrez Gene ID
100861540
Gene Name
CYP3A7-CYP3A51P readthrough
Gene Aliases
CYP3A7-3AP1, CYP3A7-CYP3AP1, CYP3A7.1L
UniGene
Gene Chromosome Location
Chr.7: 99684679-99735200 on Build GRCh38
Transcripts
NM_000765.4 NM_001256497.2

Gene Symbol
ZSCAN25
Entrez Gene ID
221785
Gene Name
zinc finger and SCAN domain containing 25
Gene Aliases
ZNF498
UniGene
Hs.446297
Gene Chromosome Location
Chr.7: 99616600-99738689 on Build GRCh38
Transcripts
NM_000765.4 NM_001256497.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6639315 c.1498G>A p.V500I Chr.7: 99705514-99705514 on Build GRCh38
5901480 c.1439G>A p.G480E Chr.7: 99705573-99705573 on Build GRCh38
1202877 c.1433G>A p.R478H Chr.7: 99705579-99705579 on Build GRCh38
5016190 c.1499_1500insT p.S501fs*>4 Chr.7: 99705512-99705513 on Build GRCh38
160308 c.1511G>A p.*504* Chr.7: 99705501-99705501 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1046248841 Chr.7: 99705664-99705665 on Build GRCh38
rs573920585 Chr.7: 99705828-99705828 on Build GRCh38
rs201591102 Chr.7: 99705529-99705529 on Build GRCh38
rs376178086 Chr.7: 99705826-99705826 on Build GRCh38
rs200727885 Chr.7: 99705542-99705542 on Build GRCh38
rs749574339 Chr.7: 99705579-99705579 on Build GRCh38
rs144829218 Chr.7: 99705680-99705680 on Build GRCh38
rs10211 Chr.7: 99705371-99705371 on Build GRCh38
rs542238455 Chr.7: 99705764-99705764 on Build GRCh38
rs111357960 Chr.7: 99705758-99705758 on Build GRCh38
rs780328060 Chr.7: 99705754-99705754 on Build GRCh38
rs554867169 Chr.7: 99705786-99705786 on Build GRCh38
rs779304133 Chr.7: 99705822-99705822 on Build GRCh38
rs536118849 Chr.7: 99705652-99705652 on Build GRCh38
rs764133850 Chr.7: 99705514-99705514 on Build GRCh38
rs368605886 Chr.7: 99705541-99705541 on Build GRCh38
rs762252052 Chr.7: 99705550-99705550 on Build GRCh38
rs201727053 Chr.7: 99705601-99705601 on Build GRCh38
rs757100968 Chr.7: 99705520-99705520 on Build GRCh38
rs759152252 Chr.7: 99705484-99705484 on Build GRCh38
rs199645882 Chr.7: 99705524-99705524 on Build GRCh38
rs764078210 Chr.7: 99705611-99705611 on Build GRCh38
rs750249078 Chr.7: 99705643-99705643 on Build GRCh38
rs12360 Chr.7: 99705471-99705471 on Build GRCh38
rs144194706 Chr.7: 99705796-99705796 on Build GRCh38
rs200849307 Chr.7: 99705599-99705599 on Build GRCh38
rs770349622 Chr.7: 99705475-99705475 on Build GRCh38
rs553783663 Chr.7: 99705726-99705726 on Build GRCh38
rs113539362 Chr.7: 99705515-99705515 on Build GRCh38
rs922765752 Chr.7: 99705558-99705558 on Build GRCh38
rs770226567 Chr.7: 99705547-99705547 on Build GRCh38
rs1046965421 Chr.7: 99705735-99705735 on Build GRCh38
rs774084637 Chr.7: 99705483-99705483 on Build GRCh38
rs1012085027 Chr.7: 99705372-99705372 on Build GRCh38
rs1026003801 Chr.7: 99705424-99705424 on Build GRCh38
rs773789511 Chr.7: 99705575-99705575 on Build GRCh38
rs367570757 Chr.7: 99705634-99705634 on Build GRCh38
rs915917299 Chr.7: 99705623-99705623 on Build GRCh38
rs768758724 Chr.7: 99705457-99705457 on Build GRCh38
rs4646468 Chr.7: 99705378-99705378 on Build GRCh38
rs1003813155 Chr.7: 99705722-99705722 on Build GRCh38
rs536656450 Chr.7: 99705528-99705528 on Build GRCh38
rs4646467 Chr.7: 99705665-99705665 on Build GRCh38
rs370160306 Chr.7: 99705426-99705426 on Build GRCh38
rs763388048 Chr.7: 99705480-99705480 on Build GRCh38
rs376299497 Chr.7: 99705481-99705481 on Build GRCh38
rs756119340 Chr.7: 99705508-99705508 on Build GRCh38
rs556304793 Chr.7: 99705637-99705637 on Build GRCh38
rs775076106 Chr.7: 99705607-99705607 on Build GRCh38
rs1005425547 Chr.7: 99705746-99705746 on Build GRCh38
rs770674379 Chr.7: 99705567-99705567 on Build GRCh38
rs370438739 Chr.7: 99705523-99705523 on Build GRCh38
rs955747041 Chr.7: 99705578-99705578 on Build GRCh38
rs199882035 Chr.7: 99705574-99705574 on Build GRCh38
rs182605954 Chr.7: 99705737-99705737 on Build GRCh38
rs1003436389 Chr.7: 99705416-99705416 on Build GRCh38
rs906367448 Chr.7: 99705395-99705395 on Build GRCh38
rs140346984 Chr.7: 99705765-99705765 on Build GRCh38
rs755214605 Chr.7: 99705808-99705808 on Build GRCh38
rs773380240 Chr.7: 99705479-99705479 on Build GRCh38
rs10263682 Chr.7: 99705610-99705610 on Build GRCh38
rs753848334 Chr.7: 99705618-99705618 on Build GRCh38
rs752315134 Chr.7: 99705504-99705504 on Build GRCh38
rs532834834 Chr.7: 99705458-99705458 on Build GRCh38
rs748227868 Chr.7: 99705533-99705533 on Build GRCh38
rs751002782 Chr.7: 99705569-99705569 on Build GRCh38
rs201845912 Chr.7: 99705590-99705590 on Build GRCh38
rs771294954 Chr.7: 99705580-99705580 on Build GRCh38

View Full Product Details