Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 157841544-157842048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD5L
Entrez Gene ID
922
Gene Name
CD5 molecule like
Gene Aliases
AIM, API6, CT-2, PRO229, SP-ALPHA, Spalpha, hAIM
UniGene
Hs.134035
Gene Chromosome Location
Chr.1: 157827071-157841844 on Build GRCh38
Transcripts
NM_005894.2 XM_005245602.1 XM_017002806.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1247715 c.19T>G p.L7V Chr.1: 157841683-157841683 on Build GRCh38
4633356 c.11T>G p.L4R Chr.1: 157841691-157841691 on Build GRCh38
1985561 c.13T>C p.F5L Chr.1: 157841689-157841689 on Build GRCh38
292852 c.12A>T p.L4L Chr.1: 157841690-157841690 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1014308241 Chr.1: 157841898-157841898 on Build GRCh38
rs576976589 Chr.1: 157841722-157841722 on Build GRCh38
rs758299958 Chr.1: 157841707-157841707 on Build GRCh38
rs760365605 Chr.1: 157841687-157841687 on Build GRCh38
rs775872188 Chr.1: 157841739-157841739 on Build GRCh38
rs1017165640 Chr.1: 157841991-157841991 on Build GRCh38
rs925193821 Chr.1: 157842015-157842015 on Build GRCh38
rs1056288051 Chr.1: 157841639-157841639 on Build GRCh38
rs546259157 Chr.1: 157841753-157841753 on Build GRCh38
rs746320706 Chr.1: 157841729-157841729 on Build GRCh38
rs373635325 Chr.1: 157841969-157841969 on Build GRCh38
rs569476905 Chr.1: 157841580-157841580 on Build GRCh38
rs781223824 Chr.1: 157841660-157841660 on Build GRCh38
rs544440054 Chr.1: 157841761-157841761 on Build GRCh38
rs928704723 Chr.1: 157841605-157841605 on Build GRCh38
rs371204785 Chr.1: 157841601-157841601 on Build GRCh38
rs867656573 Chr.1: 157841993-157841993 on Build GRCh38
rs904468652 Chr.1: 157841860-157841860 on Build GRCh38
rs1027201558 Chr.1: 157841860-157841861 on Build GRCh38
rs762576824 Chr.1: 157841645-157841645 on Build GRCh38
rs1024444939 Chr.1: 157841665-157841665 on Build GRCh38
rs2281870 Chr.1: 157841795-157841795 on Build GRCh38
rs777968122 Chr.1: 157841806-157841806 on Build GRCh38
rs143300465 Chr.1: 157841939-157841939 on Build GRCh38
rs763705151 Chr.1: 157841690-157841690 on Build GRCh38
rs945889305 Chr.1: 157841829-157841829 on Build GRCh38
rs779503779 Chr.1: 157841724-157841724 on Build GRCh38
rs775142026 Chr.1: 157841667-157841667 on Build GRCh38
rs903519264 Chr.1: 157841754-157841754 on Build GRCh38
rs894488239 Chr.1: 157841872-157841872 on Build GRCh38
rs960093759 Chr.1: 157841877-157841877 on Build GRCh38
rs887242729 Chr.1: 157841935-157841935 on Build GRCh38
rs761214538 Chr.1: 157841698-157841698 on Build GRCh38
rs781571036 Chr.1: 157841626-157841626 on Build GRCh38
rs747212862 Chr.1: 157841708-157841708 on Build GRCh38
rs542100868 Chr.1: 157841821-157841821 on Build GRCh38
rs147298497 Chr.1: 157841793-157841793 on Build GRCh38
rs1036203565 Chr.1: 157841870-157841870 on Build GRCh38
rs558693417 Chr.1: 157841718-157841718 on Build GRCh38
rs772541717 Chr.1: 157841737-157841737 on Build GRCh38
rs751994649 Chr.1: 157841699-157841699 on Build GRCh38
rs377466969 Chr.1: 157841595-157841595 on Build GRCh38
rs1018381996 Chr.1: 157841826-157841826 on Build GRCh38
rs1056231263 Chr.1: 157841862-157841862 on Build GRCh38
rs776271438 Chr.1: 157841694-157841694 on Build GRCh38
rs371637908 Chr.1: 157841642-157841642 on Build GRCh38
rs750148106 Chr.1: 157841851-157841851 on Build GRCh38
rs1043450721 Chr.1: 157841723-157841723 on Build GRCh38
rs947829378 Chr.1: 157842000-157842000 on Build GRCh38
rs770016758 Chr.1: 157841629-157841629 on Build GRCh38
rs746232893 Chr.1: 157841652-157841652 on Build GRCh38
rs369016557 Chr.1: 157841712-157841712 on Build GRCh38
rs879380469 Chr.1: 157841924-157841924 on Build GRCh38
rs2260040 Chr.1: 157841602-157841602 on Build GRCh38
rs772383437 Chr.1: 157841655-157841655 on Build GRCh38
rs931769347 Chr.1: 157841800-157841800 on Build GRCh38
rs35278075 Chr.1: 157841907-157841908 on Build GRCh38
rs540186540 Chr.1: 157841710-157841710 on Build GRCh38
rs143998261 Chr.1: 157841592-157841592 on Build GRCh38
rs368154340 Chr.1: 157841781-157841781 on Build GRCh38
rs571424618 Chr.1: 157842001-157842001 on Build GRCh38
rs995714312 Chr.1: 157841858-157841858 on Build GRCh38
rs561175539 Chr.1: 157841861-157841861 on Build GRCh38
rs750263320 Chr.1: 157841701-157841701 on Build GRCh38
rs868285054 Chr.1: 157841743-157841743 on Build GRCh38
rs777960935 Chr.1: 157841635-157841635 on Build GRCh38
rs774709929 Chr.1: 157841646-157841646 on Build GRCh38
rs139393365 Chr.1: 157841927-157841927 on Build GRCh38
rs963062487 Chr.1: 157841784-157841784 on Build GRCh38
rs748341076 Chr.1: 157841628-157841628 on Build GRCh38
rs751381137 Chr.1: 157841709-157841709 on Build GRCh38
rs868295471 Chr.1: 157841677-157841677 on Build GRCh38
rs368169849 Chr.1: 157841638-157841638 on Build GRCh38
rs867460785 Chr.1: 157841616-157841616 on Build GRCh38
rs1014422682 Chr.1: 157841664-157841664 on Build GRCh38
rs755616570 Chr.1: 157841767-157841767 on Build GRCh38
rs911687848 Chr.1: 157841823-157841823 on Build GRCh38
rs373594326 Chr.1: 157841820-157841820 on Build GRCh38

View Full Product Details